Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43824798-43825306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG5
Entrez Gene ID
64240
Gene Name
ATP binding cassette subfamily G member 5
Gene Aliases
STSL
UniGene
Hs.132992
Gene Chromosome Location
Chr.2: 43806154-43838840 on Build GRCh38
Transcripts
NM_022436.2 XM_005264480.3 XM_006712073.3 XM_006712074.3 XM_011533024.2 XM_011533025.2 XM_011533026.2 XM_011533027.2 XM_011533028.2 XM_005264364.4 XM_005264365.4

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
UniGene
Hs.371597
Gene Chromosome Location
Chr.2: 43774039-43828492 on Build GRCh38
Transcripts
NM_022436.2 XM_005264480.3 XM_006712073.3 XM_006712074.3 XM_011533024.2 XM_011533025.2 XM_011533026.2 XM_011533027.2 XM_011533028.2 XM_005264364.4 XM_005264365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3581743 c.839T>G p.M280R Chr.2: 43824954-43824954 on Build GRCh38
4433575 c.805G>A p.G269R Chr.2: 43824988-43824988 on Build GRCh38
3910354 c.804C>T p.F268F Chr.2: 43824989-43824989 on Build GRCh38
4510798 c.849C>T p.F283F Chr.2: 43824944-43824944 on Build GRCh38
5040064 c.868C>T p.P290S Chr.2: 43824925-43824925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766905376 Chr.2: 43824866-43824866 on Build GRCh38
rs769810440 Chr.2: 43825195-43825195 on Build GRCh38
rs530120175 Chr.2: 43824932-43824932 on Build GRCh38
rs953640844 Chr.2: 43825224-43825224 on Build GRCh38
rs771137662 Chr.2: 43824913-43824913 on Build GRCh38
rs772098158 Chr.2: 43824895-43824895 on Build GRCh38
rs773160445 Chr.2: 43825261-43825261 on Build GRCh38
rs530003702 Chr.2: 43825267-43825267 on Build GRCh38
rs753502791 Chr.2: 43824888-43824888 on Build GRCh38
rs775753291 Chr.2: 43824943-43824944 on Build GRCh38
rs79964831 Chr.2: 43825277-43825277 on Build GRCh38
rs936742300 Chr.2: 43824957-43824957 on Build GRCh38
rs554183818 Chr.2: 43824989-43824989 on Build GRCh38
rs370806483 Chr.2: 43824944-43824944 on Build GRCh38
rs937261413 Chr.2: 43825269-43825269 on Build GRCh38
rs368001054 Chr.2: 43825035-43825035 on Build GRCh38
rs765931771 Chr.2: 43824938-43824938 on Build GRCh38
rs758983572 Chr.2: 43825066-43825066 on Build GRCh38
rs184893733 Chr.2: 43825077-43825077 on Build GRCh38
rs975612815 Chr.2: 43824939-43824939 on Build GRCh38
rs773384981 Chr.2: 43824841-43824841 on Build GRCh38
rs779617392 Chr.2: 43824886-43824886 on Build GRCh38
rs201469377 Chr.2: 43825025-43825025 on Build GRCh38
rs200772212 Chr.2: 43824940-43824940 on Build GRCh38
rs759071636 Chr.2: 43825019-43825019 on Build GRCh38
rs751534445 Chr.2: 43825029-43825029 on Build GRCh38
rs776932087 Chr.2: 43824916-43824916 on Build GRCh38
rs566982836 Chr.2: 43824960-43824960 on Build GRCh38
rs190380979 Chr.2: 43825000-43825000 on Build GRCh38
rs753127265 Chr.2: 43824860-43824860 on Build GRCh38
rs377520529 Chr.2: 43824900-43824900 on Build GRCh38
rs998115224 Chr.2: 43825212-43825212 on Build GRCh38
rs949487920 Chr.2: 43825196-43825196 on Build GRCh38
rs112926649 Chr.2: 43824998-43824998 on Build GRCh38
rs770103215 Chr.2: 43824990-43824990 on Build GRCh38
rs925450199 Chr.2: 43825163-43825163 on Build GRCh38
rs904965047 Chr.2: 43825079-43825079 on Build GRCh38
rs192689305 Chr.2: 43824849-43824849 on Build GRCh38
rs757298700 Chr.2: 43825033-43825033 on Build GRCh38
rs879863995 Chr.2: 43825181-43825181 on Build GRCh38
rs769061566 Chr.2: 43825055-43825055 on Build GRCh38
rs774909356 Chr.2: 43825005-43825005 on Build GRCh38
rs569546465 Chr.2: 43825174-43825174 on Build GRCh38
rs748175962 Chr.2: 43824838-43824838 on Build GRCh38
rs759892269 Chr.2: 43824927-43824927 on Build GRCh38
rs975624159 Chr.2: 43825114-43825114 on Build GRCh38
rs768917242 Chr.2: 43825002-43825002 on Build GRCh38
rs757395824 Chr.2: 43824983-43824983 on Build GRCh38
rs778837844 Chr.2: 43824892-43824892 on Build GRCh38
rs765847597 Chr.2: 43825013-43825013 on Build GRCh38
rs1019691052 Chr.2: 43825106-43825106 on Build GRCh38
rs368800131 Chr.2: 43824966-43824966 on Build GRCh38
rs937763067 Chr.2: 43825097-43825097 on Build GRCh38
rs949163056 Chr.2: 43824903-43824903 on Build GRCh38
rs192604398 Chr.2: 43825052-43825052 on Build GRCh38
rs748009304 Chr.2: 43824893-43824893 on Build GRCh38
rs964329477 Chr.2: 43825111-43825111 on Build GRCh38
rs879230005 Chr.2: 43824942-43824942 on Build GRCh38
rs545176980 Chr.2: 43825010-43825010 on Build GRCh38
rs200416765 Chr.2: 43825036-43825036 on Build GRCh38
rs4148186 Chr.2: 43825064-43825064 on Build GRCh38
rs1007916481 Chr.2: 43825088-43825088 on Build GRCh38
rs539178581 Chr.2: 43824965-43824965 on Build GRCh38
rs78070897 Chr.2: 43825008-43825008 on Build GRCh38
rs747362226 Chr.2: 43825107-43825107 on Build GRCh38
rs201754827 Chr.2: 43824868-43824868 on Build GRCh38
rs146842360 Chr.2: 43825047-43825047 on Build GRCh38
rs780071302 Chr.2: 43824921-43824921 on Build GRCh38
rs749997142 Chr.2: 43824867-43824867 on Build GRCh38
rs772044301 Chr.2: 43824840-43824840 on Build GRCh38
rs139264483 Chr.2: 43824931-43824931 on Build GRCh38
rs377465364 Chr.2: 43824837-43824837 on Build GRCh38
rs114549158 Chr.2: 43825028-43825028 on Build GRCh38
rs201015371 Chr.2: 43825027-43825027 on Build GRCh38
rs749623331 Chr.2: 43825001-43825001 on Build GRCh38
rs755654485 Chr.2: 43824872-43824872 on Build GRCh38
rs759910660 Chr.2: 43824855-43824855 on Build GRCh38
rs373182673 Chr.2: 43824959-43824959 on Build GRCh38
rs758151854 Chr.2: 43825026-43825026 on Build GRCh38
rs376686791 Chr.2: 43825068-43825068 on Build GRCh38
rs528693183 Chr.2: 43825018-43825018 on Build GRCh38
rs552803459 Chr.2: 43824988-43824988 on Build GRCh38
rs375472612 Chr.2: 43824856-43824856 on Build GRCh38
rs986248628 Chr.2: 43825231-43825231 on Build GRCh38
rs369826575 Chr.2: 43824891-43824891 on Build GRCh38
rs958165628 Chr.2: 43825170-43825170 on Build GRCh38
rs184454421 Chr.2: 43824854-43824854 on Build GRCh38
rs111587587 Chr.2: 43825063-43825063 on Build GRCh38
rs1030484294 Chr.2: 43825213-43825213 on Build GRCh38
rs764907076 Chr.2: 43825021-43825021 on Build GRCh38
rs562035968 Chr.2: 43825164-43825164 on Build GRCh38
rs760788948 Chr.2: 43824845-43824845 on Build GRCh38
rs752336493 Chr.2: 43824982-43824982 on Build GRCh38
rs1020454866 Chr.2: 43825006-43825006 on Build GRCh38
rs763582454 Chr.2: 43824864-43824864 on Build GRCh38
rs577788640 Chr.2: 43825007-43825007 on Build GRCh38
rs893491597 Chr.2: 43825154-43825154 on Build GRCh38
rs141033260 Chr.2: 43824934-43824934 on Build GRCh38
rs951139635 Chr.2: 43825080-43825080 on Build GRCh38
rs373237261 Chr.2: 43824896-43824896 on Build GRCh38

View Full Product Details