Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 56183983-56184470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85A
Entrez Gene ID
114800
Gene Name
coiled-coil domain containing 85A
Gene Aliases
-
UniGene
Hs.117136
Gene Chromosome Location
Chr.2: 56184123-56386174 on Build GRCh38
Transcripts
NM_001080433.1 NR_125368.1 XM_017005442.1

Gene Symbol
LOC100129434
Entrez Gene ID
100129434
Gene Name
uncharacterized LOC100129434
Gene Aliases
-
UniGene
Hs.580330
Gene Chromosome Location
Chr.2: 56173534-56185770 on Build GRCh38
Transcripts
NM_001080433.1 NR_125368.1 XM_017005442.1

Gene Symbol
LOC107985795
Entrez Gene ID
107985795
Gene Name
uncharacterized LOC107985795
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.2: 56183193-56184133 on Build GRCh38
Transcripts
NM_001080433.1 NR_125368.1 XM_017005442.1

SNP Detail

SNP ID Chromosome Location
rs1054340335 Chr.2: 56184288-56184288 on Build GRCh38
rs954285053 Chr.2: 56184366-56184366 on Build GRCh38
rs539935384 Chr.2: 56184256-56184256 on Build GRCh38
rs892935923 Chr.2: 56184051-56184051 on Build GRCh38
rs1048808403 Chr.2: 56184241-56184241 on Build GRCh38
rs182343995 Chr.2: 56184282-56184282 on Build GRCh38
rs1011004147 Chr.2: 56184055-56184055 on Build GRCh38
rs1032405638 Chr.2: 56184365-56184365 on Build GRCh38
rs931504798 Chr.2: 56184229-56184229 on Build GRCh38
rs1028421415 Chr.2: 56184409-56184409 on Build GRCh38
rs547018220 Chr.2: 56184268-56184268 on Build GRCh38
rs972973688 Chr.2: 56184401-56184401 on Build GRCh38
rs932631383 Chr.2: 56184117-56184117 on Build GRCh38
rs1017172341 Chr.2: 56184382-56184382 on Build GRCh38
rs142674240 Chr.2: 56184031-56184031 on Build GRCh38
rs951673879 Chr.2: 56184186-56184186 on Build GRCh38
rs867889942 Chr.2: 56184168-56184168 on Build GRCh38
rs565033682 Chr.2: 56184104-56184126 on Build GRCh38
rs931429467 Chr.2: 56184415-56184415 on Build GRCh38
rs934356131 Chr.2: 56184434-56184434 on Build GRCh38
rs979482904 Chr.2: 56184089-56184089 on Build GRCh38
rs955171488 Chr.2: 56184411-56184411 on Build GRCh38
rs979239300 Chr.2: 56184362-56184362 on Build GRCh38
rs966537552 Chr.2: 56184073-56184073 on Build GRCh38
rs186996172 Chr.2: 56184333-56184333 on Build GRCh38
rs562563456 Chr.2: 56184171-56184171 on Build GRCh38
rs77586647 Chr.2: 56184151-56184151 on Build GRCh38
rs1033777270 Chr.2: 56184304-56184304 on Build GRCh38
rs1002284495 Chr.2: 56184014-56184014 on Build GRCh38
rs887067809 Chr.2: 56184373-56184373 on Build GRCh38
rs983414257 Chr.2: 56184204-56184204 on Build GRCh38
rs555694443 Chr.2: 56184011-56184011 on Build GRCh38
rs551583315 Chr.2: 56184205-56184205 on Build GRCh38
rs924309533 Chr.2: 56184425-56184425 on Build GRCh38
rs1048611976 Chr.2: 56184420-56184420 on Build GRCh38
rs985885236 Chr.2: 56184148-56184148 on Build GRCh38
rs937742235 Chr.2: 56184439-56184439 on Build GRCh38
rs908932746 Chr.2: 56184435-56184435 on Build GRCh38
rs912535826 Chr.2: 56184393-56184393 on Build GRCh38
rs1044262530 Chr.2: 56184448-56184448 on Build GRCh38
rs539882709 Chr.2: 56184096-56184096 on Build GRCh38
rs960489 Chr.2: 56184228-56184228 on Build GRCh38
rs1035044576 Chr.2: 56184370-56184370 on Build GRCh38
rs988082306 Chr.2: 56184316-56184316 on Build GRCh38
rs963774813 Chr.2: 56184121-56184121 on Build GRCh38
rs962815157 Chr.2: 56184221-56184221 on Build GRCh38
rs918851999 Chr.2: 56184408-56184408 on Build GRCh38
rs113509779 Chr.2: 56184084-56184084 on Build GRCh38
rs75829550 Chr.2: 56184263-56184263 on Build GRCh38
rs1019396214 Chr.2: 56184118-56184118 on Build GRCh38
rs549225112 Chr.2: 56184158-56184158 on Build GRCh38
rs905306650 Chr.2: 56184337-56184337 on Build GRCh38
rs948604450 Chr.2: 56184440-56184440 on Build GRCh38
rs560091771 Chr.2: 56184126-56184126 on Build GRCh38
rs1020729749 Chr.2: 56184063-56184063 on Build GRCh38
rs563868697 Chr.2: 56184039-56184039 on Build GRCh38
rs921037964 Chr.2: 56184255-56184255 on Build GRCh38
rs999150761 Chr.2: 56184026-56184026 on Build GRCh38
rs1023759685 Chr.2: 56184082-56184082 on Build GRCh38
rs960490 Chr.2: 56184022-56184022 on Build GRCh38
rs962917593 Chr.2: 56184388-56184388 on Build GRCh38
rs1004006424 Chr.2: 56184369-56184369 on Build GRCh38
rs1013435525 Chr.2: 56184326-56184326 on Build GRCh38
rs867061198 Chr.2: 56184297-56184297 on Build GRCh38
rs879408978 Chr.2: 56184336-56184336 on Build GRCh38
rs985593598 Chr.2: 56184372-56184372 on Build GRCh38
rs531561120 Chr.2: 56184192-56184192 on Build GRCh38
rs944059365 Chr.2: 56184400-56184400 on Build GRCh38
rs370051404 Chr.2: 56184341-56184341 on Build GRCh38
rs958070400 Chr.2: 56184293-56184293 on Build GRCh38
rs924249358 Chr.2: 56184267-56184267 on Build GRCh38
rs577303738 Chr.2: 56184045-56184045 on Build GRCh38

View Full Product Details