Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70809643-70810139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4F
Entrez Gene ID
165530
Gene Name
C-type lectin domain family 4 member F
Gene Aliases
CLECSF13, KCLR
UniGene
Hs.681239
Gene Chromosome Location
Chr.2: 70808643-70825235 on Build GRCh38
Transcripts
NM_001258027.1 NM_001321308.1 NM_173535.2 XM_011532635.2 XM_011532637.2 XM_011532638.2 XM_011532639.2 XM_011532640.2 XM_011532641.2 XM_011532642.2 XM_011532643.1 XM_017003519.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
443156 c.1607G>T p.G536V Chr.2: 70809790-70809790 on Build GRCh38
3991419 c.1611C>T p.S537S Chr.2: 70809786-70809786 on Build GRCh38
7361514 c.1568T>C p.V523A Chr.2: 70809829-70809829 on Build GRCh38
6092035 c.1624G>C p.D542H Chr.2: 70809773-70809773 on Build GRCh38
7361515 c.1547T>C p.L516P Chr.2: 70809850-70809850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369383205 Chr.2: 70809876-70809876 on Build GRCh38
rs782555838 Chr.2: 70809808-70809808 on Build GRCh38
rs34875126 Chr.2: 70809850-70809850 on Build GRCh38
rs201579889 Chr.2: 70809752-70809752 on Build GRCh38
rs781797471 Chr.2: 70809723-70809723 on Build GRCh38
rs375297484 Chr.2: 70809693-70809693 on Build GRCh38
rs781929303 Chr.2: 70810079-70810079 on Build GRCh38
rs782205530 Chr.2: 70809727-70809727 on Build GRCh38
rs782518965 Chr.2: 70809738-70809738 on Build GRCh38
rs782715985 Chr.2: 70809889-70809889 on Build GRCh38
rs561064679 Chr.2: 70809816-70809816 on Build GRCh38
rs373262526 Chr.2: 70809756-70809756 on Build GRCh38
rs782399642 Chr.2: 70809839-70809839 on Build GRCh38
rs199985963 Chr.2: 70809725-70809725 on Build GRCh38
rs542885903 Chr.2: 70809761-70809761 on Build GRCh38
rs568946922 Chr.2: 70809958-70809958 on Build GRCh38
rs782430983 Chr.2: 70809724-70809724 on Build GRCh38
rs781915691 Chr.2: 70809861-70809861 on Build GRCh38
rs782548920 Chr.2: 70809696-70809696 on Build GRCh38
rs782441312 Chr.2: 70809821-70809821 on Build GRCh38
rs781890521 Chr.2: 70809880-70809880 on Build GRCh38
rs201250175 Chr.2: 70809873-70809873 on Build GRCh38
rs782480125 Chr.2: 70809896-70809896 on Build GRCh38
rs782408952 Chr.2: 70809745-70809745 on Build GRCh38
rs144900238 Chr.2: 70809804-70809804 on Build GRCh38
rs782301690 Chr.2: 70809744-70809744 on Build GRCh38
rs150339964 Chr.2: 70809829-70809829 on Build GRCh38
rs552307036 Chr.2: 70809942-70809942 on Build GRCh38
rs782361898 Chr.2: 70809753-70809753 on Build GRCh38
rs781845318 Chr.2: 70809894-70809894 on Build GRCh38
rs782139934 Chr.2: 70809759-70809759 on Build GRCh38
rs368376926 Chr.2: 70809936-70809936 on Build GRCh38
rs201804036 Chr.2: 70809815-70809815 on Build GRCh38
rs782737026 Chr.2: 70809690-70809690 on Build GRCh38
rs782636606 Chr.2: 70809739-70809739 on Build GRCh38
rs782055755 Chr.2: 70809761-70809762 on Build GRCh38
rs139353507 Chr.2: 70809781-70809781 on Build GRCh38
rs782068286 Chr.2: 70809887-70809887 on Build GRCh38
rs782336643 Chr.2: 70809870-70809870 on Build GRCh38
rs782318976 Chr.2: 70809855-70809855 on Build GRCh38
rs79792059 Chr.2: 70809964-70809964 on Build GRCh38
rs782411904 Chr.2: 70809881-70809881 on Build GRCh38
rs545107860 Chr.2: 70809792-70809792 on Build GRCh38
rs781884335 Chr.2: 70809892-70809893 on Build GRCh38
rs782481443 Chr.2: 70809796-70809796 on Build GRCh38
rs540370999 Chr.2: 70809798-70809798 on Build GRCh38
rs146771126 Chr.2: 70809828-70809828 on Build GRCh38
rs782211960 Chr.2: 70809836-70809836 on Build GRCh38
rs376387699 Chr.2: 70809854-70809854 on Build GRCh38
rs532308950 Chr.2: 70809879-70809879 on Build GRCh38
rs940936830 Chr.2: 70809751-70809751 on Build GRCh38
rs35001900 Chr.2: 70809799-70809799 on Build GRCh38
rs148732839 Chr.2: 70809782-70809782 on Build GRCh38
rs889208565 Chr.2: 70809805-70809805 on Build GRCh38
rs782789953 Chr.2: 70809717-70809717 on Build GRCh38
rs145732605 Chr.2: 70809747-70809747 on Build GRCh38
rs367995245 Chr.2: 70809947-70809947 on Build GRCh38

View Full Product Details