Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73258508-73259017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
UniGene
Hs.23158
Gene Chromosome Location
Chr.2: 73254682-73284453 on Build GRCh38
Transcripts
NM_001080410.2 XM_005264158.3 XM_006711950.2 XM_011532562.1 XM_011532563.2 XM_017003421.1 XM_017003422.1 XM_017003423.1

SNP Detail

SNP ID Chromosome Location
rs534167655 Chr.2: 73258587-73258587 on Build GRCh38
rs983599007 Chr.2: 73258672-73258672 on Build GRCh38
rs944887003 Chr.2: 73258752-73258752 on Build GRCh38
rs756124881 Chr.2: 73258793-73258793 on Build GRCh38
rs775195418 Chr.2: 73258966-73258966 on Build GRCh38
rs372442356 Chr.2: 73258773-73258773 on Build GRCh38
rs879415931 Chr.2: 73258897-73258897 on Build GRCh38
rs866786695 Chr.2: 73258568-73258568 on Build GRCh38
rs370166126 Chr.2: 73258967-73258967 on Build GRCh38
rs370061489 Chr.2: 73258665-73258665 on Build GRCh38
rs761196272 Chr.2: 73258978-73258978 on Build GRCh38
rs923857854 Chr.2: 73258763-73258763 on Build GRCh38
rs140489725 Chr.2: 73258739-73258739 on Build GRCh38
rs1016047855 Chr.2: 73258684-73258684 on Build GRCh38
rs768245332 Chr.2: 73258968-73258968 on Build GRCh38
rs80319113 Chr.2: 73258885-73258885 on Build GRCh38
rs1011563615 Chr.2: 73258554-73258554 on Build GRCh38
rs759473513 Chr.2: 73258848-73258848 on Build GRCh38
rs927712397 Chr.2: 73258565-73258565 on Build GRCh38
rs767121287 Chr.2: 73258979-73258979 on Build GRCh38
rs561109380 Chr.2: 73258659-73258659 on Build GRCh38
rs996092514 Chr.2: 73258774-73258774 on Build GRCh38
rs958994434 Chr.2: 73258579-73258579 on Build GRCh38
rs950897800 Chr.2: 73258655-73258655 on Build GRCh38
rs1036342741 Chr.2: 73258654-73258654 on Build GRCh38
rs959109134 Chr.2: 73258975-73258975 on Build GRCh38
rs564585221 Chr.2: 73258785-73258785 on Build GRCh38
rs893158272 Chr.2: 73258861-73258861 on Build GRCh38
rs921575520 Chr.2: 73258740-73258740 on Build GRCh38
rs1005066392 Chr.2: 73258981-73258981 on Build GRCh38
rs761177597 Chr.2: 73258991-73258991 on Build GRCh38
rs780991069 Chr.2: 73258940-73258940 on Build GRCh38
rs559200824 Chr.2: 73258884-73258884 on Build GRCh38
rs990488757 Chr.2: 73258586-73258586 on Build GRCh38
rs765105355 Chr.2: 73258859-73258859 on Build GRCh38
rs536316404 Chr.2: 73258942-73258942 on Build GRCh38
rs531516004 Chr.2: 73258664-73258664 on Build GRCh38
rs1003280425 Chr.2: 73258953-73258953 on Build GRCh38
rs565617439 Chr.2: 73258888-73258888 on Build GRCh38
rs766208278 Chr.2: 73258692-73258692 on Build GRCh38
rs906128568 Chr.2: 73258593-73258593 on Build GRCh38
rs199625082 Chr.2: 73258990-73258990 on Build GRCh38
rs920298237 Chr.2: 73258604-73258604 on Build GRCh38
rs1053170738 Chr.2: 73258531-73258531 on Build GRCh38
rs112137477 Chr.2: 73258700-73258700 on Build GRCh38
rs138268745 Chr.2: 73258974-73258974 on Build GRCh38
rs933241501 Chr.2: 73258750-73258750 on Build GRCh38
rs893205265 Chr.2: 73258552-73258552 on Build GRCh38
rs753684556 Chr.2: 73258757-73258757 on Build GRCh38
rs769732247 Chr.2: 73258961-73258961 on Build GRCh38
rs757420604 Chr.2: 73258903-73258903 on Build GRCh38
rs542437515 Chr.2: 73258611-73258611 on Build GRCh38
rs1003091161 Chr.2: 73258599-73258599 on Build GRCh38
rs930902043 Chr.2: 73258610-73258610 on Build GRCh38
rs935190323 Chr.2: 73258810-73258810 on Build GRCh38
rs531969229 Chr.2: 73258876-73258876 on Build GRCh38

View Full Product Details