Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74365813-74366311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN1
Entrez Gene ID
1639
Gene Name
dynactin subunit 1
Gene Aliases
DAP-150, DP-150, P135
UniGene
Hs.516111
Gene Chromosome Location
Chr.2: 74361154-74392087 on Build GRCh38
Transcripts
NM_001135040.2 NM_001135041.2 NM_001190836.1 NM_001190837.1 NM_004082.4 NM_023019.3 NR_033935.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203169 c.2843G>A p.R948Q Chr.2: 74365936-74365936 on Build GRCh38
6010167 c.2860G>A p.E954K Chr.2: 74365919-74365919 on Build GRCh38
6456395 c.2746C>T p.R916W Chr.2: 74366258-74366258 on Build GRCh38
3583338 c.2842C>T p.R948* Chr.2: 74365937-74365937 on Build GRCh38
6641115 c.2748G>C p.R916R Chr.2: 74366256-74366256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747099902 Chr.2: 74366242-74366242 on Build GRCh38
rs780644304 Chr.2: 74365943-74365943 on Build GRCh38
rs749833142 Chr.2: 74366200-74366200 on Build GRCh38
rs779423676 Chr.2: 74366210-74366210 on Build GRCh38
rs751743289 Chr.2: 74365868-74365868 on Build GRCh38
rs72659382 Chr.2: 74365838-74365838 on Build GRCh38
rs773462298 Chr.2: 74366226-74366226 on Build GRCh38
rs767199008 Chr.2: 74366065-74366065 on Build GRCh38
rs754399792 Chr.2: 74366055-74366055 on Build GRCh38
rs555165454 Chr.2: 74366098-74366098 on Build GRCh38
rs769044539 Chr.2: 74365960-74365960 on Build GRCh38
rs767128134 Chr.2: 74366263-74366263 on Build GRCh38
rs780584687 Chr.2: 74366019-74366019 on Build GRCh38
rs114444981 Chr.2: 74365852-74365852 on Build GRCh38
rs778585634 Chr.2: 74365908-74365908 on Build GRCh38
rs745321425 Chr.2: 74366015-74366015 on Build GRCh38
rs199780349 Chr.2: 74365934-74365934 on Build GRCh38
rs138305126 Chr.2: 74365944-74365944 on Build GRCh38
rs765216385 Chr.2: 74365994-74365994 on Build GRCh38
rs895893019 Chr.2: 74366163-74366163 on Build GRCh38
rs941936627 Chr.2: 74366124-74366124 on Build GRCh38
rs774804530 Chr.2: 74366032-74366032 on Build GRCh38
rs763101727 Chr.2: 74365842-74365842 on Build GRCh38
rs780186070 Chr.2: 74365857-74365857 on Build GRCh38
rs1031165508 Chr.2: 74366180-74366180 on Build GRCh38
rs200545447 Chr.2: 74366037-74366037 on Build GRCh38
rs531275207 Chr.2: 74366031-74366031 on Build GRCh38
rs374197215 Chr.2: 74366066-74366066 on Build GRCh38
rs369754137 Chr.2: 74366068-74366068 on Build GRCh38
rs948832837 Chr.2: 74366164-74366164 on Build GRCh38
rs186615787 Chr.2: 74366057-74366057 on Build GRCh38
rs749706596 Chr.2: 74366023-74366023 on Build GRCh38
rs777391237 Chr.2: 74365936-74365936 on Build GRCh38
rs755686027 Chr.2: 74366271-74366271 on Build GRCh38
rs373818927 Chr.2: 74365985-74365985 on Build GRCh38
rs752851562 Chr.2: 74366011-74366011 on Build GRCh38
rs752469625 Chr.2: 74365931-74365931 on Build GRCh38
rs1051611543 Chr.2: 74366086-74366086 on Build GRCh38
rs973376180 Chr.2: 74366129-74366129 on Build GRCh38
rs751246549 Chr.2: 74366010-74366010 on Build GRCh38
rs960860261 Chr.2: 74366185-74366185 on Build GRCh38
rs979497373 Chr.2: 74365835-74365835 on Build GRCh38
rs1050338834 Chr.2: 74365969-74365969 on Build GRCh38
rs991947321 Chr.2: 74366197-74366197 on Build GRCh38
rs904724647 Chr.2: 74366245-74366245 on Build GRCh38
rs771075973 Chr.2: 74366251-74366251 on Build GRCh38
rs756911529 Chr.2: 74366060-74366060 on Build GRCh38
rs779387697 Chr.2: 74366070-74366070 on Build GRCh38
rs987924253 Chr.2: 74365884-74365884 on Build GRCh38
rs758168607 Chr.2: 74366000-74366000 on Build GRCh38
rs1023165285 Chr.2: 74366039-74366039 on Build GRCh38
rs75503452 Chr.2: 74366261-74366261 on Build GRCh38
rs567179271 Chr.2: 74366115-74366115 on Build GRCh38
rs986355786 Chr.2: 74366104-74366104 on Build GRCh38
rs72659381 Chr.2: 74366268-74366268 on Build GRCh38
rs766456562 Chr.2: 74365878-74365878 on Build GRCh38
rs754893423 Chr.2: 74365896-74365896 on Build GRCh38
rs756273697 Chr.2: 74366138-74366138 on Build GRCh38
rs774681414 Chr.2: 74365966-74365966 on Build GRCh38
rs375079576 Chr.2: 74366257-74366257 on Build GRCh38
rs201914453 Chr.2: 74366003-74366003 on Build GRCh38
rs753405772 Chr.2: 74365907-74365907 on Build GRCh38
rs761587735 Chr.2: 74366262-74366262 on Build GRCh38
rs2735283 Chr.2: 74365840-74365840 on Build GRCh38
rs910409595 Chr.2: 74366118-74366118 on Build GRCh38
rs549475401 Chr.2: 74366036-74366036 on Build GRCh38
rs752493458 Chr.2: 74365999-74365999 on Build GRCh38
rs1030103694 Chr.2: 74366095-74366095 on Build GRCh38
rs748518770 Chr.2: 74366219-74366219 on Build GRCh38
rs145130328 Chr.2: 74365974-74365974 on Build GRCh38
rs929309842 Chr.2: 74366157-74366157 on Build GRCh38
rs767725965 Chr.2: 74365984-74365984 on Build GRCh38
rs762138930 Chr.2: 74366034-74366034 on Build GRCh38
rs537761448 Chr.2: 74366254-74366254 on Build GRCh38
rs775726255 Chr.2: 74365946-74365946 on Build GRCh38
rs78785770 Chr.2: 74366078-74366078 on Build GRCh38
rs1007287725 Chr.2: 74366096-74366096 on Build GRCh38
rs767442935 Chr.2: 74365948-74365948 on Build GRCh38
rs956441865 Chr.2: 74365862-74365862 on Build GRCh38
rs924086052 Chr.2: 74365967-74365967 on Build GRCh38
rs371973468 Chr.2: 74366224-74366224 on Build GRCh38
rs2528571 Chr.2: 74365848-74365848 on Build GRCh38
rs916304569 Chr.2: 74366198-74366198 on Build GRCh38
rs550554047 Chr.2: 74366017-74366017 on Build GRCh38
rs1036199881 Chr.2: 74366058-74366062 on Build GRCh38
rs752651198 Chr.2: 74366045-74366045 on Build GRCh38
rs748058825 Chr.2: 74365862-74365862 on Build GRCh38
rs956368812 Chr.2: 74366074-74366074 on Build GRCh38
rs746572197 Chr.2: 74365937-74365937 on Build GRCh38
rs753748221 Chr.2: 74365881-74365881 on Build GRCh38
rs757386032 Chr.2: 74365871-74365871 on Build GRCh38
rs200005865 Chr.2: 74366049-74366049 on Build GRCh38
rs1057116218 Chr.2: 74366161-74366161 on Build GRCh38
rs550195526 Chr.2: 74365956-74365956 on Build GRCh38
rs909178 Chr.2: 74366208-74366208 on Build GRCh38
rs755482521 Chr.2: 74366207-74366207 on Build GRCh38
rs763956654 Chr.2: 74366258-74366258 on Build GRCh38

View Full Product Details