Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85371268-85371667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD3
Entrez Gene ID
84173
Gene Name
ELMO domain containing 3
Gene Aliases
DFNB88, LST3, RBED1, RBM29
UniGene
Hs.269990
Gene Chromosome Location
Chr.2: 85354394-85391752 on Build GRCh38
Transcripts
NM_001135021.1 NM_001135022.1 NM_001135023.1 NM_032213.4 XM_005264596.3 XM_005264597.3 XM_005264598.3 XM_017005078.1 XM_017005079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
723028 c.523C>T p.L175F Chr.2: 85371478-85371478 on Build GRCh38
1615173 c.588G>A p.W196* Chr.2: 85371543-85371543 on Build GRCh38
4831702 c.540G>A p.K180K Chr.2: 85371495-85371495 on Build GRCh38
1409779 c.573T>C p.L191L Chr.2: 85371528-85371528 on Build GRCh38
1615172 c.588G>A p.W196* Chr.2: 85371543-85371543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758849415 Chr.2: 85371378-85371378 on Build GRCh38
rs750341592 Chr.2: 85371505-85371505 on Build GRCh38
rs753809916 Chr.2: 85371482-85371482 on Build GRCh38
rs189400834 Chr.2: 85371628-85371628 on Build GRCh38
rs910363899 Chr.2: 85371337-85371337 on Build GRCh38
rs534872052 Chr.2: 85371311-85371311 on Build GRCh38
rs376101201 Chr.2: 85371317-85371319 on Build GRCh38
rs772622572 Chr.2: 85371515-85371515 on Build GRCh38
rs148943018 Chr.2: 85371537-85371537 on Build GRCh38
rs773925336 Chr.2: 85371399-85371399 on Build GRCh38
rs900922061 Chr.2: 85371305-85371305 on Build GRCh38
rs370124766 Chr.2: 85371319-85371321 on Build GRCh38
rs979178219 Chr.2: 85371633-85371633 on Build GRCh38
rs115378987 Chr.2: 85371504-85371504 on Build GRCh38
rs575583374 Chr.2: 85371472-85371472 on Build GRCh38
rs548659210 Chr.2: 85371306-85371306 on Build GRCh38
rs139255752 Chr.2: 85371582-85371582 on Build GRCh38
rs372910890 Chr.2: 85371578-85371578 on Build GRCh38
rs777617403 Chr.2: 85371302-85371302 on Build GRCh38
rs778937204 Chr.2: 85371491-85371491 on Build GRCh38
rs917283750 Chr.2: 85371437-85371437 on Build GRCh38
rs1002891878 Chr.2: 85371524-85371524 on Build GRCh38
rs987926063 Chr.2: 85371312-85371312 on Build GRCh38
rs146704263 Chr.2: 85371320-85371320 on Build GRCh38
rs765623161 Chr.2: 85371332-85371332 on Build GRCh38
rs751491844 Chr.2: 85371304-85371304 on Build GRCh38
rs747562452 Chr.2: 85371530-85371530 on Build GRCh38
rs369497831 Chr.2: 85371439-85371439 on Build GRCh38
rs780541317 Chr.2: 85371644-85371644 on Build GRCh38
rs557031260 Chr.2: 85371384-85371384 on Build GRCh38
rs961510555 Chr.2: 85371343-85371343 on Build GRCh38
rs184768722 Chr.2: 85371319-85371319 on Build GRCh38
rs116470781 Chr.2: 85371517-85371517 on Build GRCh38
rs763860824 Chr.2: 85371453-85371453 on Build GRCh38
rs147341196 Chr.2: 85371445-85371445 on Build GRCh38
rs755239764 Chr.2: 85371485-85371485 on Build GRCh38
rs758403657 Chr.2: 85371526-85371539 on Build GRCh38
rs369642265 Chr.2: 85371416-85371416 on Build GRCh38
rs960392590 Chr.2: 85371563-85371563 on Build GRCh38
rs925977582 Chr.2: 85371460-85371460 on Build GRCh38
rs764749612 Chr.2: 85371440-85371440 on Build GRCh38
rs1035274955 Chr.2: 85371531-85371531 on Build GRCh38
rs567069997 Chr.2: 85371309-85371309 on Build GRCh38
rs755545293 Chr.2: 85371601-85371601 on Build GRCh38
rs780212959 Chr.2: 85371477-85371477 on Build GRCh38
rs769339643 Chr.2: 85371596-85371596 on Build GRCh38
rs761251824 Chr.2: 85371406-85371406 on Build GRCh38
rs748378091 Chr.2: 85371505-85371505 on Build GRCh38
rs752504989 Chr.2: 85371441-85371441 on Build GRCh38
rs770524326 Chr.2: 85371411-85371411 on Build GRCh38
rs772712336 Chr.2: 85371328-85371328 on Build GRCh38
rs750753118 Chr.2: 85371347-85371347 on Build GRCh38
rs757152587 Chr.2: 85371473-85371473 on Build GRCh38
rs775977369 Chr.2: 85371432-85371432 on Build GRCh38
rs754023217 Chr.2: 85371550-85371550 on Build GRCh38
rs780663226 Chr.2: 85371503-85371503 on Build GRCh38
rs967807235 Chr.2: 85371632-85371632 on Build GRCh38
rs539320797 Chr.2: 85371361-85371361 on Build GRCh38
rs768649695 Chr.2: 85371420-85371420 on Build GRCh38

View Full Product Details