Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97560354-97560863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36B
Entrez Gene ID
57730
Gene Name
ankyrin repeat domain 36B
Gene Aliases
KIAA1641
UniGene
Hs.532921
Gene Chromosome Location
Chr.2: 97492663-97590245 on Build GRCh38
Transcripts
NM_025190.3 XM_005263995.4 XM_006712661.1 XM_011511555.1 XM_011511556.1 XM_011511557.2 XM_011511558.1 XM_017004578.1 XM_017004579.1 XM_017004580.1 XM_017004581.1 XM_017004582.1 XM_017004583.1 XM_017004584.1 XM_017004585.1 XM_017004586.1 XM_017004587.1 XM_017004588.1 XM_017004589.1 XM_017004590.1 XM_017004592.1 XM_017004593.1 XM_017004594.1 XM_017004595.1

SNP Detail

SNP ID Chromosome Location
rs375216709 Chr.2: 97560803-97560803 on Build GRCh38
rs755162021 Chr.2: 97560502-97560502 on Build GRCh38
rs368692757 Chr.2: 97560761-97560764 on Build GRCh38
rs2922587 Chr.2: 97560647-97560647 on Build GRCh38
rs761004790 Chr.2: 97560785-97560785 on Build GRCh38
rs750616166 Chr.2: 97560784-97560784 on Build GRCh38
rs747797606 Chr.2: 97560643-97560643 on Build GRCh38
rs745703584 Chr.2: 97560622-97560622 on Build GRCh38
rs762070702 Chr.2: 97560683-97560683 on Build GRCh38
rs199835974 Chr.2: 97560604-97560604 on Build GRCh38
rs759780127 Chr.2: 97560676-97560676 on Build GRCh38
rs184695107 Chr.2: 97560499-97560499 on Build GRCh38
rs1011407185 Chr.2: 97560496-97560496 on Build GRCh38
rs575707899 Chr.2: 97560568-97560568 on Build GRCh38
rs772490159 Chr.2: 97560678-97560678 on Build GRCh38
rs370555422 Chr.2: 97560578-97560578 on Build GRCh38
rs796632266 Chr.2: 97560613-97560613 on Build GRCh38
rs79824900 Chr.2: 97560437-97560437 on Build GRCh38
rs753620047 Chr.2: 97560795-97560795 on Build GRCh38
rs548389472 Chr.2: 97560393-97560393 on Build GRCh38
rs780012890 Chr.2: 97560704-97560704 on Build GRCh38
rs74805914 Chr.2: 97560636-97560636 on Build GRCh38
rs570732004 Chr.2: 97560524-97560524 on Build GRCh38
rs77143397 Chr.2: 97560542-97560542 on Build GRCh38
rs766240137 Chr.2: 97560794-97560794 on Build GRCh38
rs900251619 Chr.2: 97560512-97560513 on Build GRCh38
rs916732644 Chr.2: 97560618-97560618 on Build GRCh38
rs757841915 Chr.2: 97560831-97560831 on Build GRCh38
rs553736341 Chr.2: 97560456-97560456 on Build GRCh38
rs2442256 Chr.2: 97560754-97560754 on Build GRCh38
rs771664255 Chr.2: 97560645-97560645 on Build GRCh38
rs11681640 Chr.2: 97560661-97560661 on Build GRCh38
rs756109596 Chr.2: 97560628-97560628 on Build GRCh38
rs796528969 Chr.2: 97560612-97560612 on Build GRCh38
rs770091657 Chr.2: 97560758-97560758 on Build GRCh38
rs377631596 Chr.2: 97560751-97560752 on Build GRCh38
rs201800024 Chr.2: 97560386-97560386 on Build GRCh38
rs564617173 Chr.2: 97560747-97560747 on Build GRCh38
rs76656845 Chr.2: 97560553-97560553 on Build GRCh38
rs995023011 Chr.2: 97560840-97560840 on Build GRCh38
rs767711502 Chr.2: 97560776-97560776 on Build GRCh38
rs761866937 Chr.2: 97560770-97560770 on Build GRCh38
rs534012858 Chr.2: 97560434-97560434 on Build GRCh38
rs777282868 Chr.2: 97560832-97560832 on Build GRCh38
rs749246923 Chr.2: 97560631-97560631 on Build GRCh38
rs754923027 Chr.2: 97560802-97560802 on Build GRCh38
rs752682006 Chr.2: 97560823-97560823 on Build GRCh38
rs2261700 Chr.2: 97560842-97560842 on Build GRCh38
rs369923997 Chr.2: 97560769-97560769 on Build GRCh38
rs369487924 Chr.2: 97560648-97560648 on Build GRCh38
rs982206213 Chr.2: 97560624-97560624 on Build GRCh38
rs374371976 Chr.2: 97560734-97560734 on Build GRCh38
rs755910024 Chr.2: 97560697-97560697 on Build GRCh38
rs568128253 Chr.2: 97560407-97560407 on Build GRCh38
rs754904396 Chr.2: 97560723-97560723 on Build GRCh38
rs375514942 Chr.2: 97560493-97560493 on Build GRCh38
rs891673971 Chr.2: 97560435-97560435 on Build GRCh38
rs771750373 Chr.2: 97560731-97560731 on Build GRCh38
rs775797963 Chr.2: 97560679-97560680 on Build GRCh38
rs775863880 Chr.2: 97560761-97560761 on Build GRCh38
rs368731044 Chr.2: 97560811-97560811 on Build GRCh38
rs144890806 Chr.2: 97560658-97560658 on Build GRCh38
rs75227888 Chr.2: 97560552-97560552 on Build GRCh38
rs555785091 Chr.2: 97560567-97560567 on Build GRCh38
rs370270629 Chr.2: 97560679-97560679 on Build GRCh38
rs367884437 Chr.2: 97560678-97560678 on Build GRCh38
rs773756375 Chr.2: 97560640-97560640 on Build GRCh38
rs77767116 Chr.2: 97560412-97560412 on Build GRCh38
rs780211046 Chr.2: 97560630-97560630 on Build GRCh38
rs373994972 Chr.2: 97560587-97560587 on Build GRCh38
rs763484294 Chr.2: 97560766-97560766 on Build GRCh38
rs753761128 Chr.2: 97560711-97560711 on Build GRCh38
rs781100754 Chr.2: 97560621-97560621 on Build GRCh38
rs995915706 Chr.2: 97560533-97560533 on Build GRCh38
rs796425921 Chr.2: 97560703-97560703 on Build GRCh38
rs750242061 Chr.2: 97560687-97560687 on Build GRCh38
rs371722883 Chr.2: 97560461-97560461 on Build GRCh38
rs778457264 Chr.2: 97560725-97560725 on Build GRCh38
rs768154379 Chr.2: 97560638-97560638 on Build GRCh38
rs764191098 Chr.2: 97560670-97560670 on Build GRCh38
rs963566290 Chr.2: 97560582-97560582 on Build GRCh38
rs201940725 Chr.2: 97560822-97560822 on Build GRCh38
rs572503391 Chr.2: 97560726-97560726 on Build GRCh38

View Full Product Details