Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 99363560-99364059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF5B
Entrez Gene ID
9669
Gene Name
eukaryotic translation initiation factor 5B
Gene Aliases
IF2
UniGene
Hs.158688
Gene Chromosome Location
Chr.2: 99337371-99400265 on Build GRCh38
Transcripts
NM_015904.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4770053 c.1073A>T p.Q358L Chr.2: 99363798-99363798 on Build GRCh38
2914596 c.1093C>T p.R365C Chr.2: 99363818-99363818 on Build GRCh38
4520757 c.1074G>A p.Q358Q Chr.2: 99363799-99363799 on Build GRCh38
5620756 c.1046C>T p.A349V Chr.2: 99363771-99363771 on Build GRCh38
3584379 c.1048A>G p.K350E Chr.2: 99363773-99363773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375797022 Chr.2: 99363896-99363896 on Build GRCh38
rs776972763 Chr.2: 99363775-99363775 on Build GRCh38
rs368501708 Chr.2: 99363706-99363706 on Build GRCh38
rs758896433 Chr.2: 99363852-99363852 on Build GRCh38
rs977737722 Chr.2: 99363744-99363744 on Build GRCh38
rs762815204 Chr.2: 99363805-99363805 on Build GRCh38
rs762910916 Chr.2: 99363641-99363641 on Build GRCh38
rs578008866 Chr.2: 99363663-99363663 on Build GRCh38
rs775292334 Chr.2: 99363793-99363793 on Build GRCh38
rs966720686 Chr.2: 99363620-99363620 on Build GRCh38
rs752603680 Chr.2: 99363630-99363630 on Build GRCh38
rs527908818 Chr.2: 99363960-99363960 on Build GRCh38
rs989455717 Chr.2: 99363584-99363584 on Build GRCh38
rs769511995 Chr.2: 99363784-99363784 on Build GRCh38
rs774290174 Chr.2: 99363914-99363914 on Build GRCh38
rs766960926 Chr.2: 99363827-99363827 on Build GRCh38
rs548433328 Chr.2: 99363887-99363887 on Build GRCh38
rs780789499 Chr.2: 99363674-99363674 on Build GRCh38
rs1055903822 Chr.2: 99363984-99363984 on Build GRCh38
rs948427988 Chr.2: 99363738-99363738 on Build GRCh38
rs531826437 Chr.2: 99363866-99363866 on Build GRCh38
rs766900505 Chr.2: 99363907-99363908 on Build GRCh38
rs761403299 Chr.2: 99363677-99363679 on Build GRCh38
rs747083956 Chr.2: 99363755-99363755 on Build GRCh38
rs967154547 Chr.2: 99363991-99363991 on Build GRCh38
rs768522531 Chr.2: 99363903-99363903 on Build GRCh38
rs763519466 Chr.2: 99363642-99363642 on Build GRCh38
rs978775415 Chr.2: 99363627-99363627 on Build GRCh38
rs546134984 Chr.2: 99363687-99363687 on Build GRCh38
rs1014413962 Chr.2: 99364010-99364010 on Build GRCh38
rs746086268 Chr.2: 99363888-99363888 on Build GRCh38
rs371427237 Chr.2: 99363894-99363894 on Build GRCh38
rs552555606 Chr.2: 99364024-99364024 on Build GRCh38
rs928236911 Chr.2: 99363651-99363651 on Build GRCh38
rs371784944 Chr.2: 99363828-99363828 on Build GRCh38
rs866550212 Chr.2: 99363801-99363801 on Build GRCh38
rs1010973444 Chr.2: 99363981-99363981 on Build GRCh38
rs1022362270 Chr.2: 99363987-99363987 on Build GRCh38
rs199730736 Chr.2: 99363860-99363860 on Build GRCh38
rs772571739 Chr.2: 99363752-99363752 on Build GRCh38
rs10642 Chr.2: 99363734-99363734 on Build GRCh38
rs71413834 Chr.2: 99363730-99363730 on Build GRCh38
rs753090811 Chr.2: 99363851-99363851 on Build GRCh38
rs915196887 Chr.2: 99363594-99363594 on Build GRCh38
rs74430561 Chr.2: 99363661-99363661 on Build GRCh38
rs562742041 Chr.2: 99363772-99363772 on Build GRCh38
rs369815195 Chr.2: 99364029-99364033 on Build GRCh38
rs1001608330 Chr.2: 99363604-99363605 on Build GRCh38
rs11547668 Chr.2: 99363743-99363743 on Build GRCh38
rs547606165 Chr.2: 99363968-99363968 on Build GRCh38
rs915666759 Chr.2: 99363735-99363735 on Build GRCh38
rs1801875 Chr.2: 99363838-99363838 on Build GRCh38
rs761287210 Chr.2: 99363821-99363821 on Build GRCh38
rs903425436 Chr.2: 99364033-99364033 on Build GRCh38
rs75521402 Chr.2: 99363736-99363736 on Build GRCh38
rs3205296 Chr.2: 99363803-99363803 on Build GRCh38
rs759374647 Chr.2: 99363618-99363618 on Build GRCh38
rs749864947 Chr.2: 99363702-99363707 on Build GRCh38
rs779156736 Chr.2: 99363841-99363841 on Build GRCh38
rs764897553 Chr.2: 99363691-99363693 on Build GRCh38
rs562291227 Chr.2: 99363957-99363957 on Build GRCh38
rs750106725 Chr.2: 99363836-99363836 on Build GRCh38
rs748606443 Chr.2: 99363723-99363723 on Build GRCh38
rs771320014 Chr.2: 99363766-99363766 on Build GRCh38
rs749076771 Chr.2: 99363900-99363900 on Build GRCh38
rs367922942 Chr.2: 99363881-99363881 on Build GRCh38
rs763837602 Chr.2: 99363605-99363605 on Build GRCh38
rs918484508 Chr.2: 99363898-99363898 on Build GRCh38
rs369984953 Chr.2: 99363707-99363712 on Build GRCh38
rs751105863 Chr.2: 99363819-99363819 on Build GRCh38
rs776245567 Chr.2: 99363643-99363643 on Build GRCh38
rs1048072889 Chr.2: 99363787-99363787 on Build GRCh38
rs764562903 Chr.2: 99363963-99363963 on Build GRCh38
rs150512844 Chr.2: 99363625-99363625 on Build GRCh38
rs764081987 Chr.2: 99363808-99363808 on Build GRCh38
rs929934726 Chr.2: 99363901-99363901 on Build GRCh38
rs755316424 Chr.2: 99363697-99363697 on Build GRCh38
rs778055308 Chr.2: 99363754-99363754 on Build GRCh38
rs759784321 Chr.2: 99363782-99363782 on Build GRCh38

View Full Product Details