Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 100280838-100281357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LONRF2
Entrez Gene ID
164832
Gene Name
LON peptidase N-terminal domain and ring finger 2
Gene Aliases
RNF192
UniGene
Hs.21380
Gene Chromosome Location
Chr.2: 100273291-100322733 on Build GRCh38
Transcripts
NM_198461.3

SNP Detail

SNP ID Chromosome Location
rs766161882 Chr.2: 100281086-100281086 on Build GRCh38
rs7601873 Chr.2: 100281280-100281280 on Build GRCh38
rs934196264 Chr.2: 100281149-100281149 on Build GRCh38
rs866286483 Chr.2: 100281187-100281187 on Build GRCh38
rs1048353367 Chr.2: 100281054-100281054 on Build GRCh38
rs60454280 Chr.2: 100281278-100281281 on Build GRCh38
rs181896915 Chr.2: 100281058-100281058 on Build GRCh38
rs398060394 Chr.2: 100281280-100281283 on Build GRCh38
rs547792535 Chr.2: 100281174-100281174 on Build GRCh38
rs902770663 Chr.2: 100281173-100281173 on Build GRCh38
rs1036861892 Chr.2: 100281075-100281075 on Build GRCh38
rs778621693 Chr.2: 100281194-100281194 on Build GRCh38
rs928078262 Chr.2: 100280976-100280976 on Build GRCh38
rs966879981 Chr.2: 100280931-100280931 on Build GRCh38
rs76623699 Chr.2: 100281206-100281206 on Build GRCh38
rs960816760 Chr.2: 100280985-100280985 on Build GRCh38
rs916589830 Chr.2: 100281049-100281049 on Build GRCh38
rs7601442 Chr.2: 100280889-100280889 on Build GRCh38
rs192196142 Chr.2: 100281050-100281050 on Build GRCh38
rs1035204560 Chr.2: 100281195-100281195 on Build GRCh38
rs574704537 Chr.2: 100281281-100281284 on Build GRCh38
rs752954340 Chr.2: 100281312-100281312 on Build GRCh38
rs894600384 Chr.2: 100281219-100281219 on Build GRCh38
rs201677288 Chr.2: 100281281-100281281 on Build GRCh38
rs1024113394 Chr.2: 100281253-100281253 on Build GRCh38
rs187356015 Chr.2: 100281033-100281033 on Build GRCh38
rs935015158 Chr.2: 100281041-100281041 on Build GRCh38
rs978214179 Chr.2: 100280932-100280932 on Build GRCh38
rs1017426972 Chr.2: 100281215-100281215 on Build GRCh38
rs779733481 Chr.2: 100280974-100280974 on Build GRCh38
rs753475460 Chr.2: 100281278-100281285 on Build GRCh38
rs200286984 Chr.2: 100281279-100281279 on Build GRCh38
rs75357854 Chr.2: 100281296-100281296 on Build GRCh38
rs200634811 Chr.2: 100281277-100281280 on Build GRCh38
rs1010515684 Chr.2: 100281128-100281128 on Build GRCh38
rs2309817 Chr.2: 100281212-100281212 on Build GRCh38
rs963532749 Chr.2: 100280875-100280875 on Build GRCh38
rs547548906 Chr.2: 100281301-100281301 on Build GRCh38
rs1022528141 Chr.2: 100281166-100281166 on Build GRCh38
rs970581685 Chr.2: 100280992-100280992 on Build GRCh38
rs984769899 Chr.2: 100281332-100281332 on Build GRCh38
rs796568853 Chr.2: 100281022-100281024 on Build GRCh38
rs889476997 Chr.2: 100281118-100281118 on Build GRCh38
rs373646225 Chr.2: 100281024-100281026 on Build GRCh38
rs560549158 Chr.2: 100280975-100280975 on Build GRCh38
rs939856970 Chr.2: 100281071-100281071 on Build GRCh38
rs2309816 Chr.2: 100280997-100280997 on Build GRCh38
rs7557977 Chr.2: 100281111-100281111 on Build GRCh38
rs760676537 Chr.2: 100281076-100281076 on Build GRCh38
rs997953708 Chr.2: 100281103-100281103 on Build GRCh38

View Full Product Details