Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130152887-130153388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMPD4
Entrez Gene ID
55627
Gene Name
sphingomyelin phosphodiesterase 4
Gene Aliases
NET13, NSMASE-3, NSMASE3
UniGene
Hs.516450
Gene Chromosome Location
Chr.2: 130151392-130182750 on Build GRCh38
Transcripts
NM_001171083.2 NM_017751.4 NM_017951.4 NR_033230.2 NR_033231.2 NR_033232.2 XM_011511443.2 XM_011511444.2 XM_011511445.2 XM_017004448.1 XM_017004449.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6366071 c.2200C>T p.P734S Chr.2: 130153114-130153114 on Build GRCh38
6378078 c.2261T>C p.I754T Chr.2: 130153053-130153053 on Build GRCh38
3566381 c.2220C>T p.I740I Chr.2: 130153094-130153094 on Build GRCh38
3566382 c.2097C>T p.I699I Chr.2: 130153364-130153364 on Build GRCh38
4399155 c.2086C>T p.P696S Chr.2: 130153375-130153375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758283567 Chr.2: 130152927-130152927 on Build GRCh38
rs765167051 Chr.2: 130153326-130153326 on Build GRCh38
rs753956321 Chr.2: 130153207-130153207 on Build GRCh38
rs776147444 Chr.2: 130153037-130153037 on Build GRCh38
rs773287870 Chr.2: 130153066-130153066 on Build GRCh38
rs777580014 Chr.2: 130153347-130153347 on Build GRCh38
rs372272450 Chr.2: 130153016-130153016 on Build GRCh38
rs559232746 Chr.2: 130152953-130152953 on Build GRCh38
rs762531432 Chr.2: 130153155-130153155 on Build GRCh38
rs201205980 Chr.2: 130153307-130153307 on Build GRCh38
rs41269835 Chr.2: 130153343-130153343 on Build GRCh38
rs766299358 Chr.2: 130153076-130153076 on Build GRCh38
rs369700397 Chr.2: 130153080-130153080 on Build GRCh38
rs375396456 Chr.2: 130153062-130153062 on Build GRCh38
rs984530546 Chr.2: 130153219-130153219 on Build GRCh38
rs978772382 Chr.2: 130152957-130152957 on Build GRCh38
rs960988783 Chr.2: 130153183-130153183 on Build GRCh38
rs771940382 Chr.2: 130153352-130153352 on Build GRCh38
rs746314229 Chr.2: 130153108-130153108 on Build GRCh38
rs188562076 Chr.2: 130152992-130152992 on Build GRCh38
rs940407867 Chr.2: 130153231-130153231 on Build GRCh38
rs752992558 Chr.2: 130153194-130153194 on Build GRCh38
rs568772435 Chr.2: 130153189-130153189 on Build GRCh38
rs747737640 Chr.2: 130153124-130153124 on Build GRCh38
rs908950539 Chr.2: 130153225-130153226 on Build GRCh38
rs368168049 Chr.2: 130153364-130153364 on Build GRCh38
rs1022338686 Chr.2: 130152945-130152947 on Build GRCh38
rs760835570 Chr.2: 130153197-130153197 on Build GRCh38
rs765234023 Chr.2: 130153022-130153022 on Build GRCh38
rs766433330 Chr.2: 130152982-130152982 on Build GRCh38
rs755180132 Chr.2: 130152995-130152995 on Build GRCh38
rs761682669 Chr.2: 130153226-130153227 on Build GRCh38
rs867814872 Chr.2: 130153125-130153125 on Build GRCh38
rs553165123 Chr.2: 130152973-130152973 on Build GRCh38
rs201893351 Chr.2: 130153338-130153338 on Build GRCh38
rs557503931 Chr.2: 130153337-130153337 on Build GRCh38
rs376833804 Chr.2: 130152980-130152980 on Build GRCh38
rs200559691 Chr.2: 130153107-130153107 on Build GRCh38
rs553662129 Chr.2: 130153171-130153171 on Build GRCh38
rs759531506 Chr.2: 130153134-130153134 on Build GRCh38
rs767470899 Chr.2: 130153184-130153185 on Build GRCh38
rs766211559 Chr.2: 130152932-130152932 on Build GRCh38
rs779536559 Chr.2: 130153221-130153221 on Build GRCh38
rs35442126 Chr.2: 130153327-130153328 on Build GRCh38
rs758782981 Chr.2: 130153331-130153331 on Build GRCh38
rs535668738 Chr.2: 130153199-130153199 on Build GRCh38
rs774645435 Chr.2: 130153026-130153026 on Build GRCh38
rs542879796 Chr.2: 130152926-130152926 on Build GRCh38
rs997820584 Chr.2: 130153040-130153040 on Build GRCh38
rs757806814 Chr.2: 130153097-130153097 on Build GRCh38
rs373235873 Chr.2: 130153164-130153164 on Build GRCh38
rs368968551 Chr.2: 130153030-130153030 on Build GRCh38
rs765335407 Chr.2: 130152911-130152911 on Build GRCh38
rs759387305 Chr.2: 130153179-130153179 on Build GRCh38
rs746192514 Chr.2: 130153028-130153028 on Build GRCh38
rs372808796 Chr.2: 130153061-130153061 on Build GRCh38
rs559220661 Chr.2: 130153363-130153363 on Build GRCh38
rs370467960 Chr.2: 130153299-130153299 on Build GRCh38
rs377386891 Chr.2: 130153295-130153295 on Build GRCh38
rs576034962 Chr.2: 130153229-130153229 on Build GRCh38
rs377358034 Chr.2: 130153208-130153208 on Build GRCh38
rs747825514 Chr.2: 130153283-130153283 on Build GRCh38
rs756188829 Chr.2: 130153086-130153086 on Build GRCh38
rs771920003 Chr.2: 130153300-130153300 on Build GRCh38
rs752802268 Chr.2: 130153082-130153082 on Build GRCh38
rs200458140 Chr.2: 130153294-130153294 on Build GRCh38
rs750972713 Chr.2: 130153327-130153327 on Build GRCh38
rs758798595 Chr.2: 130153270-130153270 on Build GRCh38
rs552264997 Chr.2: 130153081-130153081 on Build GRCh38
rs143470110 Chr.2: 130153109-130153109 on Build GRCh38
rs940353024 Chr.2: 130153021-130153021 on Build GRCh38
rs201666305 Chr.2: 130152948-130152949 on Build GRCh38
rs756822260 Chr.2: 130153350-130153350 on Build GRCh38
rs770286654 Chr.2: 130153296-130153296 on Build GRCh38
rs202061208 Chr.2: 130153308-130153308 on Build GRCh38
rs762946445 Chr.2: 130153075-130153075 on Build GRCh38
rs35114946 Chr.2: 130152913-130152914 on Build GRCh38
rs769461501 Chr.2: 130153129-130153129 on Build GRCh38
rs916705816 Chr.2: 130152986-130152986 on Build GRCh38
rs749726318 Chr.2: 130153022-130153022 on Build GRCh38
rs775196106 Chr.2: 130153303-130153303 on Build GRCh38
rs780913869 Chr.2: 130153110-130153110 on Build GRCh38
rs887482170 Chr.2: 130153020-130153020 on Build GRCh38
rs750502468 Chr.2: 130153061-130153061 on Build GRCh38
rs371316712 Chr.2: 130153015-130153015 on Build GRCh38
rs939569012 Chr.2: 130152968-130152968 on Build GRCh38
rs557572416 Chr.2: 130153223-130153223 on Build GRCh38
rs751420866 Chr.2: 130153078-130153078 on Build GRCh38
rs761898713 Chr.2: 130153322-130153322 on Build GRCh38
rs146485657 Chr.2: 130153094-130153094 on Build GRCh38
rs185280937 Chr.2: 130153351-130153351 on Build GRCh38
rs772800099 Chr.2: 130153137-130153137 on Build GRCh38
rs778264373 Chr.2: 130153018-130153018 on Build GRCh38
rs761896214 Chr.2: 130152934-130152935 on Build GRCh38
rs374659255 Chr.2: 130153023-130153023 on Build GRCh38
rs774142444 Chr.2: 130153166-130153166 on Build GRCh38
rs766890210 Chr.2: 130153335-130153335 on Build GRCh38
rs748882791 Chr.2: 130153349-130153349 on Build GRCh38
rs76145869 Chr.2: 130153234-130153234 on Build GRCh38
rs4511676 Chr.2: 130153274-130153274 on Build GRCh38

View Full Product Details