Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 134945959-134946463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNT2
Entrez Gene ID
905
Gene Name
cyclin T2
Gene Aliases
CYCT2
UniGene
Hs.744115
Gene Chromosome Location
Chr.2: 134918810-134959345 on Build GRCh38
Transcripts
NM_001241.3 NM_001320748.1 NM_001320749.1 NM_058241.2 NR_037649.1 NR_135468.1 XM_017005227.1 XM_017005228.1 XM_017005229.1 XM_017005230.1 XM_017005231.1 XM_017005232.1 XM_017005233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4498920 c.530C>G p.A177G Chr.2: 134946137-134946137 on Build GRCh38
5350703 c.494-7C>T p.? Chr.2: 134946094-134946094 on Build GRCh38
5949639 c.539+2T>C p.? Chr.2: 134946148-134946148 on Build GRCh38
4807682 c.516A>G p.T172T Chr.2: 134946123-134946123 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768830735 Chr.2: 134946097-134946110 on Build GRCh38
rs776362957 Chr.2: 134946186-134946186 on Build GRCh38
rs201814332 Chr.2: 134946080-134946081 on Build GRCh38
rs982316818 Chr.2: 134946155-134946155 on Build GRCh38
rs763861413 Chr.2: 134946055-134946055 on Build GRCh38
rs755142465 Chr.2: 134946098-134946098 on Build GRCh38
rs903899222 Chr.2: 134946263-134946263 on Build GRCh38
rs1040844125 Chr.2: 134946083-134946083 on Build GRCh38
rs141802303 Chr.2: 134946102-134946102 on Build GRCh38
rs765608310 Chr.2: 134946069-134946069 on Build GRCh38
rs1037172585 Chr.2: 134946247-134946247 on Build GRCh38
rs766416935 Chr.2: 134946094-134946094 on Build GRCh38
rs149994804 Chr.2: 134946351-134946351 on Build GRCh38
rs539026309 Chr.2: 134946060-134946060 on Build GRCh38
rs758435065 Chr.2: 134946090-134946090 on Build GRCh38
rs780838978 Chr.2: 134946016-134946016 on Build GRCh38
rs748976577 Chr.2: 134946103-134946103 on Build GRCh38
rs745766765 Chr.2: 134946133-134946133 on Build GRCh38
rs943573903 Chr.2: 134946059-134946059 on Build GRCh38
rs750339545 Chr.2: 134946056-134946056 on Build GRCh38
rs865871542 Chr.2: 134946391-134946391 on Build GRCh38
rs571972480 Chr.2: 134946081-134946082 on Build GRCh38
rs552829067 Chr.2: 134946240-134946240 on Build GRCh38
rs1048305068 Chr.2: 134946324-134946324 on Build GRCh38
rs930953548 Chr.2: 134946271-134946271 on Build GRCh38
rs571009014 Chr.2: 134946250-134946250 on Build GRCh38
rs778952569 Chr.2: 134946122-134946122 on Build GRCh38
rs923368644 Chr.2: 134946237-134946237 on Build GRCh38
rs749848722 Chr.2: 134946081-134946081 on Build GRCh38
rs750953850 Chr.2: 134946081-134946081 on Build GRCh38
rs772182336 Chr.2: 134946162-134946162 on Build GRCh38
rs776528368 Chr.2: 134946140-134946141 on Build GRCh38
rs796949280 Chr.2: 134946081-134946082 on Build GRCh38
rs181927424 Chr.2: 134946010-134946010 on Build GRCh38
rs750443027 Chr.2: 134946408-134946408 on Build GRCh38
rs751688594 Chr.2: 134946097-134946097 on Build GRCh38
rs746579444 Chr.2: 134946168-134946168 on Build GRCh38
rs369414426 Chr.2: 134946053-134946053 on Build GRCh38
rs1000520004 Chr.2: 134946368-134946368 on Build GRCh38
rs779694060 Chr.2: 134946164-134946164 on Build GRCh38
rs757096592 Chr.2: 134946121-134946121 on Build GRCh38
rs762247856 Chr.2: 134946062-134946062 on Build GRCh38
rs928114897 Chr.2: 134946225-134946225 on Build GRCh38
rs760948989 Chr.2: 134946094-134946094 on Build GRCh38
rs866903276 Chr.2: 134946118-134946118 on Build GRCh38
rs938476013 Chr.2: 134946226-134946226 on Build GRCh38
rs777113344 Chr.2: 134946080-134946080 on Build GRCh38
rs531147102 Chr.2: 134946176-134946176 on Build GRCh38

View Full Product Details