Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151411229-151411729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIF1
Entrez Gene ID
55183
Gene Name
replication timing regulatory factor 1
Gene Aliases
-
UniGene
Hs.536537
Gene Chromosome Location
Chr.2: 151409883-151534200 on Build GRCh38
Transcripts
NM_001177663.1 NM_001177664.1 NM_001177665.1 NM_018151.4 XM_005246665.3 XM_011511395.2 XM_017004422.1 XM_017004423.1 XM_017004424.1 XM_017004425.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1399895 c.130G>T p.E44* Chr.2: 151411285-151411285 on Build GRCh38
1007342 c.169T>C p.Y57H Chr.2: 151411324-151411324 on Build GRCh38
1007341 c.118G>T p.E40* Chr.2: 151411273-151411273 on Build GRCh38
6154237 c.149A>T p.E50V Chr.2: 151411304-151411304 on Build GRCh38
6766463 c.147T>C p.I49I Chr.2: 151411302-151411302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866307816 Chr.2: 151411491-151411491 on Build GRCh38
rs1046395954 Chr.2: 151411705-151411705 on Build GRCh38
rs921414847 Chr.2: 151411643-151411643 on Build GRCh38
rs146223898 Chr.2: 151411702-151411702 on Build GRCh38
rs143246235 Chr.2: 151411460-151411460 on Build GRCh38
rs375522651 Chr.2: 151411401-151411402 on Build GRCh38
rs900026265 Chr.2: 151411443-151411443 on Build GRCh38
rs770381917 Chr.2: 151411283-151411285 on Build GRCh38
rs528981890 Chr.2: 151411397-151411397 on Build GRCh38
rs544135831 Chr.2: 151411506-151411506 on Build GRCh38
rs912856216 Chr.2: 151411360-151411360 on Build GRCh38
rs371737740 Chr.2: 151411357-151411357 on Build GRCh38
rs745420565 Chr.2: 151411289-151411289 on Build GRCh38
rs773889833 Chr.2: 151411348-151411351 on Build GRCh38
rs760002980 Chr.2: 151411361-151411362 on Build GRCh38
rs914783776 Chr.2: 151411551-151411551 on Build GRCh38
rs933336345 Chr.2: 151411596-151411596 on Build GRCh38
rs995288799 Chr.2: 151411465-151411465 on Build GRCh38
rs951118487 Chr.2: 151411573-151411573 on Build GRCh38
rs768068288 Chr.2: 151411384-151411384 on Build GRCh38
rs919714642 Chr.2: 151411541-151411541 on Build GRCh38
rs970150799 Chr.2: 151411476-151411476 on Build GRCh38
rs773307697 Chr.2: 151411341-151411341 on Build GRCh38
rs201379519 Chr.2: 151411395-151411395 on Build GRCh38
rs371134498 Chr.2: 151411628-151411628 on Build GRCh38
rs944298348 Chr.2: 151411406-151411406 on Build GRCh38
rs974209384 Chr.2: 151411512-151411512 on Build GRCh38
rs202100792 Chr.2: 151411328-151411328 on Build GRCh38
rs377188695 Chr.2: 151411316-151411316 on Build GRCh38
rs184378944 Chr.2: 151411632-151411632 on Build GRCh38
rs758873382 Chr.2: 151411308-151411308 on Build GRCh38
rs1020209887 Chr.2: 151411471-151411471 on Build GRCh38
rs775585377 Chr.2: 151411351-151411351 on Build GRCh38
rs762483460 Chr.2: 151411272-151411274 on Build GRCh38
rs753634523 Chr.2: 151411363-151411363 on Build GRCh38
rs749897553 Chr.2: 151411379-151411379 on Build GRCh38
rs944267782 Chr.2: 151411621-151411621 on Build GRCh38
rs1054273280 Chr.2: 151411676-151411676 on Build GRCh38
rs763152702 Chr.2: 151411342-151411342 on Build GRCh38
rs530181892 Chr.2: 151411454-151411455 on Build GRCh38
rs542991314 Chr.2: 151411665-151411665 on Build GRCh38
rs147741381 Chr.2: 151411315-151411315 on Build GRCh38
rs776824025 Chr.2: 151411461-151411461 on Build GRCh38
rs147405197 Chr.2: 151411383-151411384 on Build GRCh38
rs145765490 Chr.2: 151411339-151411339 on Build GRCh38
rs1001333592 Chr.2: 151411452-151411452 on Build GRCh38
rs374684225 Chr.2: 151411394-151411395 on Build GRCh38
rs537429388 Chr.2: 151411472-151411472 on Build GRCh38
rs892589459 Chr.2: 151411684-151411684 on Build GRCh38
rs541267077 Chr.2: 151411620-151411620 on Build GRCh38
rs936884335 Chr.2: 151411670-151411670 on Build GRCh38
rs1050995137 Chr.2: 151411608-151411608 on Build GRCh38
rs756912315 Chr.2: 151411369-151411369 on Build GRCh38
rs890939199 Chr.2: 151411408-151411408 on Build GRCh38
rs1036857362 Chr.2: 151411700-151411700 on Build GRCh38
rs557422122 Chr.2: 151411477-151411477 on Build GRCh38
rs572099346 Chr.2: 151411601-151411601 on Build GRCh38
rs764839243 Chr.2: 151411375-151411375 on Build GRCh38
rs747761997 Chr.2: 151411268-151411268 on Build GRCh38
rs780572826 Chr.2: 151411310-151411310 on Build GRCh38
rs763949243 Chr.2: 151411361-151411361 on Build GRCh38
rs566104276 Chr.2: 151411432-151411432 on Build GRCh38
rs372832713 Chr.2: 151411277-151411277 on Build GRCh38
rs563027164 Chr.2: 151411385-151411385 on Build GRCh38
rs891044762 Chr.2: 151411626-151411626 on Build GRCh38
rs567687651 Chr.2: 151411503-151411503 on Build GRCh38
rs996763843 Chr.2: 151411390-151411401 on Build GRCh38
rs921938138 Chr.2: 151411587-151411587 on Build GRCh38
rs1039816522 Chr.2: 151411438-151411438 on Build GRCh38
rs778473583 Chr.2: 151411283-151411283 on Build GRCh38
rs369094048 Chr.2: 151411345-151411345 on Build GRCh38
rs572014369 Chr.2: 151411468-151411468 on Build GRCh38
rs549082186 Chr.2: 151411402-151411402 on Build GRCh38
rs772428196 Chr.2: 151411496-151411496 on Build GRCh38
rs1047289023 Chr.2: 151411332-151411332 on Build GRCh38
rs756931357 Chr.2: 151411280-151411280 on Build GRCh38
rs528691419 Chr.2: 151411459-151411459 on Build GRCh38
rs763598295 Chr.2: 151411272-151411272 on Build GRCh38
rs1009605579 Chr.2: 151411466-151411466 on Build GRCh38
rs982376231 Chr.2: 151411580-151411580 on Build GRCh38
rs956171657 Chr.2: 151411538-151411538 on Build GRCh38
rs754070888 Chr.2: 151411384-151411385 on Build GRCh38
rs553443431 Chr.2: 151411547-151411547 on Build GRCh38
rs556028488 Chr.2: 151411583-151411583 on Build GRCh38
rs577550362 Chr.2: 151411634-151411634 on Build GRCh38

View Full Product Details