Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 152759669-152760183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL6IP6
Entrez Gene ID
151188
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 6
Gene Aliases
AIP-6, AIP6, PFAAP1
UniGene
Hs.643580
Gene Chromosome Location
Chr.2: 152717893-152761253 on Build GRCh38
Transcripts
NM_152522.5 NR_024526.2 XM_006712326.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
441266 c.638A>C p.N213T Chr.2: 152759797-152759797 on Build GRCh38
6205630 c.666A>C p.A222A Chr.2: 152759825-152759825 on Build GRCh38
7198206 c.596C>G p.T199S Chr.2: 152759755-152759755 on Build GRCh38
1007915 c.616G>A p.G206S Chr.2: 152759775-152759775 on Build GRCh38
4136092 c.636G>C p.L212F Chr.2: 152759795-152759795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559707304 Chr.2: 152759855-152759855 on Build GRCh38
rs888189470 Chr.2: 152760122-152760122 on Build GRCh38
rs371666118 Chr.2: 152759805-152759805 on Build GRCh38
rs1044174056 Chr.2: 152760113-152760113 on Build GRCh38
rs772868616 Chr.2: 152759846-152759846 on Build GRCh38
rs934527396 Chr.2: 152760051-152760051 on Build GRCh38
rs532701668 Chr.2: 152759986-152759986 on Build GRCh38
rs990739116 Chr.2: 152760056-152760056 on Build GRCh38
rs746863004 Chr.2: 152759745-152759745 on Build GRCh38
rs144699421 Chr.2: 152759798-152759798 on Build GRCh38
rs753053720 Chr.2: 152759808-152759808 on Build GRCh38
rs781163954 Chr.2: 152759838-152759838 on Build GRCh38
rs950592441 Chr.2: 152759989-152759989 on Build GRCh38
rs766015346 Chr.2: 152759750-152759753 on Build GRCh38
rs781347653 Chr.2: 152759722-152759722 on Build GRCh38
rs377486862 Chr.2: 152759703-152759703 on Build GRCh38
rs569395528 Chr.2: 152760109-152760109 on Build GRCh38
rs774647971 Chr.2: 152759789-152759789 on Build GRCh38
rs964682311 Chr.2: 152759922-152759922 on Build GRCh38
rs745953973 Chr.2: 152759825-152759825 on Build GRCh38
rs755387049 Chr.2: 152759715-152759715 on Build GRCh38
rs367604437 Chr.2: 152759724-152759724 on Build GRCh38
rs769535236 Chr.2: 152759845-152759845 on Build GRCh38
rs963261428 Chr.2: 152760049-152760049 on Build GRCh38
rs113883448 Chr.2: 152759741-152759742 on Build GRCh38
rs780162517 Chr.2: 152759743-152759743 on Build GRCh38
rs776026049 Chr.2: 152759871-152759871 on Build GRCh38
rs747930229 Chr.2: 152759842-152759842 on Build GRCh38
rs764935873 Chr.2: 152759702-152759702 on Build GRCh38
rs773002096 Chr.2: 152759735-152759735 on Build GRCh38
rs1016555258 Chr.2: 152759756-152759756 on Build GRCh38
rs968622711 Chr.2: 152759936-152759936 on Build GRCh38
rs761143948 Chr.2: 152759877-152759877 on Build GRCh38
rs998005319 Chr.2: 152759945-152759945 on Build GRCh38
rs773758163 Chr.2: 152759777-152759778 on Build GRCh38
rs1011799336 Chr.2: 152759854-152759854 on Build GRCh38
rs916888229 Chr.2: 152760081-152760081 on Build GRCh38
rs983327166 Chr.2: 152760010-152760010 on Build GRCh38
rs761653552 Chr.2: 152759777-152759777 on Build GRCh38
rs368411477 Chr.2: 152759862-152759862 on Build GRCh38
rs761733100 Chr.2: 152759708-152759709 on Build GRCh38
rs776353229 Chr.2: 152759758-152759758 on Build GRCh38
rs948374891 Chr.2: 152760110-152760110 on Build GRCh38
rs562060847 Chr.2: 152759726-152759727 on Build GRCh38
rs544421451 Chr.2: 152760116-152760116 on Build GRCh38
rs916262310 Chr.2: 152759707-152759707 on Build GRCh38
rs748093120 Chr.2: 152760015-152760015 on Build GRCh38
rs768837570 Chr.2: 152759725-152759725 on Build GRCh38
rs112189535 Chr.2: 152760033-152760033 on Build GRCh38
rs779067094 Chr.2: 152759824-152759824 on Build GRCh38
rs758724708 Chr.2: 152760006-152760006 on Build GRCh38
rs373963152 Chr.2: 152759823-152759823 on Build GRCh38
rs753105044 Chr.2: 152759723-152759723 on Build GRCh38
rs762517055 Chr.2: 152759735-152759736 on Build GRCh38
rs769480444 Chr.2: 152759784-152759784 on Build GRCh38
rs776356898 Chr.2: 152759700-152759700 on Build GRCh38
rs754925855 Chr.2: 152759835-152759835 on Build GRCh38
rs370972436 Chr.2: 152759704-152759704 on Build GRCh38
rs374346170 Chr.2: 152759706-152759706 on Build GRCh38
rs768501444 Chr.2: 152759750-152759750 on Build GRCh38
rs898073942 Chr.2: 152759769-152759769 on Build GRCh38
rs768377601 Chr.2: 152759727-152759727 on Build GRCh38
rs879514252 Chr.2: 152760090-152760090 on Build GRCh38
rs773794941 Chr.2: 152759859-152759859 on Build GRCh38
rs142877091 Chr.2: 152759804-152759804 on Build GRCh38
rs954807539 Chr.2: 152759860-152759860 on Build GRCh38
rs528639824 Chr.2: 152760105-152760105 on Build GRCh38
rs113218221 Chr.2: 152759887-152759887 on Build GRCh38
rs748877617 Chr.2: 152759852-152759852 on Build GRCh38
rs767952131 Chr.2: 152759866-152759866 on Build GRCh38
rs752016361 Chr.2: 152759710-152759710 on Build GRCh38
rs573859223 Chr.2: 152760115-152760115 on Build GRCh38
rs778173653 Chr.2: 152759809-152759809 on Build GRCh38
rs771095178 Chr.2: 152759735-152759735 on Build GRCh38
rs552788162 Chr.2: 152760018-152760018 on Build GRCh38
rs1018087047 Chr.2: 152759911-152759911 on Build GRCh38
rs909064347 Chr.2: 152760014-152760017 on Build GRCh38
rs978996513 Chr.2: 152759979-152759979 on Build GRCh38
rs992510279 Chr.2: 152760075-152760075 on Build GRCh38

View Full Product Details