Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159780067-159780607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD302
Entrez Gene ID
9936
Gene Name
CD302 molecule
Gene Aliases
BIMLEC, CLEC13A, DCL-1, DCL1
UniGene
Hs.130014
Gene Chromosome Location
Chr.2: 159768628-159798255 on Build GRCh38
Transcripts
NM_001198763.1 NM_001198764.1 NM_014880.4 NM_001198759.1 NM_001198760.1

Gene Symbol
LY75-CD302
Entrez Gene ID
100526664
Gene Name
LY75-CD302 readthrough
Gene Aliases
-
UniGene
Hs.153563
Gene Chromosome Location
Chr.2: 159768628-159904756 on Build GRCh38
Transcripts
NM_001198763.1 NM_001198764.1 NM_014880.4 NM_001198759.1 NM_001198760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3743932 c.391C>G p.L131V Chr.2: 159780083-159780083 on Build GRCh38
476090 c.391C>A p.L131M Chr.2: 159780083-159780083 on Build GRCh38
3300386 c.364G>A p.E122K Chr.2: 159780110-159780110 on Build GRCh38
1400360 c.335T>G p.F112C Chr.2: 159780139-159780139 on Build GRCh38
1008180 c.300G>A p.A100A Chr.2: 159780174-159780174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950767143 Chr.2: 159780556-159780556 on Build GRCh38
rs183106874 Chr.2: 159780467-159780467 on Build GRCh38
rs771264039 Chr.2: 159780549-159780549 on Build GRCh38
rs756081226 Chr.2: 159780172-159780172 on Build GRCh38
rs755032369 Chr.2: 159780117-159780117 on Build GRCh38
rs182350513 Chr.2: 159780216-159780216 on Build GRCh38
rs200960588 Chr.2: 159780199-159780199 on Build GRCh38
rs771576401 Chr.2: 159780209-159780209 on Build GRCh38
rs146987171 Chr.2: 159780174-159780174 on Build GRCh38
rs754295690 Chr.2: 159780119-159780119 on Build GRCh38
rs757142786 Chr.2: 159780221-159780223 on Build GRCh38
rs992626516 Chr.2: 159780169-159780169 on Build GRCh38
rs138157162 Chr.2: 159780390-159780390 on Build GRCh38
rs1052836678 Chr.2: 159780391-159780391 on Build GRCh38
rs920661426 Chr.2: 159780098-159780098 on Build GRCh38
rs905529919 Chr.2: 159780459-159780459 on Build GRCh38
rs866342565 Chr.2: 159780356-159780356 on Build GRCh38
rs113204764 Chr.2: 159780521-159780521 on Build GRCh38
rs983479305 Chr.2: 159780343-159780343 on Build GRCh38
rs1048187484 Chr.2: 159780136-159780136 on Build GRCh38
rs1033950477 Chr.2: 159780514-159780514 on Build GRCh38
rs548907383 Chr.2: 159780456-159780456 on Build GRCh38
rs943239151 Chr.2: 159780331-159780331 on Build GRCh38
rs909223695 Chr.2: 159780297-159780297 on Build GRCh38
rs946609860 Chr.2: 159780419-159780420 on Build GRCh38
rs146355410 Chr.2: 159780567-159780567 on Build GRCh38
rs780202336 Chr.2: 159780157-159780157 on Build GRCh38
rs141287355 Chr.2: 159780121-159780121 on Build GRCh38
rs200600963 Chr.2: 159780441-159780442 on Build GRCh38
rs559817677 Chr.2: 159780546-159780547 on Build GRCh38
rs973960964 Chr.2: 159780295-159780295 on Build GRCh38
rs753792338 Chr.2: 159780135-159780139 on Build GRCh38
rs758507302 Chr.2: 159780155-159780155 on Build GRCh38
rs34844327 Chr.2: 159780442-159780442 on Build GRCh38
rs1023103919 Chr.2: 159780266-159780266 on Build GRCh38
rs960285723 Chr.2: 159780255-159780255 on Build GRCh38
rs779065808 Chr.2: 159780125-159780125 on Build GRCh38
rs189086687 Chr.2: 159780265-159780265 on Build GRCh38
rs1011239982 Chr.2: 159780437-159780437 on Build GRCh38
rs964752520 Chr.2: 159780573-159780573 on Build GRCh38
rs34439472 Chr.2: 159780441-159780441 on Build GRCh38
rs749138371 Chr.2: 159780175-159780175 on Build GRCh38
rs376800091 Chr.2: 159780379-159780381 on Build GRCh38
rs1011143117 Chr.2: 159780201-159780201 on Build GRCh38
rs201544040 Chr.2: 159780111-159780111 on Build GRCh38
rs750674228 Chr.2: 159780149-159780149 on Build GRCh38
rs950852552 Chr.2: 159780319-159780319 on Build GRCh38
rs747066009 Chr.2: 159780162-159780162 on Build GRCh38
rs1013731649 Chr.2: 159780524-159780524 on Build GRCh38
rs191839959 Chr.2: 159780450-159780450 on Build GRCh38
rs747239894 Chr.2: 159780360-159780360 on Build GRCh38
rs901643541 Chr.2: 159780378-159780378 on Build GRCh38
rs984841775 Chr.2: 159780566-159780566 on Build GRCh38
rs545644627 Chr.2: 159780315-159780315 on Build GRCh38
rs766747478 Chr.2: 159780325-159780325 on Build GRCh38
rs66740317 Chr.2: 159780442-159780443 on Build GRCh38
rs532518878 Chr.2: 159780274-159780274 on Build GRCh38
rs745776472 Chr.2: 159780108-159780108 on Build GRCh38
rs149145819 Chr.2: 159780498-159780498 on Build GRCh38
rs902258486 Chr.2: 159780217-159780217 on Build GRCh38
rs770771531 Chr.2: 159780179-159780179 on Build GRCh38
rs999234468 Chr.2: 159780242-159780242 on Build GRCh38
rs911845621 Chr.2: 159780141-159780141 on Build GRCh38
rs767523154 Chr.2: 159780109-159780111 on Build GRCh38
rs920727064 Chr.2: 159780318-159780318 on Build GRCh38
rs773948445 Chr.2: 159780181-159780181 on Build GRCh38
rs3771718 Chr.2: 159780397-159780397 on Build GRCh38
rs758434233 Chr.2: 159780463-159780465 on Build GRCh38
rs944623569 Chr.2: 159780377-159780377 on Build GRCh38
rs1038940176 Chr.2: 159780385-159780385 on Build GRCh38
rs187609856 Chr.2: 159780571-159780571 on Build GRCh38
rs546218440 Chr.2: 159780518-159780518 on Build GRCh38
rs1034616569 Chr.2: 159780247-159780247 on Build GRCh38

View Full Product Details