Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168908013-168908520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC2
Entrez Gene ID
57818
Gene Name
glucose-6-phosphatase catalytic subunit 2
Gene Aliases
IGRP
UniGene
Hs.283963
Gene Chromosome Location
Chr.2: 168901223-168910000 on Build GRCh38
Transcripts
NM_001081686.1 NM_021176.2 XM_011511564.2 XM_011511565.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
229689 c.1019C>A p.P340H Chr.2: 168908030-168908030 on Build GRCh38
3569605 c.1018C>T p.P340S Chr.2: 168908029-168908029 on Build GRCh38
4686023 c.1050C>T p.S350S Chr.2: 168908061-168908061 on Build GRCh38
7089972 c.1020C>A p.P340P Chr.2: 168908031-168908031 on Build GRCh38
1009110 c.1014C>A p.F338L Chr.2: 168908025-168908025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758997713 Chr.2: 168908094-168908094 on Build GRCh38
rs968524053 Chr.2: 168908234-168908234 on Build GRCh38
rs924000489 Chr.2: 168908266-168908266 on Build GRCh38
rs1046786155 Chr.2: 168908154-168908154 on Build GRCh38
rs780589028 Chr.2: 168908095-168908095 on Build GRCh38
rs750777548 Chr.2: 168908088-168908088 on Build GRCh38
rs2947933 Chr.2: 168908438-168908438 on Build GRCh38
rs754077966 Chr.2: 168908061-168908061 on Build GRCh38
rs757496063 Chr.2: 168908063-168908063 on Build GRCh38
rs878878851 Chr.2: 168908229-168908229 on Build GRCh38
rs78164968 Chr.2: 168908251-168908251 on Build GRCh38
rs2947932 Chr.2: 168908402-168908402 on Build GRCh38
rs3770570 Chr.2: 168908279-168908279 on Build GRCh38
rs2947931 Chr.2: 168908395-168908395 on Build GRCh38
rs1002543896 Chr.2: 168908467-168908467 on Build GRCh38
rs942994717 Chr.2: 168908373-168908373 on Build GRCh38
rs886824546 Chr.2: 168908464-168908464 on Build GRCh38
rs189856873 Chr.2: 168908126-168908126 on Build GRCh38
rs548273066 Chr.2: 168908181-168908181 on Build GRCh38
rs958831698 Chr.2: 168908190-168908190 on Build GRCh38
rs760776009 Chr.2: 168908055-168908055 on Build GRCh38
rs369261389 Chr.2: 168908122-168908122 on Build GRCh38
rs896612169 Chr.2: 168908376-168908376 on Build GRCh38
rs910487070 Chr.2: 168908242-168908242 on Build GRCh38
rs2232328 Chr.2: 168908036-168908036 on Build GRCh38
rs929043226 Chr.2: 168908354-168908354 on Build GRCh38
rs140247430 Chr.2: 168908233-168908233 on Build GRCh38
rs11554658 Chr.2: 168908044-168908044 on Build GRCh38
rs777732226 Chr.2: 168908084-168908084 on Build GRCh38
rs2232329 Chr.2: 168908106-168908106 on Build GRCh38
rs528505004 Chr.2: 168908177-168908177 on Build GRCh38
rs1039925877 Chr.2: 168908389-168908389 on Build GRCh38
rs1037354711 Chr.2: 168908243-168908243 on Build GRCh38
rs553336703 Chr.2: 168908403-168908403 on Build GRCh38
rs768454510 Chr.2: 168908068-168908069 on Build GRCh38
rs376540601 Chr.2: 168908118-168908118 on Build GRCh38
rs746860504 Chr.2: 168908349-168908349 on Build GRCh38
rs765702651 Chr.2: 168908075-168908075 on Build GRCh38
rs769183844 Chr.2: 168908448-168908448 on Build GRCh38
rs987173277 Chr.2: 168908348-168908348 on Build GRCh38
rs1013718370 Chr.2: 168908428-168908428 on Build GRCh38
rs1023818908 Chr.2: 168908450-168908450 on Build GRCh38
rs550514595 Chr.2: 168908192-168908192 on Build GRCh38
rs367851926 Chr.2: 168908060-168908060 on Build GRCh38
rs993287550 Chr.2: 168908046-168908046 on Build GRCh38
rs559301659 Chr.2: 168908146-168908146 on Build GRCh38
rs151134573 Chr.2: 168908183-168908183 on Build GRCh38
rs1046823054 Chr.2: 168908462-168908462 on Build GRCh38
rs979532744 Chr.2: 168908239-168908239 on Build GRCh38
rs771392103 Chr.2: 168908210-168908210 on Build GRCh38
rs746746462 Chr.2: 168908067-168908067 on Build GRCh38
rs34631431 Chr.2: 168908452-168908452 on Build GRCh38
rs941654712 Chr.2: 168908242-168908243 on Build GRCh38
rs368183167 Chr.2: 168908303-168908303 on Build GRCh38
rs142220775 Chr.2: 168908182-168908182 on Build GRCh38
rs866965940 Chr.2: 168908325-168908325 on Build GRCh38
rs1032327978 Chr.2: 168908132-168908132 on Build GRCh38
rs2632369 Chr.2: 168908213-168908213 on Build GRCh38
rs775478622 Chr.2: 168908051-168908051 on Build GRCh38
rs371472092 Chr.2: 168908454-168908454 on Build GRCh38
rs762906384 Chr.2: 168908041-168908041 on Build GRCh38
rs754509276 Chr.2: 168908110-168908110 on Build GRCh38
rs766251315 Chr.2: 168908043-168908043 on Build GRCh38
rs779799830 Chr.2: 168908331-168908331 on Build GRCh38
rs886802335 Chr.2: 168908174-168908174 on Build GRCh38
rs756530700 Chr.2: 168908073-168908073 on Build GRCh38
rs923094802 Chr.2: 168908191-168908191 on Build GRCh38
rs894131081 Chr.2: 168908491-168908491 on Build GRCh38
rs555457669 Chr.2: 168908484-168908484 on Build GRCh38

View Full Product Details