Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169081029-169081535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS9
Entrez Gene ID
10170
Gene Name
dehydrogenase/reductase 9
Gene Aliases
3-alpha-HSD, 3ALPHA-HSD, RDH-E2, RDH-TBE, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4
UniGene
Hs.179608
Gene Chromosome Location
Chr.2: 169067035-169096167 on Build GRCh38
Transcripts
NM_001142270.1 NM_001142271.1 NM_001289763.1 NM_199204.1 XM_011510475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6437500 c.134T>C p.L45P Chr.2: 169081535-169081535 on Build GRCh38
3569702 c.126C>T p.H42H Chr.2: 169081527-169081527 on Build GRCh38
5961179 c.133C>T p.L45F Chr.2: 169081534-169081534 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs574665131 Chr.2: 169081273-169081273 on Build GRCh38
rs986015573 Chr.2: 169081098-169081098 on Build GRCh38
rs552407862 Chr.2: 169081255-169081255 on Build GRCh38
rs577115311 Chr.2: 169081413-169081413 on Build GRCh38
rs530215580 Chr.2: 169081505-169081505 on Build GRCh38
rs890274391 Chr.2: 169081423-169081423 on Build GRCh38
rs141571021 Chr.2: 169081467-169081467 on Build GRCh38
rs892568748 Chr.2: 169081143-169081143 on Build GRCh38
rs996820892 Chr.2: 169081213-169081213 on Build GRCh38
rs765726632 Chr.2: 169081102-169081102 on Build GRCh38
rs992981870 Chr.2: 169081339-169081339 on Build GRCh38
rs183965693 Chr.2: 169081164-169081164 on Build GRCh38
rs755182120 Chr.2: 169081341-169081341 on Build GRCh38
rs959802987 Chr.2: 169081265-169081265 on Build GRCh38
rs934556402 Chr.2: 169081371-169081371 on Build GRCh38
rs555126171 Chr.2: 169081234-169081234 on Build GRCh38
rs545782441 Chr.2: 169081448-169081448 on Build GRCh38
rs1008247673 Chr.2: 169081154-169081154 on Build GRCh38
rs76168410 Chr.2: 169081259-169081259 on Build GRCh38
rs558883196 Chr.2: 169081457-169081457 on Build GRCh38
rs556967984 Chr.2: 169081365-169081365 on Build GRCh38
rs952910331 Chr.2: 169081241-169081241 on Build GRCh38
rs770890539 Chr.2: 169081475-169081475 on Build GRCh38
rs566081001 Chr.2: 169081197-169081197 on Build GRCh38
rs552586082 Chr.2: 169081190-169081190 on Build GRCh38
rs550184144 Chr.2: 169081087-169081087 on Build GRCh38
rs541496413 Chr.2: 169081492-169081492 on Build GRCh38
rs1027659436 Chr.2: 169081231-169081231 on Build GRCh38
rs73026859 Chr.2: 169081198-169081198 on Build GRCh38
rs1042697096 Chr.2: 169081355-169081355 on Build GRCh38
rs1030256248 Chr.2: 169081075-169081075 on Build GRCh38
rs780132884 Chr.2: 169081191-169081191 on Build GRCh38
rs538646336 Chr.2: 169081171-169081171 on Build GRCh38
rs561474045 Chr.2: 169081499-169081499 on Build GRCh38
rs1052907670 Chr.2: 169081400-169081400 on Build GRCh38
rs537287534 Chr.2: 169081349-169081349 on Build GRCh38
rs903773999 Chr.2: 169081316-169081316 on Build GRCh38
rs751861272 Chr.2: 169081253-169081253 on Build GRCh38
rs910175657 Chr.2: 169081101-169081101 on Build GRCh38
rs901478979 Chr.2: 169081375-169081375 on Build GRCh38
rs1043940616 Chr.2: 169081277-169081277 on Build GRCh38
rs962924475 Chr.2: 169081140-169081140 on Build GRCh38
rs972719364 Chr.2: 169081163-169081163 on Build GRCh38
rs750907322 Chr.2: 169081130-169081130 on Build GRCh38
rs530338270 Chr.2: 169081062-169081062 on Build GRCh38
rs916631760 Chr.2: 169081235-169081235 on Build GRCh38

View Full Product Details