Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 186635866-186636416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGAV
Entrez Gene ID
3685
Gene Name
integrin subunit alpha V
Gene Aliases
CD51, MSK8, VNRA, VTNR
UniGene
Hs.436873
Gene Chromosome Location
Chr.2: 186590063-186680902 on Build GRCh38
Transcripts
NM_001144999.2 NM_001145000.2 NM_002210.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
476643 c.723G>A p.R241R Chr.2: 186636173-186636173 on Build GRCh38
4171129 c.554G>A p.S185N Chr.2: 186636142-186636142 on Build GRCh38
6688099 c.632-8delT p.? Chr.2: 186636074-186636074 on Build GRCh38
3575179 c.525C>T p.I175I Chr.2: 186636113-186636113 on Build GRCh38
4171128 c.692G>A p.S231N Chr.2: 186636142-186636142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772289319 Chr.2: 186636292-186636292 on Build GRCh38
rs755379590 Chr.2: 186636170-186636170 on Build GRCh38
rs775092026 Chr.2: 186636126-186636126 on Build GRCh38
rs200647242 Chr.2: 186636114-186636114 on Build GRCh38
rs376680231 Chr.2: 186636172-186636172 on Build GRCh38
rs3795865 Chr.2: 186635955-186635955 on Build GRCh38
rs778900176 Chr.2: 186636210-186636210 on Build GRCh38
rs200818475 Chr.2: 186636219-186636219 on Build GRCh38
rs372449821 Chr.2: 186636171-186636171 on Build GRCh38
rs958208845 Chr.2: 186636010-186636010 on Build GRCh38
rs868199933 Chr.2: 186636143-186636143 on Build GRCh38
rs907177773 Chr.2: 186635949-186635949 on Build GRCh38
rs200674397 Chr.2: 186636133-186636133 on Build GRCh38
rs760398503 Chr.2: 186636129-186636129 on Build GRCh38
rs1045775440 Chr.2: 186635935-186635935 on Build GRCh38
rs1006385905 Chr.2: 186635976-186635976 on Build GRCh38
rs976274379 Chr.2: 186636006-186636006 on Build GRCh38
rs1026907883 Chr.2: 186635917-186635917 on Build GRCh38
rs755459803 Chr.2: 186636064-186636064 on Build GRCh38
rs894436054 Chr.2: 186635898-186635898 on Build GRCh38
rs1045638313 Chr.2: 186636269-186636269 on Build GRCh38
rs774086562 Chr.2: 186635968-186635968 on Build GRCh38
rs761237786 Chr.2: 186636094-186636094 on Build GRCh38
rs149545302 Chr.2: 186636125-186636125 on Build GRCh38
rs1006251713 Chr.2: 186636379-186636379 on Build GRCh38
rs572216722 Chr.2: 186635982-186635982 on Build GRCh38
rs928635778 Chr.2: 186636326-186636326 on Build GRCh38
rs200437349 Chr.2: 186636036-186636036 on Build GRCh38
rs749008407 Chr.2: 186636265-186636265 on Build GRCh38
rs756467330 Chr.2: 186636182-186636182 on Build GRCh38
rs900838934 Chr.2: 186636251-186636257 on Build GRCh38
rs1039192288 Chr.2: 186636381-186636381 on Build GRCh38
rs958176366 Chr.2: 186636380-186636380 on Build GRCh38
rs60491688 Chr.2: 186636359-186636359 on Build GRCh38
rs201504027 Chr.2: 186636103-186636103 on Build GRCh38
rs774333133 Chr.2: 186636116-186636116 on Build GRCh38
rs748413227 Chr.2: 186636090-186636090 on Build GRCh38
rs536208489 Chr.2: 186636347-186636347 on Build GRCh38
rs575317661 Chr.2: 186635963-186635963 on Build GRCh38
rs61765168 Chr.2: 186636223-186636223 on Build GRCh38
rs752096729 Chr.2: 186636054-186636054 on Build GRCh38
rs761467033 Chr.2: 186636146-186636146 on Build GRCh38
rs200856961 Chr.2: 186636101-186636101 on Build GRCh38
rs139119479 Chr.2: 186636200-186636200 on Build GRCh38
rs138081971 Chr.2: 186635911-186635911 on Build GRCh38
rs897025900 Chr.2: 186635896-186635896 on Build GRCh38
rs367683819 Chr.2: 186636142-186636142 on Build GRCh38
rs999250831 Chr.2: 186636278-186636278 on Build GRCh38
rs759468300 Chr.2: 186636121-186636121 on Build GRCh38
rs781341683 Chr.2: 186636086-186636086 on Build GRCh38
rs201616731 Chr.2: 186636173-186636173 on Build GRCh38
rs757397558 Chr.2: 186636207-186636207 on Build GRCh38
rs777936327 Chr.2: 186636095-186636095 on Build GRCh38
rs1023787801 Chr.2: 186636389-186636389 on Build GRCh38
rs746828061 Chr.2: 186636224-186636224 on Build GRCh38
rs199724726 Chr.2: 186636132-186636132 on Build GRCh38
rs969659547 Chr.2: 186636391-186636391 on Build GRCh38
rs764832885 Chr.2: 186636157-186636157 on Build GRCh38
rs756796092 Chr.2: 186636040-186636040 on Build GRCh38
rs763674292 Chr.2: 186636035-186636035 on Build GRCh38
rs1019729435 Chr.2: 186635961-186635961 on Build GRCh38
rs150746738 Chr.2: 186636158-186636158 on Build GRCh38
rs777910458 Chr.2: 186636187-186636187 on Build GRCh38
rs780755475 Chr.2: 186636050-186636050 on Build GRCh38
rs532651553 Chr.2: 186636327-186636327 on Build GRCh38
rs929357667 Chr.2: 186636339-186636339 on Build GRCh38
rs199813220 Chr.2: 186636113-186636113 on Build GRCh38
rs756635013 Chr.2: 186636218-186636218 on Build GRCh38
rs761916404 Chr.2: 186635960-186635960 on Build GRCh38
rs989663087 Chr.2: 186636021-186636021 on Build GRCh38
rs752959691 Chr.2: 186636178-186636178 on Build GRCh38
rs964962710 Chr.2: 186635985-186635985 on Build GRCh38

View Full Product Details