Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200440507-200441018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATS2L
Entrez Gene ID
26010
Gene Name
spermatogenesis associated serine rich 2 like
Gene Aliases
DNAPTP6, SGNP
UniGene
Hs.120323
Gene Chromosome Location
Chr.2: 200305881-200482263 on Build GRCh38
Transcripts
NM_001100422.1 NM_001100423.1 NM_001100424.1 NM_001282735.1 NM_001282743.1 NM_001282744.1 NM_015535.2 XM_005246456.1 XM_005246457.1 XM_005246458.1 XM_005246459.1 XM_011510938.1 XM_011510939.1 XM_017003783.1 XM_017003784.1 XM_017003785.1 XM_017003786.1 XM_017003787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7261213 c.780C>T p.R260R Chr.2: 200440686-200440686 on Build GRCh38
1404393 c.787G>A p.V263I Chr.2: 200440693-200440693 on Build GRCh38
2906292 c.717C>T p.R239R Chr.2: 200440713-200440713 on Build GRCh38
2906295 c.780A>T p.L260F Chr.2: 200440776-200440776 on Build GRCh38
2906294 c.717C>T p.R239R Chr.2: 200440713-200440713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779683408 Chr.2: 200440919-200440919 on Build GRCh38
rs776723156 Chr.2: 200440819-200440819 on Build GRCh38
rs370727542 Chr.2: 200440693-200440693 on Build GRCh38
rs765351636 Chr.2: 200440714-200440714 on Build GRCh38
rs199514057 Chr.2: 200440774-200440774 on Build GRCh38
rs923823839 Chr.2: 200440993-200440993 on Build GRCh38
rs752867720 Chr.2: 200440715-200440715 on Build GRCh38
rs370049321 Chr.2: 200440743-200440743 on Build GRCh38
rs563497956 Chr.2: 200440892-200440892 on Build GRCh38
rs879126571 Chr.2: 200440639-200440639 on Build GRCh38
rs747784161 Chr.2: 200440777-200440777 on Build GRCh38
rs758608580 Chr.2: 200440717-200440717 on Build GRCh38
rs778323726 Chr.2: 200440757-200440757 on Build GRCh38
rs779591064 Chr.2: 200440638-200440638 on Build GRCh38
rs962249976 Chr.2: 200440674-200440674 on Build GRCh38
rs746786378 Chr.2: 200440799-200440799 on Build GRCh38
rs573137209 Chr.2: 200440614-200440614 on Build GRCh38
rs767873809 Chr.2: 200440629-200440629 on Build GRCh38
rs993806310 Chr.2: 200440925-200440925 on Build GRCh38
rs757748453 Chr.2: 200440725-200440725 on Build GRCh38
rs530801530 Chr.2: 200440930-200440930 on Build GRCh38
rs375661298 Chr.2: 200440721-200440721 on Build GRCh38
rs113069017 Chr.2: 200440814-200440814 on Build GRCh38
rs201830093 Chr.2: 200440828-200440828 on Build GRCh38
rs766395304 Chr.2: 200440695-200440695 on Build GRCh38
rs753171419 Chr.2: 200440607-200440607 on Build GRCh38
rs757736783 Chr.2: 200440617-200440617 on Build GRCh38
rs562876187 Chr.2: 200440744-200440744 on Build GRCh38
rs753069675 Chr.2: 200440711-200440712 on Build GRCh38
rs923884789 Chr.2: 200440707-200440707 on Build GRCh38
rs772927866 Chr.2: 200440792-200440792 on Build GRCh38
rs376826553 Chr.2: 200440628-200440628 on Build GRCh38
rs949012909 Chr.2: 200440902-200440902 on Build GRCh38
rs771764549 Chr.2: 200440691-200440691 on Build GRCh38
rs190537548 Chr.2: 200440781-200440781 on Build GRCh38
rs778134482 Chr.2: 200440613-200440613 on Build GRCh38
rs749954837 Chr.2: 200440571-200440571 on Build GRCh38
rs747774985 Chr.2: 200440683-200440683 on Build GRCh38
rs750329896 Chr.2: 200440712-200440712 on Build GRCh38
rs1028989249 Chr.2: 200440533-200440533 on Build GRCh38
rs946010638 Chr.2: 200440637-200440637 on Build GRCh38
rs539223309 Chr.2: 200440798-200440798 on Build GRCh38
rs746810680 Chr.2: 200440912-200440912 on Build GRCh38
rs889620274 Chr.2: 200440811-200440811 on Build GRCh38
rs779782846 Chr.2: 200440875-200440875 on Build GRCh38
rs773975917 Chr.2: 200440678-200440678 on Build GRCh38
rs768369547 Chr.2: 200440669-200440669 on Build GRCh38
rs545179741 Chr.2: 200440856-200440856 on Build GRCh38
rs372942551 Chr.2: 200440713-200440713 on Build GRCh38
rs764390276 Chr.2: 200440718-200440718 on Build GRCh38
rs773018155 Chr.2: 200440692-200440692 on Build GRCh38
rs183762345 Chr.2: 200440541-200440541 on Build GRCh38
rs748769319 Chr.2: 200440665-200440665 on Build GRCh38
rs952924537 Chr.2: 200440532-200440532 on Build GRCh38
rs1005797534 Chr.2: 200440992-200440992 on Build GRCh38
rs916276894 Chr.2: 200440869-200440869 on Build GRCh38
rs888718195 Chr.2: 200440941-200440941 on Build GRCh38
rs755560576 Chr.2: 200440918-200440918 on Build GRCh38

View Full Product Details