Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207827656-207828150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHM3
Entrez Gene ID
389072
Gene Name
pleckstrin homology domain containing M3
Gene Aliases
DAPR, PLEKHM1L
UniGene
Hs.159188
Gene Chromosome Location
Chr.2: 207821288-208025675 on Build GRCh38
Transcripts
NM_001080475.2 XM_005246545.3 XM_011511160.2 XM_017004072.1 XM_017004073.1

SNP Detail

SNP ID Chromosome Location
rs1011171171 Chr.2: 207827776-207827776 on Build GRCh38
rs746883255 Chr.2: 207827981-207827981 on Build GRCh38
rs927111315 Chr.2: 207827992-207827993 on Build GRCh38
rs984003590 Chr.2: 207827980-207827980 on Build GRCh38
rs941392130 Chr.2: 207827731-207827731 on Build GRCh38
rs768112401 Chr.2: 207827847-207827847 on Build GRCh38
rs933765681 Chr.2: 207828021-207828021 on Build GRCh38
rs890036002 Chr.2: 207827701-207827701 on Build GRCh38
rs530454022 Chr.2: 207827901-207827901 on Build GRCh38
rs541630968 Chr.2: 207828120-207828120 on Build GRCh38
rs918603264 Chr.2: 207828047-207828047 on Build GRCh38
rs560520494 Chr.2: 207827944-207827944 on Build GRCh38
rs148786316 Chr.2: 207827685-207827685 on Build GRCh38
rs73983606 Chr.2: 207827985-207827985 on Build GRCh38
rs552351332 Chr.2: 207828028-207828028 on Build GRCh38
rs888450374 Chr.2: 207827707-207827707 on Build GRCh38
rs376792483 Chr.2: 207828041-207828041 on Build GRCh38
rs897371647 Chr.2: 207827760-207827760 on Build GRCh38
rs146154097 Chr.2: 207827682-207827682 on Build GRCh38
rs960800040 Chr.2: 207827950-207827950 on Build GRCh38
rs990777694 Chr.2: 207827974-207827974 on Build GRCh38
rs777791940 Chr.2: 207827951-207827951 on Build GRCh38
rs1037041552 Chr.2: 207827759-207827759 on Build GRCh38
rs550784300 Chr.2: 207828067-207828067 on Build GRCh38
rs75842382 Chr.2: 207828032-207828032 on Build GRCh38
rs981203117 Chr.2: 207827680-207827680 on Build GRCh38
rs188021026 Chr.2: 207827894-207827894 on Build GRCh38
rs761631553 Chr.2: 207827979-207827979 on Build GRCh38
rs112284760 Chr.2: 207827743-207827743 on Build GRCh38
rs971234480 Chr.2: 207828096-207828096 on Build GRCh38
rs889990050 Chr.2: 207827919-207827919 on Build GRCh38
rs150726386 Chr.2: 207828041-207828041 on Build GRCh38
rs180758429 Chr.2: 207828031-207828031 on Build GRCh38
rs922375684 Chr.2: 207828003-207828003 on Build GRCh38
rs1022846191 Chr.2: 207827782-207827782 on Build GRCh38
rs939881865 Chr.2: 207827987-207827987 on Build GRCh38
rs546996068 Chr.2: 207827936-207827936 on Build GRCh38
rs1055641474 Chr.2: 207827697-207827697 on Build GRCh38
rs916614056 Chr.2: 207827975-207827975 on Build GRCh38
rs373197047 Chr.2: 207828009-207828009 on Build GRCh38
rs1035096544 Chr.2: 207827935-207827935 on Build GRCh38
rs934219194 Chr.2: 207828064-207828064 on Build GRCh38
rs1037298417 Chr.2: 207827994-207827994 on Build GRCh38

View Full Product Details