Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210017228-210017729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPE
Entrez Gene ID
6120
Gene Name
ribulose-5-phosphate-3-epimerase
Gene Aliases
RPE2-1
UniGene
Hs.282260
Gene Chromosome Location
Chr.2: 210002565-210029156 on Build GRCh38
Transcripts
NM_001278282.1 NM_001278283.1 NM_001278285.1 NM_001278286.1 NM_001278288.1 NM_001278289.1 NM_001318926.1 NM_001318927.1 NM_001318928.1 NM_001318929.1 NM_001318930.1 NM_001318931.1 NM_006916.2 NM_199229.2 XM_006712677.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7423148 c.364A>G p.I122V Chr.2: 210017509-210017509 on Build GRCh38
7423147 c.514A>G p.I172V Chr.2: 210017509-210017509 on Build GRCh38
4090748 c.519G>T p.E173D Chr.2: 210017514-210017514 on Build GRCh38
6893331 c.520G>T p.V174F Chr.2: 210017515-210017515 on Build GRCh38
1641813 c.406T>C p.C136R Chr.2: 210017551-210017551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367899750 Chr.2: 210017597-210017597 on Build GRCh38
rs757755882 Chr.2: 210017465-210017465 on Build GRCh38
rs199875715 Chr.2: 210017451-210017451 on Build GRCh38
rs905894978 Chr.2: 210017389-210017389 on Build GRCh38
rs964691416 Chr.2: 210017571-210017571 on Build GRCh38
rs909981865 Chr.2: 210017368-210017377 on Build GRCh38
rs988084028 Chr.2: 210017285-210017285 on Build GRCh38
rs561241245 Chr.2: 210017517-210017517 on Build GRCh38
rs957900609 Chr.2: 210017674-210017674 on Build GRCh38
rs1011470472 Chr.2: 210017308-210017308 on Build GRCh38
rs1033478085 Chr.2: 210017640-210017640 on Build GRCh38
rs779936212 Chr.2: 210017598-210017598 on Build GRCh38
rs776855287 Chr.2: 210017544-210017544 on Build GRCh38
rs1000273422 Chr.2: 210017617-210017617 on Build GRCh38
rs985990712 Chr.2: 210017270-210017270 on Build GRCh38
rs751889198 Chr.2: 210017588-210017588 on Build GRCh38
rs947300100 Chr.2: 210017606-210017606 on Build GRCh38
rs13406045 Chr.2: 210017384-210017384 on Build GRCh38
rs1036146881 Chr.2: 210017468-210017468 on Build GRCh38
rs758080688 Chr.2: 210017452-210017452 on Build GRCh38
rs764614736 Chr.2: 210017558-210017558 on Build GRCh38
rs1043057306 Chr.2: 210017316-210017316 on Build GRCh38
rs764376339 Chr.2: 210017455-210017455 on Build GRCh38
rs546764801 Chr.2: 210017621-210017621 on Build GRCh38
rs541419383 Chr.2: 210017279-210017279 on Build GRCh38
rs769009455 Chr.2: 210017519-210017519 on Build GRCh38
rs773670837 Chr.2: 210017265-210017265 on Build GRCh38
rs891698937 Chr.2: 210017305-210017305 on Build GRCh38
rs994308524 Chr.2: 210017490-210017490 on Build GRCh38
rs960130753 Chr.2: 210017470-210017470 on Build GRCh38
rs746371990 Chr.2: 210017426-210017431 on Build GRCh38
rs762534839 Chr.2: 210017575-210017575 on Build GRCh38
rs1057145601 Chr.2: 210017701-210017701 on Build GRCh38
rs767945637 Chr.2: 210017590-210017590 on Build GRCh38
rs759542962 Chr.2: 210017550-210017550 on Build GRCh38
rs200970819 Chr.2: 210017535-210017535 on Build GRCh38
rs1025847765 Chr.2: 210017547-210017547 on Build GRCh38
rs551929409 Chr.2: 210017670-210017670 on Build GRCh38
rs369233313 Chr.2: 210017432-210017432 on Build GRCh38
rs760979943 Chr.2: 210017284-210017284 on Build GRCh38
rs773123860 Chr.2: 210017527-210017527 on Build GRCh38
rs368588776 Chr.2: 210017543-210017543 on Build GRCh38
rs949075668 Chr.2: 210017412-210017412 on Build GRCh38
rs756097904 Chr.2: 210017482-210017482 on Build GRCh38
rs202066051 Chr.2: 210017532-210017532 on Build GRCh38
rs764923712 Chr.2: 210017449-210017449 on Build GRCh38
rs77397340 Chr.2: 210017591-210017591 on Build GRCh38
rs62001904 Chr.2: 210017559-210017559 on Build GRCh38
rs571626807 Chr.2: 210017679-210017679 on Build GRCh38
rs1049076263 Chr.2: 210017540-210017540 on Build GRCh38
rs756882510 Chr.2: 210017339-210017339 on Build GRCh38
rs745937680 Chr.2: 210017467-210017467 on Build GRCh38
rs1028429185 Chr.2: 210017257-210017257 on Build GRCh38
rs905785095 Chr.2: 210017678-210017678 on Build GRCh38
rs773141918 Chr.2: 210017423-210017423 on Build GRCh38
rs528566114 Chr.2: 210017647-210017647 on Build GRCh38
rs751935847 Chr.2: 210017463-210017463 on Build GRCh38
rs774986952 Chr.2: 210017568-210017568 on Build GRCh38
rs757667816 Chr.2: 210017589-210017589 on Build GRCh38
rs770875515 Chr.2: 210017542-210017542 on Build GRCh38
rs2075116 Chr.2: 210017447-210017447 on Build GRCh38
rs149656701 Chr.2: 210017530-210017530 on Build GRCh38
rs780210815 Chr.2: 210017493-210017493 on Build GRCh38
rs370180062 Chr.2: 210017580-210017580 on Build GRCh38
rs377324307 Chr.2: 210017429-210017429 on Build GRCh38
rs566587944 Chr.2: 210017644-210017644 on Build GRCh38
rs766288582 Chr.2: 210017439-210017439 on Build GRCh38
rs911484812 Chr.2: 210017566-210017566 on Build GRCh38
rs1046019201 Chr.2: 210017445-210017448 on Build GRCh38
rs373744535 Chr.2: 210017355-210017355 on Build GRCh38

View Full Product Details