Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215319668-215320216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATIC
Entrez Gene ID
471
Gene Name
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
Gene Aliases
AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH
UniGene
Hs.90280
Gene Chromosome Location
Chr.2: 215311956-215359745 on Build GRCh38
Transcripts
NM_004044.6 XM_017004187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1658951 c.236G>A p.R79H Chr.2: 215319677-215319677 on Build GRCh38
3577493 c.244C>T p.P82S Chr.2: 215319685-215319685 on Build GRCh38
2717143 c.244delC p.P82fs*15 Chr.2: 215319685-215319685 on Build GRCh38
3695251 c.229C>T p.L77L Chr.2: 215319670-215319670 on Build GRCh38
4091080 c.232G>T p.A78S Chr.2: 215319673-215319673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919217607 Chr.2: 215319833-215319833 on Build GRCh38
rs778329781 Chr.2: 215320140-215320140 on Build GRCh38
rs184655279 Chr.2: 215319986-215319986 on Build GRCh38
rs901290882 Chr.2: 215320050-215320050 on Build GRCh38
rs75086019 Chr.2: 215319722-215319722 on Build GRCh38
rs748166811 Chr.2: 215319773-215319773 on Build GRCh38
rs74794855 Chr.2: 215319917-215319917 on Build GRCh38
rs898514599 Chr.2: 215319793-215319793 on Build GRCh38
rs756403064 Chr.2: 215319762-215319762 on Build GRCh38
rs749755254 Chr.2: 215319766-215319766 on Build GRCh38
rs1040203602 Chr.2: 215320039-215320039 on Build GRCh38
rs888950484 Chr.2: 215319845-215319845 on Build GRCh38
rs372474957 Chr.2: 215319765-215319765 on Build GRCh38
rs540220274 Chr.2: 215320097-215320097 on Build GRCh38
rs969976234 Chr.2: 215319955-215319955 on Build GRCh38
rs930554852 Chr.2: 215319835-215319835 on Build GRCh38
rs374183545 Chr.2: 215319706-215319706 on Build GRCh38
rs533578957 Chr.2: 215319784-215319784 on Build GRCh38
rs145156457 Chr.2: 215319891-215319891 on Build GRCh38
rs1022520347 Chr.2: 215319951-215319951 on Build GRCh38
rs1023001339 Chr.2: 215320129-215320129 on Build GRCh38
rs959424429 Chr.2: 215319963-215319963 on Build GRCh38
rs376889224 Chr.2: 215319739-215319739 on Build GRCh38
rs1041116277 Chr.2: 215320069-215320069 on Build GRCh38
rs750998008 Chr.2: 215319754-215319754 on Build GRCh38
rs369513149 Chr.2: 215319703-215319703 on Build GRCh38
rs1052478234 Chr.2: 215320065-215320065 on Build GRCh38
rs1010375526 Chr.2: 215320120-215320120 on Build GRCh38
rs985131289 Chr.2: 215320020-215320020 on Build GRCh38
rs188322235 Chr.2: 215320075-215320075 on Build GRCh38
rs762571910 Chr.2: 215319747-215319747 on Build GRCh38
rs763654938 Chr.2: 215319752-215319752 on Build GRCh38
rs759249668 Chr.2: 215319728-215319728 on Build GRCh38
rs568343379 Chr.2: 215319927-215319927 on Build GRCh38
rs535344945 Chr.2: 215319937-215319937 on Build GRCh38
rs755448727 Chr.2: 215319769-215319769 on Build GRCh38
rs574248544 Chr.2: 215320078-215320078 on Build GRCh38
rs747810267 Chr.2: 215320155-215320155 on Build GRCh38
rs979169072 Chr.2: 215319707-215319707 on Build GRCh38
rs778243464 Chr.2: 215319750-215319751 on Build GRCh38
rs780532278 Chr.2: 215319701-215319701 on Build GRCh38
rs980953629 Chr.2: 215319960-215319960 on Build GRCh38
rs147500924 Chr.2: 215320019-215320019 on Build GRCh38
rs139082550 Chr.2: 215319826-215319826 on Build GRCh38
rs768224425 Chr.2: 215320100-215320100 on Build GRCh38
rs1049846568 Chr.2: 215319818-215319818 on Build GRCh38
rs535039357 Chr.2: 215319741-215319741 on Build GRCh38
rs767062809 Chr.2: 215319758-215319758 on Build GRCh38
rs1021841451 Chr.2: 215320131-215320131 on Build GRCh38
rs939052047 Chr.2: 215319819-215319819 on Build GRCh38
rs934070045 Chr.2: 215320051-215320051 on Build GRCh38
rs145404605 Chr.2: 215320191-215320191 on Build GRCh38
rs756428011 Chr.2: 215319743-215319743 on Build GRCh38
rs960442676 Chr.2: 215319984-215319984 on Build GRCh38
rs760301346 Chr.2: 215319729-215319729 on Build GRCh38
rs995475360 Chr.2: 215319816-215319816 on Build GRCh38
rs1040222301 Chr.2: 215319875-215319875 on Build GRCh38
rs529604608 Chr.2: 215319745-215319745 on Build GRCh38
rs776551356 Chr.2: 215319727-215319727 on Build GRCh38
rs111784213 Chr.2: 215320026-215320026 on Build GRCh38
rs189148901 Chr.2: 215319896-215319896 on Build GRCh38
rs889782768 Chr.2: 215320041-215320041 on Build GRCh38
rs773244918 Chr.2: 215319776-215319776 on Build GRCh38
rs145610902 Chr.2: 215320106-215320106 on Build GRCh38
rs756981785 Chr.2: 215319757-215319757 on Build GRCh38
rs1037366782 Chr.2: 215320004-215320004 on Build GRCh38
rs193130247 Chr.2: 215319976-215319976 on Build GRCh38
rs752911895 Chr.2: 215319703-215319704 on Build GRCh38
rs748443665 Chr.2: 215319720-215319720 on Build GRCh38
rs1002826243 Chr.2: 215320153-215320153 on Build GRCh38
rs774470167 Chr.2: 215319713-215319713 on Build GRCh38
rs376959214 Chr.2: 215319774-215319774 on Build GRCh38
rs16853782 Chr.2: 215319759-215319759 on Build GRCh38
rs901238579 Chr.2: 215319883-215319883 on Build GRCh38
rs779099499 Chr.2: 215319772-215319772 on Build GRCh38
rs1035144813 Chr.2: 215320166-215320166 on Build GRCh38
rs780088259 Chr.2: 215319985-215319985 on Build GRCh38
rs1020236528 Chr.2: 215319863-215319863 on Build GRCh38
rs943219685 Chr.2: 215320036-215320036 on Build GRCh38
rs1048938023 Chr.2: 215319836-215319836 on Build GRCh38
rs751894254 Chr.2: 215319702-215319702 on Build GRCh38
rs755837732 Chr.2: 215319974-215319974 on Build GRCh38
rs943188048 Chr.2: 215319868-215319868 on Build GRCh38

View Full Product Details