Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216082063-216082556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM169
Entrez Gene ID
92691
Gene Name
transmembrane protein 169
Gene Aliases
-
UniGene
Hs.334916
Gene Chromosome Location
Chr.2: 216081866-216102783 on Build GRCh38
Transcripts
NM_001142310.1 NM_001142311.1 NM_001142312.1 NM_138390.3 XM_006712851.1 XM_011512173.2

SNP Detail

SNP ID Chromosome Location
rs367682194 Chr.2: 216082209-216082209 on Build GRCh38
rs959921576 Chr.2: 216082244-216082244 on Build GRCh38
rs759213964 Chr.2: 216082470-216082470 on Build GRCh38
rs562008502 Chr.2: 216082343-216082343 on Build GRCh38
rs1014051906 Chr.2: 216082254-216082254 on Build GRCh38
rs971558215 Chr.2: 216082414-216082414 on Build GRCh38
rs975453376 Chr.2: 216082337-216082337 on Build GRCh38
rs571976154 Chr.2: 216082529-216082529 on Build GRCh38
rs930643903 Chr.2: 216082461-216082461 on Build GRCh38
rs966778723 Chr.2: 216082185-216082185 on Build GRCh38
rs983872027 Chr.2: 216082308-216082308 on Build GRCh38
rs57310304 Chr.2: 216082339-216082339 on Build GRCh38
rs917461728 Chr.2: 216082376-216082376 on Build GRCh38
rs550030999 Chr.2: 216082301-216082301 on Build GRCh38
rs557106393 Chr.2: 216082090-216082090 on Build GRCh38
rs900982729 Chr.2: 216082534-216082534 on Build GRCh38
rs1039547778 Chr.2: 216082179-216082179 on Build GRCh38
rs558439506 Chr.2: 216082469-216082469 on Build GRCh38
rs776655706 Chr.2: 216082362-216082362 on Build GRCh38
rs913394498 Chr.2: 216082386-216082386 on Build GRCh38
rs539587067 Chr.2: 216082152-216082152 on Build GRCh38
rs765052723 Chr.2: 216082523-216082523 on Build GRCh38
rs967040295 Chr.2: 216082221-216082221 on Build GRCh38
rs951176393 Chr.2: 216082294-216082294 on Build GRCh38
rs1001883399 Chr.2: 216082210-216082210 on Build GRCh38
rs558241200 Chr.2: 216082214-216082214 on Build GRCh38
rs889224183 Chr.2: 216082166-216082166 on Build GRCh38
rs1043128667 Chr.2: 216082479-216082479 on Build GRCh38
rs904583377 Chr.2: 216082481-216082481 on Build GRCh38
rs909560201 Chr.2: 216082329-216082329 on Build GRCh38
rs963763917 Chr.2: 216082335-216082335 on Build GRCh38
rs1034248997 Chr.2: 216082213-216082213 on Build GRCh38
rs543803674 Chr.2: 216082377-216082377 on Build GRCh38
rs904510752 Chr.2: 216082164-216082164 on Build GRCh38
rs1025395628 Chr.2: 216082260-216082260 on Build GRCh38
rs919521553 Chr.2: 216082454-216082454 on Build GRCh38
rs1020630914 Chr.2: 216082217-216082217 on Build GRCh38
rs573054174 Chr.2: 216082311-216082311 on Build GRCh38
rs532954883 Chr.2: 216082522-216082522 on Build GRCh38
rs946153065 Chr.2: 216082446-216082446 on Build GRCh38
rs775198280 Chr.2: 216082531-216082531 on Build GRCh38
rs910631533 Chr.2: 216082491-216082491 on Build GRCh38
rs773424660 Chr.2: 216082168-216082168 on Build GRCh38
rs184036099 Chr.2: 216082124-216082124 on Build GRCh38
rs538400263 Chr.2: 216082371-216082371 on Build GRCh38
rs1006666737 Chr.2: 216082175-216082175 on Build GRCh38
rs930653242 Chr.2: 216082117-216082117 on Build GRCh38
rs186060573 Chr.2: 216082497-216082497 on Build GRCh38
rs1040440611 Chr.2: 216082506-216082506 on Build GRCh38
rs1051898527 Chr.2: 216082086-216082086 on Build GRCh38
rs575688901 Chr.2: 216082100-216082100 on Build GRCh38
rs901095950 Chr.2: 216082192-216082192 on Build GRCh38

View Full Product Details