Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218659247-218659750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCS1L
Entrez Gene ID
617
Gene Name
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
Gene Aliases
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1
UniGene
Hs.471401
Gene Chromosome Location
Chr.2: 218659656-218663443 on Build GRCh38
Transcripts
NM_001079866.1 NM_001257342.1 NM_001257343.1 NM_001257344.1 NM_001318836.1 NM_001320717.1 NM_004328.4 XM_005246749.3 XM_017004631.1 XM_017004632.1 XM_017004633.1 NM_001105537.2 NR_073599.1 NR_073600.1 XM_006712730.3 XM_011511784.2 XM_011511785.2 XM_011511786.2 XM_011511787.2 XM_011511788.2 XM_011511789.2 XM_017004871.1 XM_017004872.1

Gene Symbol
ZNF142
Entrez Gene ID
7701
Gene Name
zinc finger protein 142
Gene Aliases
pHZ-49
UniGene
Hs.657969
Gene Chromosome Location
Chr.2: 218637592-218659632 on Build GRCh38
Transcripts
NM_001079866.1 NM_001257342.1 NM_001257343.1 NM_001257344.1 NM_001318836.1 NM_001320717.1 NM_004328.4 XM_005246749.3 XM_017004631.1 XM_017004632.1 XM_017004633.1 NM_001105537.2 NR_073599.1 NR_073600.1 XM_006712730.3 XM_011511784.2 XM_011511785.2 XM_011511786.2 XM_011511787.2 XM_011511788.2 XM_011511789.2 XM_017004871.1 XM_017004872.1

SNP Detail

SNP ID Chromosome Location
rs555081900 Chr.2: 218659395-218659395 on Build GRCh38
rs1033936890 Chr.2: 218659359-218659359 on Build GRCh38
rs28384379 Chr.2: 218659583-218659583 on Build GRCh38
rs909837004 Chr.2: 218659468-218659468 on Build GRCh38
rs1010896493 Chr.2: 218659543-218659543 on Build GRCh38
rs770357884 Chr.2: 218659559-218659569 on Build GRCh38
rs1055327948 Chr.2: 218659714-218659714 on Build GRCh38
rs180771102 Chr.2: 218659348-218659348 on Build GRCh38
rs183751599 Chr.2: 218659599-218659599 on Build GRCh38
rs918318860 Chr.2: 218659485-218659485 on Build GRCh38
rs966847628 Chr.2: 218659405-218659405 on Build GRCh38
rs59299161 Chr.2: 218659478-218659478 on Build GRCh38
rs906709368 Chr.2: 218659336-218659336 on Build GRCh38
rs960194557 Chr.2: 218659298-218659298 on Build GRCh38
rs1029473436 Chr.2: 218659426-218659426 on Build GRCh38
rs764316987 Chr.2: 218659293-218659293 on Build GRCh38
rs1020936475 Chr.2: 218659556-218659556 on Build GRCh38
rs540816120 Chr.2: 218659632-218659632 on Build GRCh38
rs941028497 Chr.2: 218659470-218659470 on Build GRCh38
rs981183136 Chr.2: 218659659-218659659 on Build GRCh38
rs754029162 Chr.2: 218659329-218659329 on Build GRCh38
rs1043372935 Chr.2: 218659353-218659353 on Build GRCh38
rs1016238696 Chr.2: 218659426-218659426 on Build GRCh38
rs1021029658 Chr.2: 218659404-218659404 on Build GRCh38
rs371893965 Chr.2: 218659340-218659340 on Build GRCh38
rs567005931 Chr.2: 218659418-218659418 on Build GRCh38
rs954346259 Chr.2: 218659648-218659648 on Build GRCh38
rs954015234 Chr.2: 218659434-218659434 on Build GRCh38
rs953894576 Chr.2: 218659587-218659587 on Build GRCh38
rs889188875 Chr.2: 218659415-218659415 on Build GRCh38
rs906666176 Chr.2: 218659511-218659511 on Build GRCh38
rs1016788474 Chr.2: 218659601-218659601 on Build GRCh38
rs927071534 Chr.2: 218659681-218659681 on Build GRCh38
rs938140522 Chr.2: 218659708-218659708 on Build GRCh38
rs929447867 Chr.2: 218659328-218659328 on Build GRCh38
rs894084076 Chr.2: 218659531-218659531 on Build GRCh38
rs971475866 Chr.2: 218659650-218659650 on Build GRCh38
rs918237681 Chr.2: 218659628-218659628 on Build GRCh38
rs992100094 Chr.2: 218659325-218659325 on Build GRCh38
rs1037746368 Chr.2: 218659691-218659691 on Build GRCh38
rs13011338 Chr.2: 218659366-218659366 on Build GRCh38
rs977336226 Chr.2: 218659421-218659421 on Build GRCh38
rs555711517 Chr.2: 218659494-218659494 on Build GRCh38
rs998240247 Chr.2: 218659571-218659571 on Build GRCh38
rs575906291 Chr.2: 218659496-218659496 on Build GRCh38
rs902530079 Chr.2: 218659568-218659568 on Build GRCh38
rs1055380285 Chr.2: 218659524-218659524 on Build GRCh38
rs533044382 Chr.2: 218659313-218659313 on Build GRCh38
rs546136541 Chr.2: 218659527-218659527 on Build GRCh38
rs879280893 Chr.2: 218659449-218659449 on Build GRCh38
rs1052356216 Chr.2: 218659368-218659368 on Build GRCh38
rs962632190 Chr.2: 218659622-218659622 on Build GRCh38
rs533735569 Chr.2: 218659389-218659389 on Build GRCh38
rs979597626 Chr.2: 218659521-218659521 on Build GRCh38
rs1023572500 Chr.2: 218659496-218659497 on Build GRCh38
rs1010948755 Chr.2: 218659387-218659387 on Build GRCh38
rs1030290151 Chr.2: 218659448-218659448 on Build GRCh38
rs897946493 Chr.2: 218659702-218659702 on Build GRCh38
rs972639117 Chr.2: 218659624-218659624 on Build GRCh38
rs967620458 Chr.2: 218659305-218659305 on Build GRCh38
rs942094975 Chr.2: 218659645-218659645 on Build GRCh38
rs1045909633 Chr.2: 218659497-218659497 on Build GRCh38
rs938188346 Chr.2: 218659522-218659522 on Build GRCh38

View Full Product Details