Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218662783-218663280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCS1L
Entrez Gene ID
617
Gene Name
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
Gene Aliases
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1
UniGene
Hs.471401
Gene Chromosome Location
Chr.2: 218659656-218663443 on Build GRCh38
Transcripts
NM_001079866.1 NM_001257342.1 NM_001257343.1 NM_001257344.1 NM_001318836.1 NM_001320717.1 NM_004328.4 XM_005246748.2 XM_005246749.3 XM_006712678.1 XM_017004631.1 XM_017004632.1 XM_017004633.1 XM_017004634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7226170 c.1017T>C p.P339P Chr.2: 218663143-218663143 on Build GRCh38
4624580 c.904C>T p.Q302* Chr.2: 218662897-218662897 on Build GRCh38
6611127 c.974G>A p.R325H Chr.2: 218662967-218662967 on Build GRCh38
3407576 c.973C>T p.R325C Chr.2: 218662966-218662966 on Build GRCh38
1163610 c.1075T>A p.W359R Chr.2: 218663201-218663201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780931624 Chr.2: 218663044-218663044 on Build GRCh38
rs565428815 Chr.2: 218663167-218663167 on Build GRCh38
rs559519953 Chr.2: 218663008-218663008 on Build GRCh38
rs752599536 Chr.2: 218663200-218663200 on Build GRCh38
rs771920503 Chr.2: 218663089-218663089 on Build GRCh38
rs779797289 Chr.2: 218663038-218663038 on Build GRCh38
rs770483323 Chr.2: 218663100-218663100 on Build GRCh38
rs767137147 Chr.2: 218662873-218662873 on Build GRCh38
rs751769260 Chr.2: 218662942-218662943 on Build GRCh38
rs980333147 Chr.2: 218663029-218663029 on Build GRCh38
rs115594405 Chr.2: 218663016-218663016 on Build GRCh38
rs759832405 Chr.2: 218663048-218663049 on Build GRCh38
rs777504463 Chr.2: 218663221-218663221 on Build GRCh38
rs35843327 Chr.2: 218663143-218663143 on Build GRCh38
rs779805975 Chr.2: 218662978-218662978 on Build GRCh38
rs777854469 Chr.2: 218662958-218662958 on Build GRCh38
rs758285505 Chr.2: 218663205-218663205 on Build GRCh38
rs121908574 Chr.2: 218663183-218663183 on Build GRCh38
rs563252324 Chr.2: 218663045-218663045 on Build GRCh38
rs554510882 Chr.2: 218662845-218662845 on Build GRCh38
rs774076186 Chr.2: 218662972-218662972 on Build GRCh38
rs773582375 Chr.2: 218663157-218663157 on Build GRCh38
rs906812769 Chr.2: 218663173-218663175 on Build GRCh38
rs773812835 Chr.2: 218662900-218662900 on Build GRCh38
rs746710484 Chr.2: 218663238-218663238 on Build GRCh38
rs199791659 Chr.2: 218662820-218662820 on Build GRCh38
rs926239823 Chr.2: 218663078-218663078 on Build GRCh38
rs754988448 Chr.2: 218663182-218663182 on Build GRCh38
rs1000629747 Chr.2: 218662887-218662887 on Build GRCh38
rs771655220 Chr.2: 218662983-218662983 on Build GRCh38
rs747778414 Chr.2: 218663052-218663052 on Build GRCh38
rs373812068 Chr.2: 218663005-218663005 on Build GRCh38
rs587777278 Chr.2: 218662894-218662894 on Build GRCh38
rs756064291 Chr.2: 218663034-218663034 on Build GRCh38
rs377378571 Chr.2: 218663160-218663160 on Build GRCh38
rs779892991 Chr.2: 218662966-218662966 on Build GRCh38
rs752868592 Chr.2: 218663010-218663010 on Build GRCh38
rs543655130 Chr.2: 218662969-218662969 on Build GRCh38
rs33946522 Chr.2: 218662989-218662989 on Build GRCh38
rs200593699 Chr.2: 218662992-218662992 on Build GRCh38
rs1008992738 Chr.2: 218662927-218662927 on Build GRCh38
rs372817977 Chr.2: 218662960-218662960 on Build GRCh38
rs752921674 Chr.2: 218662933-218662933 on Build GRCh38
rs769721223 Chr.2: 218663158-218663158 on Build GRCh38
rs749094061 Chr.2: 218662967-218662967 on Build GRCh38
rs761731970 Chr.2: 218663164-218663164 on Build GRCh38
rs778870782 Chr.2: 218663049-218663049 on Build GRCh38
rs1004254276 Chr.2: 218663152-218663152 on Build GRCh38
rs765324054 Chr.2: 218662929-218662929 on Build GRCh38
rs761284622 Chr.2: 218662903-218662903 on Build GRCh38
rs754225834 Chr.2: 218662914-218662914 on Build GRCh38
rs550497120 Chr.2: 218663162-218663162 on Build GRCh38
rs912520138 Chr.2: 218663064-218663064 on Build GRCh38
rs386833858 Chr.2: 218662973-218662973 on Build GRCh38
rs887329900 Chr.2: 218662829-218662829 on Build GRCh38
rs781639231 Chr.2: 218662964-218662964 on Build GRCh38
rs770298779 Chr.2: 218663239-218663239 on Build GRCh38
rs936010395 Chr.2: 218663079-218663079 on Build GRCh38
rs766737254 Chr.2: 218662911-218662911 on Build GRCh38
rs541260525 Chr.2: 218662940-218662943 on Build GRCh38
rs1053496823 Chr.2: 218663120-218663120 on Build GRCh38
rs199661869 Chr.2: 218663245-218663245 on Build GRCh38
rs574554623 Chr.2: 218663009-218663009 on Build GRCh38
rs746449962 Chr.2: 218663098-218663098 on Build GRCh38
rs887110312 Chr.2: 218663145-218663145 on Build GRCh38
rs148941016 Chr.2: 218663170-218663170 on Build GRCh38
rs1044684262 Chr.2: 218662808-218662808 on Build GRCh38
rs373671954 Chr.2: 218663129-218663129 on Build GRCh38
rs1057516518 Chr.2: 218662966-218662967 on Build GRCh38
rs745785523 Chr.2: 218662959-218662959 on Build GRCh38
rs777411117 Chr.2: 218663094-218663094 on Build GRCh38
rs1033866368 Chr.2: 218663247-218663247 on Build GRCh38
rs768258849 Chr.2: 218662838-218662838 on Build GRCh38
rs758434684 Chr.2: 218662950-218662950 on Build GRCh38
rs377502595 Chr.2: 218663012-218663012 on Build GRCh38
rs767723013 Chr.2: 218663107-218663108 on Build GRCh38
rs945913428 Chr.2: 218663043-218663043 on Build GRCh38
rs948907642 Chr.2: 218662805-218662805 on Build GRCh38
rs761345871 Chr.2: 218662864-218662864 on Build GRCh38
rs111718078 Chr.2: 218663229-218663229 on Build GRCh38
rs749499100 Chr.2: 218663211-218663211 on Build GRCh38
rs141618813 Chr.2: 218662918-218662918 on Build GRCh38
rs369467055 Chr.2: 218663021-218663021 on Build GRCh38
rs760112313 Chr.2: 218663131-218663131 on Build GRCh38
rs146731467 Chr.2: 218662993-218662993 on Build GRCh38
rs931417861 Chr.2: 218663141-218663141 on Build GRCh38

View Full Product Details