Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219237094-219237600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L
Entrez Gene ID
79411
Gene Name
galactosidase beta 1 like
Gene Aliases
-
UniGene
Hs.181173
Gene Chromosome Location
Chr.2: 219236598-219245429 on Build GRCh38
Transcripts
NM_001286423.1 NM_001286427.1 NM_024506.4 XM_011511813.2 XM_017004895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
442341 c.1790G>A p.G597E Chr.2: 219237247-219237247 on Build GRCh38
3044715 c.1655G>T p.G552V Chr.2: 219237546-219237546 on Build GRCh38
6671361 c.1665T>C p.F555F Chr.2: 219237536-219237536 on Build GRCh38
4530755 c.1724G>A p.R575Q Chr.2: 219237313-219237313 on Build GRCh38
5824621 c.1875T>C p.N625N Chr.2: 219237162-219237162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938998546 Chr.2: 219237188-219237188 on Build GRCh38
rs764369890 Chr.2: 219237257-219237257 on Build GRCh38
rs773230732 Chr.2: 219237523-219237523 on Build GRCh38
rs951834217 Chr.2: 219237483-219237483 on Build GRCh38
rs757747400 Chr.2: 219237555-219237555 on Build GRCh38
rs776920943 Chr.2: 219237484-219237484 on Build GRCh38
rs773144807 Chr.2: 219237571-219237571 on Build GRCh38
rs73077102 Chr.2: 219237136-219237136 on Build GRCh38
rs917499125 Chr.2: 219237249-219237249 on Build GRCh38
rs201165210 Chr.2: 219237313-219237313 on Build GRCh38
rs750224228 Chr.2: 219237277-219237277 on Build GRCh38
rs367718312 Chr.2: 219237287-219237287 on Build GRCh38
rs976606358 Chr.2: 219237441-219237441 on Build GRCh38
rs377757936 Chr.2: 219237353-219237353 on Build GRCh38
rs773572669 Chr.2: 219237247-219237247 on Build GRCh38
rs750644392 Chr.2: 219237162-219237162 on Build GRCh38
rs775017098 Chr.2: 219237227-219237227 on Build GRCh38
rs781602676 Chr.2: 219237570-219237570 on Build GRCh38
rs746939167 Chr.2: 219237398-219237398 on Build GRCh38
rs536796641 Chr.2: 219237549-219237549 on Build GRCh38
rs369797599 Chr.2: 219237536-219237536 on Build GRCh38
rs771360156 Chr.2: 219237482-219237482 on Build GRCh38
rs760465253 Chr.2: 219237526-219237526 on Build GRCh38
rs1033887141 Chr.2: 219237246-219237247 on Build GRCh38
rs773516908 Chr.2: 219237314-219237314 on Build GRCh38
rs764689569 Chr.2: 219237127-219237127 on Build GRCh38
rs765254114 Chr.2: 219237273-219237273 on Build GRCh38
rs751406171 Chr.2: 219237180-219237180 on Build GRCh38
rs757073267 Chr.2: 219237182-219237182 on Build GRCh38
rs574090998 Chr.2: 219237528-219237528 on Build GRCh38
rs922458907 Chr.2: 219237451-219237451 on Build GRCh38
rs560232852 Chr.2: 219237228-219237228 on Build GRCh38
rs140515508 Chr.2: 219237295-219237295 on Build GRCh38
rs138413810 Chr.2: 219237170-219237170 on Build GRCh38
rs748682402 Chr.2: 219237304-219237304 on Build GRCh38
rs763062232 Chr.2: 219237506-219237506 on Build GRCh38
rs754622826 Chr.2: 219237372-219237372 on Build GRCh38
rs761723547 Chr.2: 219237348-219237348 on Build GRCh38
rs754749397 Chr.2: 219237538-219237538 on Build GRCh38
rs144432913 Chr.2: 219237275-219237275 on Build GRCh38
rs779494101 Chr.2: 219237218-219237218 on Build GRCh38
rs771935064 Chr.2: 219237235-219237237 on Build GRCh38
rs769334330 Chr.2: 219237226-219237226 on Build GRCh38
rs868298488 Chr.2: 219237307-219237307 on Build GRCh38
rs746144984 Chr.2: 219237485-219237485 on Build GRCh38
rs746559824 Chr.2: 219237445-219237445 on Build GRCh38
rs138649763 Chr.2: 219237501-219237501 on Build GRCh38
rs779827171 Chr.2: 219237286-219237286 on Build GRCh38
rs568780321 Chr.2: 219237335-219237335 on Build GRCh38
rs775430327 Chr.2: 219237369-219237370 on Build GRCh38
rs758040894 Chr.2: 219237379-219237379 on Build GRCh38
rs750230745 Chr.2: 219237352-219237352 on Build GRCh38
rs571531765 Chr.2: 219237417-219237417 on Build GRCh38
rs761071051 Chr.2: 219237316-219237316 on Build GRCh38
rs745537696 Chr.2: 219237167-219237167 on Build GRCh38
rs778322417 Chr.2: 219237399-219237399 on Build GRCh38
rs767454459 Chr.2: 219237351-219237351 on Build GRCh38
rs745520438 Chr.2: 219237215-219237215 on Build GRCh38
rs996037613 Chr.2: 219237440-219237440 on Build GRCh38
rs185114654 Chr.2: 219237222-219237222 on Build GRCh38
rs377561268 Chr.2: 219237403-219237403 on Build GRCh38
rs200650413 Chr.2: 219237245-219237245 on Build GRCh38
rs777658245 Chr.2: 219237177-219237177 on Build GRCh38
rs374134175 Chr.2: 219237366-219237366 on Build GRCh38
rs61733623 Chr.2: 219237252-219237252 on Build GRCh38
rs201208828 Chr.2: 219237511-219237511 on Build GRCh38
rs764028007 Chr.2: 219237512-219237512 on Build GRCh38
rs534248238 Chr.2: 219237439-219237439 on Build GRCh38
rs769474345 Chr.2: 219237216-219237216 on Build GRCh38
rs141680556 Chr.2: 219237276-219237276 on Build GRCh38
rs373655923 Chr.2: 219237197-219237197 on Build GRCh38
rs201866761 Chr.2: 219237554-219237554 on Build GRCh38
rs767877997 Chr.2: 219237159-219237159 on Build GRCh38
rs752199098 Chr.2: 219237545-219237545 on Build GRCh38
rs768315289 Chr.2: 219237293-219237293 on Build GRCh38
rs755759702 Chr.2: 219237282-219237282 on Build GRCh38
rs758397955 Chr.2: 219237176-219237176 on Build GRCh38
rs753960401 Chr.2: 219237254-219237254 on Build GRCh38
rs991831167 Chr.2: 219237550-219237550 on Build GRCh38
rs778586373 Chr.2: 219237134-219237135 on Build GRCh38
rs747426884 Chr.2: 219237468-219237468 on Build GRCh38
rs746984049 Chr.2: 219237166-219237166 on Build GRCh38
rs142912134 Chr.2: 219237206-219237206 on Build GRCh38
rs891797648 Chr.2: 219237357-219237357 on Build GRCh38
rs371070419 Chr.2: 219237120-219237120 on Build GRCh38
rs199890080 Chr.2: 219237504-219237504 on Build GRCh38
rs142350544 Chr.2: 219237133-219237133 on Build GRCh38
rs759686734 Chr.2: 219237329-219237329 on Build GRCh38
rs1014638859 Chr.2: 219237521-219237521 on Build GRCh38
rs766386541 Chr.2: 219237168-219237168 on Build GRCh38
rs183755555 Chr.2: 219237578-219237578 on Build GRCh38
rs888367513 Chr.2: 219237260-219237260 on Build GRCh38
rs756297788 Chr.2: 219237163-219237163 on Build GRCh38
rs776863493 Chr.2: 219237327-219237327 on Build GRCh38
rs766233849 Chr.2: 219237358-219237358 on Build GRCh38
rs769025641 Chr.2: 219237184-219237184 on Build GRCh38
rs769999302 Chr.2: 219237486-219237486 on Build GRCh38
rs368363660 Chr.2: 219237374-219237374 on Build GRCh38
rs377210014 Chr.2: 219237533-219237533 on Build GRCh38
rs768697985 Chr.2: 219237467-219237468 on Build GRCh38
rs758868943 Chr.2: 219237384-219237384 on Build GRCh38
rs371589764 Chr.2: 219237318-219237318 on Build GRCh38
rs1027437468 Chr.2: 219237144-219237144 on Build GRCh38
rs374960051 Chr.2: 219237330-219237330 on Build GRCh38
rs764953087 Chr.2: 219237540-219237540 on Build GRCh38

View Full Product Details