Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 224470277-224470800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL3
Entrez Gene ID
8452
Gene Name
cullin 3
Gene Aliases
CUL-3, PHA2E
UniGene
Hs.372286
Gene Chromosome Location
Chr.2: 224470150-224585397 on Build GRCh38
Transcripts
NM_001257197.1 NM_001257198.1 NM_003590.4

SNP Detail

SNP ID Chromosome Location
rs905496426 Chr.2: 224470700-224470700 on Build GRCh38
rs886055678 Chr.2: 224470459-224470460 on Build GRCh38
rs899861706 Chr.2: 224470511-224470511 on Build GRCh38
rs1018725074 Chr.2: 224470380-224470380 on Build GRCh38
rs571981348 Chr.2: 224470520-224470520 on Build GRCh38
rs899010687 Chr.2: 224470638-224470638 on Build GRCh38
rs969747965 Chr.2: 224470593-224470594 on Build GRCh38
rs924536991 Chr.2: 224470695-224470695 on Build GRCh38
rs191987903 Chr.2: 224470432-224470432 on Build GRCh38
rs868308694 Chr.2: 224470446-224470446 on Build GRCh38
rs1010177252 Chr.2: 224470731-224470731 on Build GRCh38
rs1029824675 Chr.2: 224470395-224470395 on Build GRCh38
rs2396092 Chr.2: 224470573-224470573 on Build GRCh38
rs1045541323 Chr.2: 224470330-224470330 on Build GRCh38
rs574153874 Chr.2: 224470599-224470599 on Build GRCh38
rs562201390 Chr.2: 224470350-224470350 on Build GRCh38
rs1010912351 Chr.2: 224470378-224470378 on Build GRCh38
rs903050266 Chr.2: 224470332-224470332 on Build GRCh38
rs982979231 Chr.2: 224470577-224470577 on Build GRCh38
rs914413627 Chr.2: 224470738-224470738 on Build GRCh38
rs933292242 Chr.2: 224470367-224470367 on Build GRCh38
rs991671820 Chr.2: 224470603-224470603 on Build GRCh38
rs139247052 Chr.2: 224470744-224470744 on Build GRCh38
rs940993118 Chr.2: 224470435-224470435 on Build GRCh38
rs1037545969 Chr.2: 224470631-224470631 on Build GRCh38
rs10612010 Chr.2: 224470713-224470714 on Build GRCh38
rs1052912068 Chr.2: 224470459-224470459 on Build GRCh38
rs537457130 Chr.2: 224470644-224470644 on Build GRCh38
rs929155648 Chr.2: 224470641-224470641 on Build GRCh38
rs909635981 Chr.2: 224470431-224470431 on Build GRCh38
rs549963856 Chr.2: 224470417-224470417 on Build GRCh38
rs893757131 Chr.2: 224470373-224470373 on Build GRCh38
rs1039822576 Chr.2: 224470745-224470745 on Build GRCh38
rs5839066 Chr.2: 224470509-224470509 on Build GRCh38
rs749918111 Chr.2: 224470710-224470710 on Build GRCh38
rs977025239 Chr.2: 224470391-224470391 on Build GRCh38
rs949953803 Chr.2: 224470448-224470448 on Build GRCh38
rs893881920 Chr.2: 224470482-224470482 on Build GRCh38
rs982916176 Chr.2: 224470399-224470399 on Build GRCh38
rs562788492 Chr.2: 224470770-224470770 on Build GRCh38
rs547601761 Chr.2: 224470382-224470382 on Build GRCh38
rs999536214 Chr.2: 224470333-224470333 on Build GRCh38
rs1026460390 Chr.2: 224470441-224470441 on Build GRCh38
rs933404090 Chr.2: 224470570-224470570 on Build GRCh38
rs915618253 Chr.2: 224470437-224470437 on Build GRCh38
rs541115656 Chr.2: 224470491-224470491 on Build GRCh38
rs374191606 Chr.2: 224470580-224470580 on Build GRCh38
rs993267933 Chr.2: 224470428-224470428 on Build GRCh38
rs981283535 Chr.2: 224470691-224470691 on Build GRCh38
rs1032521345 Chr.2: 224470512-224470512 on Build GRCh38
rs1018269280 Chr.2: 224470495-224470495 on Build GRCh38
rs764772276 Chr.2: 224470649-224470649 on Build GRCh38
rs934605106 Chr.2: 224470715-224470715 on Build GRCh38
rs969670556 Chr.2: 224470499-224470499 on Build GRCh38
rs886055679 Chr.2: 224470479-224470479 on Build GRCh38
rs913996416 Chr.2: 224470532-224470532 on Build GRCh38
rs949738072 Chr.2: 224470661-224470661 on Build GRCh38
rs567732119 Chr.2: 224470397-224470397 on Build GRCh38
rs898953115 Chr.2: 224470423-224470423 on Build GRCh38
rs991165537 Chr.2: 224470436-224470436 on Build GRCh38
rs1029939029 Chr.2: 224470572-224470572 on Build GRCh38
rs757151773 Chr.2: 224470675-224470675 on Build GRCh38
rs146239785 Chr.2: 224470313-224470313 on Build GRCh38
rs536464065 Chr.2: 224470398-224470398 on Build GRCh38
rs1054292471 Chr.2: 224470734-224470734 on Build GRCh38
rs558805673 Chr.2: 224470492-224470492 on Build GRCh38
rs902999581 Chr.2: 224470454-224470454 on Build GRCh38
rs1031102069 Chr.2: 224470342-224470342 on Build GRCh38
rs1045634838 Chr.2: 224470449-224470449 on Build GRCh38
rs4234054 Chr.2: 224470456-224470456 on Build GRCh38
rs1014366309 Chr.2: 224470479-224470479 on Build GRCh38

View Full Product Details