Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237575296-237575793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB17
Entrez Gene ID
64284
Gene Name
RAB17, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.44278
Gene Chromosome Location
Chr.2: 237574322-237591126 on Build GRCh38
Transcripts
NM_022449.3 NR_033308.1 XM_006712689.1 XM_017004693.1 XM_017004694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7175157 c.454G>T p.D152Y Chr.2: 237575462-237575462 on Build GRCh38
442561 c.471G>A p.L157L Chr.2: 237575445-237575445 on Build GRCh38
6383988 c.452C>T p.A151V Chr.2: 237575464-237575464 on Build GRCh38
1692236 c.487G>A p.A163T Chr.2: 237575429-237575429 on Build GRCh38
7353179 c.491A>G p.K164R Chr.2: 237575425-237575425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376729614 Chr.2: 237575407-237575407 on Build GRCh38
rs908871256 Chr.2: 237575598-237575598 on Build GRCh38
rs369913434 Chr.2: 237575490-237575491 on Build GRCh38
rs35980448 Chr.2: 237575566-237575567 on Build GRCh38
rs950520054 Chr.2: 237575578-237575578 on Build GRCh38
rs116394440 Chr.2: 237575430-237575430 on Build GRCh38
rs994623927 Chr.2: 237575605-237575605 on Build GRCh38
rs551966752 Chr.2: 237575518-237575519 on Build GRCh38
rs919002129 Chr.2: 237575321-237575321 on Build GRCh38
rs987541068 Chr.2: 237575657-237575657 on Build GRCh38
rs566785323 Chr.2: 237575406-237575406 on Build GRCh38
rs367968871 Chr.2: 237575453-237575453 on Build GRCh38
rs761962024 Chr.2: 237575455-237575455 on Build GRCh38
rs893117711 Chr.2: 237575704-237575704 on Build GRCh38
rs928954955 Chr.2: 237575329-237575329 on Build GRCh38
rs926130545 Chr.2: 237575472-237575472 on Build GRCh38
rs553213262 Chr.2: 237575604-237575604 on Build GRCh38
rs747788987 Chr.2: 237575416-237575416 on Build GRCh38
rs571582972 Chr.2: 237575625-237575625 on Build GRCh38
rs767284036 Chr.2: 237575382-237575382 on Build GRCh38
rs41270747 Chr.2: 237575425-237575425 on Build GRCh38
rs771660639 Chr.2: 237575424-237575424 on Build GRCh38
rs140150815 Chr.2: 237575357-237575357 on Build GRCh38
rs765832427 Chr.2: 237575403-237575403 on Build GRCh38
rs1022904653 Chr.2: 237575535-237575535 on Build GRCh38
rs372061703 Chr.2: 237575378-237575378 on Build GRCh38
rs568969245 Chr.2: 237575431-237575431 on Build GRCh38
rs749422328 Chr.2: 237575438-237575438 on Build GRCh38
rs202010764 Chr.2: 237575460-237575460 on Build GRCh38
rs138383903 Chr.2: 237575394-237575394 on Build GRCh38
rs764596449 Chr.2: 237575508-237575508 on Build GRCh38
rs758083213 Chr.2: 237575517-237575517 on Build GRCh38
rs751697747 Chr.2: 237575377-237575377 on Build GRCh38
rs921521764 Chr.2: 237575673-237575673 on Build GRCh38
rs908923235 Chr.2: 237575355-237575355 on Build GRCh38
rs370434872 Chr.2: 237575342-237575342 on Build GRCh38
rs1026183388 Chr.2: 237575576-237575576 on Build GRCh38
rs936134211 Chr.2: 237575491-237575491 on Build GRCh38
rs756008721 Chr.2: 237575400-237575400 on Build GRCh38
rs202064784 Chr.2: 237575701-237575701 on Build GRCh38
rs746347621 Chr.2: 237575377-237575377 on Build GRCh38
rs1011906748 Chr.2: 237575325-237575325 on Build GRCh38
rs781705545 Chr.2: 237575524-237575524 on Build GRCh38
rs775838317 Chr.2: 237575433-237575433 on Build GRCh38
rs561367060 Chr.2: 237575320-237575320 on Build GRCh38
rs1053561730 Chr.2: 237575504-237575504 on Build GRCh38
rs537980921 Chr.2: 237575464-237575464 on Build GRCh38
rs370756013 Chr.2: 237575564-237575564 on Build GRCh38
rs763106085 Chr.2: 237575371-237575371 on Build GRCh38
rs1054555538 Chr.2: 237575703-237575703 on Build GRCh38
rs1012922223 Chr.2: 237575723-237575723 on Build GRCh38
rs947739458 Chr.2: 237575516-237575516 on Build GRCh38
rs754574468 Chr.2: 237575507-237575507 on Build GRCh38
rs967379608 Chr.2: 237575696-237575696 on Build GRCh38
rs60484328 Chr.2: 237575697-237575697 on Build GRCh38
rs533569172 Chr.2: 237575587-237575587 on Build GRCh38
rs13414095 Chr.2: 237575701-237575701 on Build GRCh38
rs984858415 Chr.2: 237575333-237575333 on Build GRCh38
rs774830303 Chr.2: 237575442-237575442 on Build GRCh38
rs768610341 Chr.2: 237575449-237575449 on Build GRCh38
rs767334922 Chr.2: 237575462-237575462 on Build GRCh38
rs78478325 Chr.2: 237575547-237575547 on Build GRCh38
rs147196366 Chr.2: 237575463-237575463 on Build GRCh38
rs545195707 Chr.2: 237575665-237575665 on Build GRCh38
rs778694655 Chr.2: 237575415-237575415 on Build GRCh38
rs367868362 Chr.2: 237575509-237575509 on Build GRCh38
rs766383536 Chr.2: 237575469-237575469 on Build GRCh38
rs10202062 Chr.2: 237575579-237575579 on Build GRCh38
rs1051698317 Chr.2: 237575693-237575693 on Build GRCh38
rs146887202 Chr.2: 237575707-237575707 on Build GRCh38
rs769998175 Chr.2: 237575349-237575349 on Build GRCh38
rs1022852245 Chr.2: 237575771-237575771 on Build GRCh38
rs899032817 Chr.2: 237575588-237575588 on Build GRCh38
rs573206612 Chr.2: 237575627-237575627 on Build GRCh38
rs529923135 Chr.2: 237575332-237575332 on Build GRCh38
rs934294075 Chr.2: 237575678-237575678 on Build GRCh38
rs200427826 Chr.2: 237575465-237575465 on Build GRCh38
rs764424565 Chr.2: 237575372-237575372 on Build GRCh38
rs760855976 Chr.2: 237575330-237575330 on Build GRCh38
rs565180815 Chr.2: 237575677-237575677 on Build GRCh38
rs753715396 Chr.2: 237575490-237575490 on Build GRCh38
rs953467510 Chr.2: 237575631-237575631 on Build GRCh38
rs1004762980 Chr.2: 237575340-237575340 on Build GRCh38
rs1029075877 Chr.2: 237575615-237575615 on Build GRCh38
rs527608420 Chr.2: 237575648-237575648 on Build GRCh38
rs757711254 Chr.2: 237575418-237575418 on Build GRCh38

View Full Product Details