Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238426866-238427391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB1
Entrez Gene ID
51665
Gene Name
ankyrin repeat and SOCS box containing 1
Gene Aliases
ASB-1
UniGene
Hs.516788
Gene Chromosome Location
Chr.2: 238426741-238452250 on Build GRCh38
Transcripts
NM_001040445.1 XM_005246080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6241507 c.14G>T p.G5V Chr.2: 238427084-238427084 on Build GRCh38
6501944 c.45C>T p.S15S Chr.2: 238427115-238427115 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs996464236 Chr.2: 238427198-238427198 on Build GRCh38
rs1013713109 Chr.2: 238426910-238426910 on Build GRCh38
rs932245298 Chr.2: 238427003-238427011 on Build GRCh38
rs887927899 Chr.2: 238427177-238427177 on Build GRCh38
rs1029945835 Chr.2: 238427098-238427099 on Build GRCh38
rs28588698 Chr.2: 238427363-238427363 on Build GRCh38
rs746993463 Chr.2: 238427362-238427362 on Build GRCh38
rs949486218 Chr.2: 238427025-238427025 on Build GRCh38
rs758562718 Chr.2: 238427048-238427048 on Build GRCh38
rs1006813414 Chr.2: 238427189-238427189 on Build GRCh38
rs888055869 Chr.2: 238427345-238427345 on Build GRCh38
rs975074580 Chr.2: 238426989-238426989 on Build GRCh38
rs896521656 Chr.2: 238427013-238427013 on Build GRCh38
rs958552387 Chr.2: 238427369-238427369 on Build GRCh38
rs1029316513 Chr.2: 238427203-238427203 on Build GRCh38
rs915996279 Chr.2: 238427244-238427244 on Build GRCh38
rs112204292 Chr.2: 238427255-238427255 on Build GRCh38
rs929309164 Chr.2: 238427325-238427325 on Build GRCh38
rs1037830692 Chr.2: 238427260-238427260 on Build GRCh38
rs968883598 Chr.2: 238427137-238427137 on Build GRCh38
rs990413793 Chr.2: 238427236-238427236 on Build GRCh38
rs545535041 Chr.2: 238426966-238426966 on Build GRCh38
rs370485939 Chr.2: 238427095-238427095 on Build GRCh38
rs978609288 Chr.2: 238426976-238426976 on Build GRCh38
rs955045758 Chr.2: 238427208-238427208 on Build GRCh38
rs913041739 Chr.2: 238426968-238426968 on Build GRCh38
rs990224841 Chr.2: 238427092-238427092 on Build GRCh38
rs546848246 Chr.2: 238427054-238427054 on Build GRCh38
rs1045801369 Chr.2: 238427097-238427097 on Build GRCh38
rs527262869 Chr.2: 238427146-238427146 on Build GRCh38
rs1046001589 Chr.2: 238426900-238426900 on Build GRCh38
rs890719696 Chr.2: 238427298-238427298 on Build GRCh38
rs900713252 Chr.2: 238427262-238427262 on Build GRCh38
rs372433030 Chr.2: 238427102-238427102 on Build GRCh38
rs34337994 Chr.2: 238427210-238427210 on Build GRCh38
rs551551395 Chr.2: 238427230-238427230 on Build GRCh38
rs570935150 Chr.2: 238427096-238427096 on Build GRCh38
rs899602105 Chr.2: 238427194-238427194 on Build GRCh38
rs1006867098 Chr.2: 238426931-238426931 on Build GRCh38
rs907477503 Chr.2: 238426919-238426919 on Build GRCh38
rs569233993 Chr.2: 238427174-238427174 on Build GRCh38
rs950942382 Chr.2: 238426930-238426930 on Build GRCh38
rs929509115 Chr.2: 238427125-238427125 on Build GRCh38
rs942120992 Chr.2: 238427238-238427238 on Build GRCh38
rs924431946 Chr.2: 238427051-238427051 on Build GRCh38
rs957217080 Chr.2: 238427221-238427221 on Build GRCh38
rs896534276 Chr.2: 238426899-238426899 on Build GRCh38
rs1005262187 Chr.2: 238426956-238426956 on Build GRCh38
rs955329786 Chr.2: 238427005-238427005 on Build GRCh38
rs1032022268 Chr.2: 238427220-238427220 on Build GRCh38
rs922502506 Chr.2: 238426995-238426995 on Build GRCh38
rs71426528 Chr.2: 238426934-238426934 on Build GRCh38
rs927151964 Chr.2: 238427150-238427150 on Build GRCh38
rs1006318585 Chr.2: 238427090-238427090 on Build GRCh38
rs894454877 Chr.2: 238426893-238426893 on Build GRCh38
rs988946809 Chr.2: 238427122-238427122 on Build GRCh38
rs935853093 Chr.2: 238427083-238427083 on Build GRCh38
rs374121868 Chr.2: 238427295-238427295 on Build GRCh38
rs974345562 Chr.2: 238426942-238426942 on Build GRCh38
rs954428887 Chr.2: 238427111-238427111 on Build GRCh38
rs952319405 Chr.2: 238426953-238426953 on Build GRCh38
rs947262882 Chr.2: 238426976-238426977 on Build GRCh38
rs948817800 Chr.2: 238427259-238427259 on Build GRCh38
rs564228023 Chr.2: 238426982-238426982 on Build GRCh38
rs777993118 Chr.2: 238427289-238427289 on Build GRCh38
rs1026892782 Chr.2: 238426920-238426920 on Build GRCh38
rs370306772 Chr.2: 238427341-238427341 on Build GRCh38
rs978553803 Chr.2: 238427149-238427149 on Build GRCh38
rs1048449893 Chr.2: 238427157-238427157 on Build GRCh38
rs936067688 Chr.2: 238426887-238426887 on Build GRCh38
rs977317798 Chr.2: 238427037-238427037 on Build GRCh38
rs6711952 Chr.2: 238426964-238426964 on Build GRCh38
rs550984560 Chr.2: 238427117-238427117 on Build GRCh38
rs968496446 Chr.2: 238427344-238427344 on Build GRCh38
rs1013349367 Chr.2: 238426898-238426898 on Build GRCh38
rs777905978 Chr.2: 238427082-238427082 on Build GRCh38
rs1017113083 Chr.2: 238426962-238426962 on Build GRCh38
rs1047947901 Chr.2: 238427338-238427338 on Build GRCh38
rs528237776 Chr.2: 238427008-238427008 on Build GRCh38
rs1016393999 Chr.2: 238427094-238427094 on Build GRCh38
rs1026520020 Chr.2: 238426941-238426941 on Build GRCh38
rs927161903 Chr.2: 238426986-238426986 on Build GRCh38
rs1054952690 Chr.2: 238426890-238426890 on Build GRCh38
rs990774110 Chr.2: 238427025-238427026 on Build GRCh38
rs907447732 Chr.2: 238427124-238427124 on Build GRCh38
rs757400364 Chr.2: 238427319-238427319 on Build GRCh38
rs1016668266 Chr.2: 238427190-238427190 on Build GRCh38

View Full Product Details