Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241189925-241190415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO7
Entrez Gene ID
50636
Gene Name
anoctamin 7
Gene Aliases
D-TMPP, DTMPP, IPCA-5, IPCA5, NGEP, PCANAP5, PCANAP5L, TMEM16G
UniGene
Hs.163909
Gene Chromosome Location
Chr.2: 241188509-241239602 on Build GRCh38
Transcripts
NM_001001666.3 NM_001001891.3 XM_011511263.2 XM_011511267.2 XM_017004229.1 XM_017004230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4536411 c.229G>A p.E77K Chr.2: 241190130-241190130 on Build GRCh38
4311849 c.261C>T p.H87H Chr.2: 241190162-241190162 on Build GRCh38
1018870 c.173G>A p.R58Q Chr.2: 241190074-241190074 on Build GRCh38
6399022 c.159C>A p.S53R Chr.2: 241190060-241190060 on Build GRCh38
4411828 c.206T>C p.I69T Chr.2: 241190107-241190107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771494446 Chr.2: 241190173-241190173 on Build GRCh38
rs777904354 Chr.2: 241190136-241190136 on Build GRCh38
rs4675985 Chr.2: 241190354-241190354 on Build GRCh38
rs200080175 Chr.2: 241190034-241190034 on Build GRCh38
rs569629834 Chr.2: 241190285-241190285 on Build GRCh38
rs754323255 Chr.2: 241190053-241190053 on Build GRCh38
rs952261493 Chr.2: 241190001-241190001 on Build GRCh38
rs751098531 Chr.2: 241190039-241190039 on Build GRCh38
rs769595495 Chr.2: 241190083-241190083 on Build GRCh38
rs201884340 Chr.2: 241190073-241190073 on Build GRCh38
rs745742731 Chr.2: 241190080-241190080 on Build GRCh38
rs866549830 Chr.2: 241190130-241190130 on Build GRCh38
rs969886969 Chr.2: 241190244-241190244 on Build GRCh38
rs1040926009 Chr.2: 241190364-241190364 on Build GRCh38
rs752979533 Chr.2: 241190070-241190070 on Build GRCh38
rs552003479 Chr.2: 241190349-241190349 on Build GRCh38
rs200885969 Chr.2: 241190178-241190178 on Build GRCh38
rs999133773 Chr.2: 241190195-241190195 on Build GRCh38
rs775132010 Chr.2: 241190033-241190033 on Build GRCh38
rs923464668 Chr.2: 241190372-241190372 on Build GRCh38
rs11695874 Chr.2: 241190072-241190072 on Build GRCh38
rs763725932 Chr.2: 241190037-241190037 on Build GRCh38
rs558745232 Chr.2: 241190065-241190065 on Build GRCh38
rs553293936 Chr.2: 241190382-241190382 on Build GRCh38
rs150023062 Chr.2: 241190138-241190138 on Build GRCh38
rs1003136568 Chr.2: 241190258-241190258 on Build GRCh38
rs570599792 Chr.2: 241190350-241190350 on Build GRCh38
rs1019617430 Chr.2: 241190012-241190012 on Build GRCh38
rs932119756 Chr.2: 241190394-241190394 on Build GRCh38
rs765882040 Chr.2: 241190182-241190182 on Build GRCh38
rs763430518 Chr.2: 241190192-241190192 on Build GRCh38
rs919256901 Chr.2: 241190337-241190337 on Build GRCh38
rs367770225 Chr.2: 241190119-241190119 on Build GRCh38
rs370288925 Chr.2: 241190160-241190160 on Build GRCh38
rs866019321 Chr.2: 241190167-241190167 on Build GRCh38
rs537185089 Chr.2: 241190297-241190297 on Build GRCh38
rs758800409 Chr.2: 241190135-241190135 on Build GRCh38
rs143588052 Chr.2: 241190076-241190076 on Build GRCh38
rs907397744 Chr.2: 241190170-241190170 on Build GRCh38
rs572772782 Chr.2: 241190198-241190198 on Build GRCh38
rs769704521 Chr.2: 241190117-241190117 on Build GRCh38
rs776114773 Chr.2: 241190187-241190187 on Build GRCh38
rs2302054 Chr.2: 241190100-241190100 on Build GRCh38
rs537939066 Chr.2: 241190027-241190027 on Build GRCh38
rs189471056 Chr.2: 241190102-241190102 on Build GRCh38
rs757291441 Chr.2: 241190143-241190143 on Build GRCh38
rs377407324 Chr.2: 241190315-241190315 on Build GRCh38
rs760062062 Chr.2: 241190118-241190118 on Build GRCh38
rs374671478 Chr.2: 241190111-241190111 on Build GRCh38
rs748248235 Chr.2: 241190108-241190108 on Build GRCh38
rs1035509279 Chr.2: 241190296-241190296 on Build GRCh38
rs148628229 Chr.2: 241190162-241190162 on Build GRCh38
rs151218672 Chr.2: 241190040-241190040 on Build GRCh38
rs148106236 Chr.2: 241190077-241190077 on Build GRCh38
rs756164274 Chr.2: 241190222-241190222 on Build GRCh38
rs61749471 Chr.2: 241190168-241190168 on Build GRCh38
rs868738251 Chr.2: 241190342-241190342 on Build GRCh38
rs2302053 Chr.2: 241190030-241190030 on Build GRCh38
rs758751051 Chr.2: 241190071-241190071 on Build GRCh38
rs371479862 Chr.2: 241190147-241190147 on Build GRCh38
rs971526115 Chr.2: 241190023-241190023 on Build GRCh38
rs34069570 Chr.2: 241190109-241190109 on Build GRCh38
rs770838286 Chr.2: 241190013-241190013 on Build GRCh38
rs759295773 Chr.2: 241190025-241190025 on Build GRCh38
rs766844190 Chr.2: 241190046-241190046 on Build GRCh38
rs373712262 Chr.2: 241190215-241190215 on Build GRCh38
rs543636867 Chr.2: 241190148-241190148 on Build GRCh38
rs775324576 Chr.2: 241190087-241190087 on Build GRCh38
rs373077663 Chr.2: 241189953-241189953 on Build GRCh38
rs996164948 Chr.2: 241189952-241189952 on Build GRCh38
rs529634514 Chr.2: 241190243-241190243 on Build GRCh38
rs779353305 Chr.2: 241190067-241190067 on Build GRCh38
rs761253562 Chr.2: 241190108-241190108 on Build GRCh38
rs371262700 Chr.2: 241190095-241190095 on Build GRCh38
rs1005090007 Chr.2: 241190008-241190008 on Build GRCh38
rs773907613 Chr.2: 241190163-241190163 on Build GRCh38
rs535849624 Chr.2: 241189944-241189944 on Build GRCh38
rs369590766 Chr.2: 241190199-241190199 on Build GRCh38
rs781278110 Chr.2: 241190145-241190145 on Build GRCh38
rs910640783 Chr.2: 241190354-241190358 on Build GRCh38
rs1028332520 Chr.2: 241189961-241189961 on Build GRCh38
rs753175781 Chr.2: 241190121-241190121 on Build GRCh38
rs972657457 Chr.2: 241190331-241190331 on Build GRCh38
rs371321043 Chr.2: 241189976-241189976 on Build GRCh38
rs776596277 Chr.2: 241190021-241190021 on Build GRCh38
rs548218549 Chr.2: 241190245-241190245 on Build GRCh38
rs559632164 Chr.2: 241190217-241190217 on Build GRCh38
rs978025342 Chr.2: 241190283-241190283 on Build GRCh38
rs768339208 Chr.2: 241190099-241190099 on Build GRCh38
rs760256346 Chr.2: 241190179-241190179 on Build GRCh38
rs775557322 Chr.2: 241190057-241190057 on Build GRCh38

View Full Product Details