Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241356077-241356593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARP2
Entrez Gene ID
9855
Gene Name
FERM, ARH/RhoGEF and pleckstrin domain protein 2
Gene Aliases
FIR, FRG, PLEKHC3
UniGene
Hs.726316
Gene Chromosome Location
Chr.2: 241356249-241494842 on Build GRCh38
Transcripts
NM_001282983.1 NM_001282984.1 NM_014808.3 XM_005247050.3 XM_011512233.1 XM_017005416.1 XM_017005417.1 XM_017005419.1 XM_017005420.1

SNP Detail

SNP ID Chromosome Location
rs1041150577 Chr.2: 241356500-241356500 on Build GRCh38
rs897336760 Chr.2: 241356572-241356572 on Build GRCh38
rs898148483 Chr.2: 241356179-241356179 on Build GRCh38
rs371240585 Chr.2: 241356542-241356542 on Build GRCh38
rs922551916 Chr.2: 241356550-241356550 on Build GRCh38
rs967278294 Chr.2: 241356165-241356165 on Build GRCh38
rs1019499848 Chr.2: 241356273-241356273 on Build GRCh38
rs1028386120 Chr.2: 241356204-241356204 on Build GRCh38
rs2286278 Chr.2: 241356233-241356233 on Build GRCh38
rs934321508 Chr.2: 241356365-241356365 on Build GRCh38
rs1003362219 Chr.2: 241356481-241356481 on Build GRCh38
rs981656377 Chr.2: 241356298-241356298 on Build GRCh38
rs182421171 Chr.2: 241356355-241356355 on Build GRCh38
rs996675575 Chr.2: 241356277-241356277 on Build GRCh38
rs1002962770 Chr.2: 241356557-241356557 on Build GRCh38
rs982349667 Chr.2: 241356239-241356239 on Build GRCh38
rs997119928 Chr.2: 241356414-241356414 on Build GRCh38
rs902452942 Chr.2: 241356259-241356259 on Build GRCh38
rs560889552 Chr.2: 241356481-241356481 on Build GRCh38
rs984074210 Chr.2: 241356229-241356229 on Build GRCh38
rs1053727943 Chr.2: 241356513-241356513 on Build GRCh38
rs923803170 Chr.2: 241356508-241356508 on Build GRCh38
rs538353929 Chr.2: 241356556-241356556 on Build GRCh38
rs959645917 Chr.2: 241356487-241356487 on Build GRCh38
rs938169148 Chr.2: 241356257-241356257 on Build GRCh38
rs1036730587 Chr.2: 241356178-241356178 on Build GRCh38
rs902864541 Chr.2: 241356356-241356356 on Build GRCh38
rs368771269 Chr.2: 241356424-241356424 on Build GRCh38
rs901078526 Chr.2: 241356397-241356397 on Build GRCh38
rs1019869813 Chr.2: 241356168-241356168 on Build GRCh38
rs1021580278 Chr.2: 241356150-241356150 on Build GRCh38
rs888243954 Chr.2: 241356431-241356431 on Build GRCh38
rs982788395 Chr.2: 241356309-241356309 on Build GRCh38
rs1042743051 Chr.2: 241356269-241356269 on Build GRCh38
rs1041386701 Chr.2: 241356385-241356385 on Build GRCh38
rs578115837 Chr.2: 241356333-241356333 on Build GRCh38
rs1035713592 Chr.2: 241356483-241356483 on Build GRCh38
rs545622457 Chr.2: 241356338-241356338 on Build GRCh38
rs973393070 Chr.2: 241356307-241356307 on Build GRCh38
rs908836821 Chr.2: 241356232-241356232 on Build GRCh38
rs915421467 Chr.2: 241356489-241356489 on Build GRCh38
rs928134176 Chr.2: 241356253-241356253 on Build GRCh38
rs965208280 Chr.2: 241356174-241356174 on Build GRCh38
rs961329083 Chr.2: 241356291-241356291 on Build GRCh38
rs1040709438 Chr.2: 241356539-241356539 on Build GRCh38
rs915702656 Chr.2: 241356345-241356345 on Build GRCh38
rs991441274 Chr.2: 241356488-241356488 on Build GRCh38
rs544319482 Chr.2: 241356324-241356324 on Build GRCh38
rs2286279 Chr.2: 241356106-241356106 on Build GRCh38
rs1011228444 Chr.2: 241356107-241356107 on Build GRCh38
rs141174816 Chr.2: 241356154-241356154 on Build GRCh38
rs902813801 Chr.2: 241356271-241356271 on Build GRCh38
rs931089919 Chr.2: 241356196-241356196 on Build GRCh38
rs1005602403 Chr.2: 241356440-241356440 on Build GRCh38
rs571361733 Chr.2: 241356532-241356532 on Build GRCh38
rs928783502 Chr.2: 241356302-241356302 on Build GRCh38
rs947069133 Chr.2: 241356268-241356268 on Build GRCh38
rs1055975508 Chr.2: 241356260-241356260 on Build GRCh38
rs1015555461 Chr.2: 241356286-241356286 on Build GRCh38
rs748020483 Chr.2: 241356510-241356510 on Build GRCh38
rs978526159 Chr.2: 241356521-241356521 on Build GRCh38
rs894216592 Chr.2: 241356265-241356265 on Build GRCh38
rs994723855 Chr.2: 241356573-241356573 on Build GRCh38
rs1044410625 Chr.2: 241356533-241356533 on Build GRCh38
rs928218987 Chr.2: 241356314-241356314 on Build GRCh38
rs959716632 Chr.2: 241356332-241356357 on Build GRCh38
rs893151354 Chr.2: 241356417-241356417 on Build GRCh38
rs975918921 Chr.2: 241356201-241356201 on Build GRCh38
rs947123149 Chr.2: 241356353-241356353 on Build GRCh38
rs984129569 Chr.2: 241356285-241356285 on Build GRCh38
rs902966778 Chr.2: 241356427-241356427 on Build GRCh38
rs1028856319 Chr.2: 241356280-241356280 on Build GRCh38
rs1049467423 Chr.2: 241356203-241356203 on Build GRCh38
rs1036150176 Chr.2: 241356559-241356559 on Build GRCh38
rs932591837 Chr.2: 241356562-241356562 on Build GRCh38
rs1015606352 Chr.2: 241356455-241356455 on Build GRCh38
rs1041380311 Chr.2: 241356251-241356251 on Build GRCh38
rs549621012 Chr.2: 241356514-241356514 on Build GRCh38
rs952792246 Chr.2: 241356281-241356281 on Build GRCh38
rs924341485 Chr.2: 241356528-241356528 on Build GRCh38
rs999186603 Chr.2: 241356272-241356272 on Build GRCh38
rs998729582 Chr.2: 241356166-241356166 on Build GRCh38
rs938206498 Chr.2: 241356319-241356319 on Build GRCh38
rs185454962 Chr.2: 241356420-241356420 on Build GRCh38
rs531285692 Chr.2: 241356482-241356482 on Build GRCh38
rs549682579 Chr.2: 241356461-241356461 on Build GRCh38
rs562952681 Chr.2: 241356329-241356329 on Build GRCh38
rs1011624388 Chr.2: 241356274-241356274 on Build GRCh38
rs901021784 Chr.2: 241356276-241356276 on Build GRCh38
rs1054075875 Chr.2: 241356383-241356383 on Build GRCh38
rs952739894 Chr.2: 241356205-241356205 on Build GRCh38
rs943839605 Chr.2: 241356497-241356497 on Build GRCh38
rs1049601407 Chr.2: 241356565-241356565 on Build GRCh38

View Full Product Details