Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241686341-241686884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTYMK
Entrez Gene ID
1841
Gene Name
deoxythymidylate kinase
Gene Aliases
CDC8, PP3731, TMPK, TYMK
UniGene
Hs.471873
Gene Chromosome Location
Chr.2: 241675742-241686968 on Build GRCh38
Transcripts
NM_001165031.1 NM_001320902.1 NM_001320903.1 NM_001320904.1 NM_001320905.1 NM_012145.3 NR_033255.1 NR_135492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6157797 c.6G>T p.A2A Chr.2: 241686778-241686778 on Build GRCh38
3771843 c.19G>T p.A7S Chr.2: 241686765-241686765 on Build GRCh38
4596823 c.16G>A p.G6R Chr.2: 241686768-241686768 on Build GRCh38
5555347 c.102C>T p.G34G Chr.2: 241686682-241686682 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775312315 Chr.2: 241686633-241686633 on Build GRCh38
rs1040467237 Chr.2: 241686448-241686448 on Build GRCh38
rs757368570 Chr.2: 241686618-241686618 on Build GRCh38
rs763538564 Chr.2: 241686768-241686768 on Build GRCh38
rs763214806 Chr.2: 241686647-241686648 on Build GRCh38
rs11555213 Chr.2: 241686681-241686681 on Build GRCh38
rs765871836 Chr.2: 241686691-241686691 on Build GRCh38
rs946743253 Chr.2: 241686390-241686390 on Build GRCh38
rs923912912 Chr.2: 241686405-241686405 on Build GRCh38
rs757412726 Chr.2: 241686710-241686710 on Build GRCh38
rs562230284 Chr.2: 241686858-241686858 on Build GRCh38
rs540945389 Chr.2: 241686845-241686845 on Build GRCh38
rs767123575 Chr.2: 241686649-241686649 on Build GRCh38
rs978991979 Chr.2: 241686515-241686515 on Build GRCh38
rs777297560 Chr.2: 241686805-241686805 on Build GRCh38
rs1017174801 Chr.2: 241686398-241686398 on Build GRCh38
rs771863067 Chr.2: 241686802-241686802 on Build GRCh38
rs887013063 Chr.2: 241686370-241686371 on Build GRCh38
rs959848262 Chr.2: 241686592-241686592 on Build GRCh38
rs1022472985 Chr.2: 241686796-241686796 on Build GRCh38
rs370711359 Chr.2: 241686735-241686735 on Build GRCh38
rs771602837 Chr.2: 241686754-241686754 on Build GRCh38
rs780311405 Chr.2: 241686627-241686627 on Build GRCh38
rs541885726 Chr.2: 241686621-241686621 on Build GRCh38
rs1042422898 Chr.2: 241686401-241686401 on Build GRCh38
rs763591671 Chr.2: 241686821-241686821 on Build GRCh38
rs373456554 Chr.2: 241686445-241686446 on Build GRCh38
rs576938061 Chr.2: 241686425-241686426 on Build GRCh38
rs758841162 Chr.2: 241686699-241686699 on Build GRCh38
rs769019188 Chr.2: 241686823-241686823 on Build GRCh38
rs761359926 Chr.2: 241686644-241686644 on Build GRCh38
rs901023275 Chr.2: 241686478-241686478 on Build GRCh38
rs779996055 Chr.2: 241686747-241686747 on Build GRCh38
rs774013958 Chr.2: 241686750-241686750 on Build GRCh38
rs111965544 Chr.2: 241686523-241686523 on Build GRCh38
rs781369163 Chr.2: 241686712-241686712 on Build GRCh38
rs778819243 Chr.2: 241686792-241686792 on Build GRCh38
rs751863088 Chr.2: 241686704-241686704 on Build GRCh38
rs766543742 Chr.2: 241686776-241686777 on Build GRCh38
rs946265181 Chr.2: 241686509-241686509 on Build GRCh38
rs774870983 Chr.2: 241686834-241686834 on Build GRCh38
rs753622988 Chr.2: 241686366-241686366 on Build GRCh38
rs1015233151 Chr.2: 241686553-241686553 on Build GRCh38
rs968194268 Chr.2: 241686814-241686814 on Build GRCh38
rs7370850 Chr.2: 241686520-241686520 on Build GRCh38
rs747136824 Chr.2: 241686610-241686610 on Build GRCh38
rs564539865 Chr.2: 241686807-241686807 on Build GRCh38
rs933962626 Chr.2: 241686632-241686632 on Build GRCh38
rs756137538 Chr.2: 241686738-241686738 on Build GRCh38
rs75122573 Chr.2: 241686516-241686516 on Build GRCh38
rs975196783 Chr.2: 241686414-241686414 on Build GRCh38
rs987738144 Chr.2: 241686471-241686471 on Build GRCh38
rs28630677 Chr.2: 241686546-241686546 on Build GRCh38
rs34795545 Chr.2: 241686716-241686717 on Build GRCh38
rs944784068 Chr.2: 241686441-241686441 on Build GRCh38
rs758791909 Chr.2: 241686603-241686603 on Build GRCh38
rs751834968 Chr.2: 241686775-241686775 on Build GRCh38
rs770502073 Chr.2: 241686764-241686764 on Build GRCh38
rs760297067 Chr.2: 241686759-241686759 on Build GRCh38
rs568831972 Chr.2: 241686491-241686491 on Build GRCh38
rs978360754 Chr.2: 241686841-241686841 on Build GRCh38
rs762141486 Chr.2: 241686778-241686778 on Build GRCh38
rs376486491 Chr.2: 241686766-241686766 on Build GRCh38
rs768498689 Chr.2: 241686748-241686748 on Build GRCh38
rs753218438 Chr.2: 241686698-241686698 on Build GRCh38
rs954671833 Chr.2: 241686456-241686456 on Build GRCh38
rs1015715035 Chr.2: 241686687-241686702 on Build GRCh38
rs745912790 Chr.2: 241686714-241686714 on Build GRCh38
rs773311128 Chr.2: 241686633-241686633 on Build GRCh38
rs1040520559 Chr.2: 241686700-241686700 on Build GRCh38
rs890585857 Chr.2: 241686826-241686826 on Build GRCh38
rs921887682 Chr.2: 241686861-241686861 on Build GRCh38
rs767655288 Chr.2: 241686779-241686779 on Build GRCh38
rs756929774 Chr.2: 241686732-241686732 on Build GRCh38
rs747875141 Chr.2: 241686751-241686751 on Build GRCh38
rs780049385 Chr.2: 241686787-241686787 on Build GRCh38
rs555247939 Chr.2: 241686571-241686571 on Build GRCh38
rs979390866 Chr.2: 241686658-241686658 on Build GRCh38
rs753839699 Chr.2: 241686789-241686789 on Build GRCh38
rs996292274 Chr.2: 241686595-241686595 on Build GRCh38
rs961000805 Chr.2: 241686570-241686570 on Build GRCh38
rs563449193 Chr.2: 241686637-241686637 on Build GRCh38
rs1049583958 Chr.2: 241686760-241686760 on Build GRCh38
rs756190575 Chr.2: 241686786-241686786 on Build GRCh38
rs750602620 Chr.2: 241686785-241686785 on Build GRCh38
rs1016774256 Chr.2: 241686573-241686573 on Build GRCh38
rs915495732 Chr.2: 241686372-241686372 on Build GRCh38
rs772876127 Chr.2: 241686756-241686756 on Build GRCh38
rs754840879 Chr.2: 241686790-241686790 on Build GRCh38
rs550713289 Chr.2: 241686363-241686363 on Build GRCh38
rs573520579 Chr.2: 241686579-241686579 on Build GRCh38
rs778175645 Chr.2: 241686605-241686605 on Build GRCh38
rs760170346 Chr.2: 241686687-241686687 on Build GRCh38
rs899623354 Chr.2: 241686580-241686580 on Build GRCh38
rs949749568 Chr.2: 241686549-241686550 on Build GRCh38
rs768398398 Chr.2: 241686638-241686638 on Build GRCh38
rs747919089 Chr.2: 241686800-241686800 on Build GRCh38
rs1012373058 Chr.2: 241686794-241686794 on Build GRCh38
rs770620579 Chr.2: 241686808-241686808 on Build GRCh38
rs776272909 Chr.2: 241686819-241686819 on Build GRCh38
rs963526130 Chr.2: 241686411-241686411 on Build GRCh38
rs528759558 Chr.2: 241686843-241686843 on Build GRCh38
rs774056171 Chr.2: 241686641-241686641 on Build GRCh38
rs764583881 Chr.2: 241686773-241686773 on Build GRCh38
rs990997254 Chr.2: 241686599-241686599 on Build GRCh38
rs1028996099 Chr.2: 241686428-241686428 on Build GRCh38

View Full Product Details