Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 329166-329703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOX12
Entrez Gene ID
6666
Gene Name
SRY-box 12
Gene Aliases
SOX22
UniGene
Hs.43627
Gene Chromosome Location
Chr.20: 325571-330228 on Build GRCh38
Transcripts
NM_006943.3

SNP Detail

SNP ID Chromosome Location
rs1043413531 Chr.20: 329605-329605 on Build GRCh38
rs368955451 Chr.20: 329360-329360 on Build GRCh38
rs960113166 Chr.20: 329238-329238 on Build GRCh38
rs1044300521 Chr.20: 329200-329200 on Build GRCh38
rs536212154 Chr.20: 329274-329274 on Build GRCh38
rs532901133 Chr.20: 329527-329527 on Build GRCh38
rs1008266801 Chr.20: 329314-329314 on Build GRCh38
rs1018274632 Chr.20: 329573-329573 on Build GRCh38
rs945478521 Chr.20: 329508-329508 on Build GRCh38
rs574505291 Chr.20: 329482-329482 on Build GRCh38
rs551157171 Chr.20: 329375-329375 on Build GRCh38
rs992577957 Chr.20: 329413-329413 on Build GRCh38
rs913661975 Chr.20: 329536-329536 on Build GRCh38
rs994472969 Chr.20: 329666-329666 on Build GRCh38
rs764870075 Chr.20: 329358-329358 on Build GRCh38
rs954884905 Chr.20: 329307-329307 on Build GRCh38
rs905231046 Chr.20: 329240-329240 on Build GRCh38
rs975225783 Chr.20: 329334-329334 on Build GRCh38
rs376038400 Chr.20: 329266-329266 on Build GRCh38
rs776432819 Chr.20: 329219-329220 on Build GRCh38
rs948577791 Chr.20: 329437-329437 on Build GRCh38
rs763136295 Chr.20: 329244-329244 on Build GRCh38
rs984512836 Chr.20: 329288-329290 on Build GRCh38
rs1053877111 Chr.20: 329264-329264 on Build GRCh38
rs567357812 Chr.20: 329263-329263 on Build GRCh38
rs1000835030 Chr.20: 329257-329257 on Build GRCh38
rs980435578 Chr.20: 329463-329463 on Build GRCh38
rs963021331 Chr.20: 329369-329369 on Build GRCh38
rs544211389 Chr.20: 329440-329440 on Build GRCh38
rs925821627 Chr.20: 329464-329464 on Build GRCh38
rs552561809 Chr.20: 329437-329438 on Build GRCh38
rs55960158 Chr.20: 329282-329282 on Build GRCh38
rs767256687 Chr.20: 329298-329298 on Build GRCh38
rs892191111 Chr.20: 329377-329377 on Build GRCh38
rs769171825 Chr.20: 329616-329616 on Build GRCh38
rs892460307 Chr.20: 329501-329501 on Build GRCh38
rs532151671 Chr.20: 329560-329560 on Build GRCh38
rs539636618 Chr.20: 329497-329497 on Build GRCh38
rs147933544 Chr.20: 329517-329518 on Build GRCh38
rs558516535 Chr.20: 329385-329385 on Build GRCh38
rs773222418 Chr.20: 329217-329217 on Build GRCh38
rs748531812 Chr.20: 329655-329655 on Build GRCh38
rs1027143802 Chr.20: 329225-329225 on Build GRCh38
rs572959340 Chr.20: 329323-329323 on Build GRCh38
rs1001713915 Chr.20: 329211-329211 on Build GRCh38
rs6050323 Chr.20: 329625-329625 on Build GRCh38
rs372208473 Chr.20: 329520-329521 on Build GRCh38
rs777499590 Chr.20: 329378-329378 on Build GRCh38
rs922508934 Chr.20: 329517-329517 on Build GRCh38
rs752532967 Chr.20: 329338-329338 on Build GRCh38
rs1001746529 Chr.20: 329433-329433 on Build GRCh38
rs759526020 Chr.20: 329291-329291 on Build GRCh38
rs1051856704 Chr.20: 329526-329526 on Build GRCh38
rs922401110 Chr.20: 329355-329355 on Build GRCh38
rs917093230 Chr.20: 329426-329426 on Build GRCh38
rs13043818 Chr.20: 329386-329386 on Build GRCh38
rs992967925 Chr.20: 329248-329248 on Build GRCh38
rs1014826708 Chr.20: 329445-329445 on Build GRCh38
rs187518923 Chr.20: 329479-329479 on Build GRCh38
rs1025753135 Chr.20: 329475-329475 on Build GRCh38
rs950442083 Chr.20: 329389-329389 on Build GRCh38
rs951523276 Chr.20: 329478-329478 on Build GRCh38
rs115791418 Chr.20: 329667-329667 on Build GRCh38
rs747561305 Chr.20: 329555-329555 on Build GRCh38

View Full Product Details