Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 67000415-67000961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35D1
Entrez Gene ID
23169
Gene Name
solute carrier family 35 member D1
Gene Aliases
UGTREL7
UniGene
Hs.213642
Gene Chromosome Location
Chr.1: 66999332-67054397 on Build GRCh38
Transcripts
NM_015139.2 XM_006710478.1 XM_011541070.1

SNP Detail

SNP ID Chromosome Location
rs1011021492 Chr.1: 67000742-67000742 on Build GRCh38
rs564640749 Chr.1: 67000623-67000623 on Build GRCh38
rs958381775 Chr.1: 67000906-67000906 on Build GRCh38
rs1025336224 Chr.1: 67000870-67000870 on Build GRCh38
rs936714937 Chr.1: 67000532-67000532 on Build GRCh38
rs771499327 Chr.1: 67000503-67000503 on Build GRCh38
rs965176033 Chr.1: 67000465-67000465 on Build GRCh38
rs1027070992 Chr.1: 67000717-67000717 on Build GRCh38
rs547269205 Chr.1: 67000868-67000868 on Build GRCh38
rs939326825 Chr.1: 67000657-67000657 on Build GRCh38
rs1021483863 Chr.1: 67000457-67000457 on Build GRCh38
rs945351192 Chr.1: 67000919-67000919 on Build GRCh38
rs775962582 Chr.1: 67000909-67000909 on Build GRCh38
rs951319865 Chr.1: 67000444-67000444 on Build GRCh38
rs533678925 Chr.1: 67000760-67000760 on Build GRCh38
rs528082939 Chr.1: 67000921-67000921 on Build GRCh38
rs985319315 Chr.1: 67000500-67000500 on Build GRCh38
rs117769761 Chr.1: 67000522-67000522 on Build GRCh38
rs964979038 Chr.1: 67000506-67000506 on Build GRCh38
rs544769342 Chr.1: 67000622-67000622 on Build GRCh38
rs904325030 Chr.1: 67000793-67000793 on Build GRCh38
rs562202606 Chr.1: 67000529-67000529 on Build GRCh38
rs918328640 Chr.1: 67000539-67000539 on Build GRCh38
rs1010484693 Chr.1: 67000445-67000445 on Build GRCh38
rs892528360 Chr.1: 67000711-67000711 on Build GRCh38
rs145206449 Chr.1: 67000621-67000621 on Build GRCh38
rs975640756 Chr.1: 67000805-67000805 on Build GRCh38
rs938155347 Chr.1: 67000650-67000650 on Build GRCh38
rs965208480 Chr.1: 67000792-67000792 on Build GRCh38
rs951134098 Chr.1: 67000884-67000884 on Build GRCh38
rs114329922 Chr.1: 67000923-67000923 on Build GRCh38
rs1006684645 Chr.1: 67000811-67000811 on Build GRCh38
rs965304383 Chr.1: 67000853-67000853 on Build GRCh38
rs894201716 Chr.1: 67000694-67000694 on Build GRCh38
rs184748428 Chr.1: 67000924-67000924 on Build GRCh38
rs762534156 Chr.1: 67000755-67000755 on Build GRCh38
rs372562279 Chr.1: 67000840-67000840 on Build GRCh38
rs1022818272 Chr.1: 67000746-67000746 on Build GRCh38
rs570427498 Chr.1: 67000696-67000696 on Build GRCh38
rs982398338 Chr.1: 67000920-67000920 on Build GRCh38
rs1018137884 Chr.1: 67000835-67000835 on Build GRCh38
rs149155563 Chr.1: 67000842-67000842 on Build GRCh38
rs906792014 Chr.1: 67000647-67000647 on Build GRCh38
rs969511370 Chr.1: 67000912-67000912 on Build GRCh38
rs112800156 Chr.1: 67000838-67000838 on Build GRCh38
rs906731028 Chr.1: 67000635-67000635 on Build GRCh38
rs982625086 Chr.1: 67000447-67000447 on Build GRCh38
rs747719749 Chr.1: 67000477-67000477 on Build GRCh38
rs910023130 Chr.1: 67000595-67000595 on Build GRCh38
rs929763585 Chr.1: 67000540-67000540 on Build GRCh38
rs984681814 Chr.1: 67000573-67000573 on Build GRCh38
rs886743269 Chr.1: 67000734-67000734 on Build GRCh38
rs1019487555 Chr.1: 67000769-67000769 on Build GRCh38
rs976421658 Chr.1: 67000527-67000527 on Build GRCh38
rs376314733 Chr.1: 67000890-67000890 on Build GRCh38
rs913879327 Chr.1: 67000566-67000566 on Build GRCh38

View Full Product Details