Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6075958-6076457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT1
Entrez Gene ID
55612
Gene Name
fermitin family member 1
Gene Aliases
C20orf42, DTGCU2, KIND1, UNC112A, URP1
UniGene
Hs.472054
Gene Chromosome Location
Chr.20: 6074845-6123544 on Build GRCh38
Transcripts
NM_017671.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161046 c.1650A>T p.R550S Chr.20: 6076456-6076456 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs6139909 Chr.20: 6076018-6076018 on Build GRCh38
rs1043261835 Chr.20: 6076150-6076150 on Build GRCh38
rs527703164 Chr.20: 6076246-6076246 on Build GRCh38
rs6053878 Chr.20: 6076271-6076271 on Build GRCh38
rs775060736 Chr.20: 6076405-6076406 on Build GRCh38
rs549079931 Chr.20: 6076248-6076248 on Build GRCh38
rs188548418 Chr.20: 6076390-6076390 on Build GRCh38
rs550396481 Chr.20: 6076389-6076389 on Build GRCh38
rs879128472 Chr.20: 6076264-6076264 on Build GRCh38
rs578179571 Chr.20: 6076138-6076138 on Build GRCh38
rs988439414 Chr.20: 6076374-6076374 on Build GRCh38
rs781104279 Chr.20: 6076241-6076241 on Build GRCh38
rs41282956 Chr.20: 6076097-6076097 on Build GRCh38
rs373822971 Chr.20: 6076179-6076179 on Build GRCh38
rs969484281 Chr.20: 6076328-6076328 on Build GRCh38
rs750904222 Chr.20: 6076395-6076395 on Build GRCh38
rs1013570977 Chr.20: 6076298-6076298 on Build GRCh38
rs1052709630 Chr.20: 6076100-6076100 on Build GRCh38
rs938430063 Chr.20: 6076345-6076345 on Build GRCh38
rs969181299 Chr.20: 6076055-6076055 on Build GRCh38
rs748927308 Chr.20: 6076418-6076418 on Build GRCh38
rs565068120 Chr.20: 6076137-6076137 on Build GRCh38
rs918624771 Chr.20: 6076014-6076014 on Build GRCh38
rs970381681 Chr.20: 6076277-6076277 on Build GRCh38
rs368803013 Chr.20: 6076118-6076118 on Build GRCh38
rs755186326 Chr.20: 6076020-6076020 on Build GRCh38
rs886056897 Chr.20: 6076277-6076277 on Build GRCh38
rs899611340 Chr.20: 6076149-6076149 on Build GRCh38
rs1024997394 Chr.20: 6076310-6076310 on Build GRCh38
rs1006847575 Chr.20: 6076256-6076256 on Build GRCh38
rs543610282 Chr.20: 6076115-6076115 on Build GRCh38
rs769454885 Chr.20: 6076292-6076292 on Build GRCh38
rs374876522 Chr.20: 6076420-6076420 on Build GRCh38
rs879231203 Chr.20: 6076101-6076101 on Build GRCh38
rs1052824024 Chr.20: 6076351-6076351 on Build GRCh38
rs770860248 Chr.20: 6076213-6076213 on Build GRCh38
rs371657740 Chr.20: 6076430-6076430 on Build GRCh38
rs1040785409 Chr.20: 6076139-6076139 on Build GRCh38
rs767059789 Chr.20: 6076424-6076424 on Build GRCh38
rs957898941 Chr.20: 6076367-6076367 on Build GRCh38
rs370805467 Chr.20: 6076190-6076191 on Build GRCh38
rs1007022220 Chr.20: 6076164-6076164 on Build GRCh38
rs936470110 Chr.20: 6076305-6076305 on Build GRCh38
rs756695878 Chr.20: 6076423-6076423 on Build GRCh38
rs886056896 Chr.20: 6076222-6076222 on Build GRCh38
rs545499046 Chr.20: 6076159-6076159 on Build GRCh38
rs758757189 Chr.20: 6076323-6076323 on Build GRCh38
rs980565611 Chr.20: 6076334-6076334 on Build GRCh38
rs909025380 Chr.20: 6076051-6076051 on Build GRCh38
rs928489784 Chr.20: 6076304-6076304 on Build GRCh38
rs531871016 Chr.20: 6076375-6076375 on Build GRCh38
rs142538462 Chr.20: 6076027-6076027 on Build GRCh38
rs928364319 Chr.20: 6076074-6076074 on Build GRCh38
rs113475128 Chr.20: 6076275-6076275 on Build GRCh38
rs1044639904 Chr.20: 6076073-6076073 on Build GRCh38
rs775250903 Chr.20: 6076408-6076408 on Build GRCh38
rs936618603 Chr.20: 6076114-6076114 on Build GRCh38
rs147620065 Chr.20: 6076169-6076169 on Build GRCh38
rs762424632 Chr.20: 6076411-6076412 on Build GRCh38
rs8116177 Chr.20: 6076071-6076071 on Build GRCh38
rs1043120859 Chr.20: 6076401-6076401 on Build GRCh38

View Full Product Details