Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 14326326-14326872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLRT3
Entrez Gene ID
23767
Gene Name
fibronectin leucine rich transmembrane protein 3
Gene Aliases
HH21
UniGene
Hs.41296
Gene Chromosome Location
Chr.20: 14322985-14337847 on Build GRCh38
Transcripts
NM_013281.3 NM_198391.2 XM_005260682.4 XM_011529204.2 XM_011529205.2 NM_080676.5 XM_017027677.1

Gene Symbol
MACROD2
Entrez Gene ID
140733
Gene Name
MACRO domain containing 2
Gene Aliases
C20orf133
UniGene
Hs.661576
Gene Chromosome Location
Chr.20: 13995476-16053197 on Build GRCh38
Transcripts
NM_013281.3 NM_198391.2 XM_005260682.4 XM_011529204.2 XM_011529205.2 NM_080676.5 XM_017027677.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6665201 c.944G>A p.W315* Chr.20: 14326563-14326563 on Build GRCh38
281192 c.907C>T p.R303C Chr.20: 14326600-14326600 on Build GRCh38
1494983 c.1109T>A p.I370K Chr.20: 14326398-14326398 on Build GRCh38
3910877 c.917C>T p.P306L Chr.20: 14326590-14326590 on Build GRCh38
1410457 c.793A>G p.N265D Chr.20: 14326714-14326714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748518747 Chr.20: 14326544-14326544 on Build GRCh38
rs764742743 Chr.20: 14326783-14326783 on Build GRCh38
rs759473524 Chr.20: 14326428-14326428 on Build GRCh38
rs765363483 Chr.20: 14326643-14326643 on Build GRCh38
rs752008590 Chr.20: 14326667-14326667 on Build GRCh38
rs757500775 Chr.20: 14326823-14326823 on Build GRCh38
rs765357979 Chr.20: 14326757-14326757 on Build GRCh38
rs149124349 Chr.20: 14326676-14326676 on Build GRCh38
rs373003336 Chr.20: 14326610-14326610 on Build GRCh38
rs375651610 Chr.20: 14326555-14326555 on Build GRCh38
rs748572291 Chr.20: 14326394-14326394 on Build GRCh38
rs376150945 Chr.20: 14326725-14326725 on Build GRCh38
rs762893918 Chr.20: 14326580-14326580 on Build GRCh38
rs944637009 Chr.20: 14326485-14326485 on Build GRCh38
rs769769921 Chr.20: 14326561-14326561 on Build GRCh38
rs763048882 Chr.20: 14326558-14326558 on Build GRCh38
rs746662796 Chr.20: 14326410-14326410 on Build GRCh38
rs369748001 Chr.20: 14326599-14326599 on Build GRCh38
rs756352350 Chr.20: 14326489-14326489 on Build GRCh38
rs779566098 Chr.20: 14326716-14326716 on Build GRCh38
rs758993460 Chr.20: 14326656-14326656 on Build GRCh38
rs749185378 Chr.20: 14326718-14326718 on Build GRCh38
rs8120693 Chr.20: 14326378-14326378 on Build GRCh38
rs868467242 Chr.20: 14326796-14326796 on Build GRCh38
rs6033944 Chr.20: 14326469-14326469 on Build GRCh38
rs750500844 Chr.20: 14326365-14326365 on Build GRCh38
rs188277793 Chr.20: 14326382-14326382 on Build GRCh38
rs779346527 Chr.20: 14326524-14326524 on Build GRCh38
rs904764203 Chr.20: 14326766-14326766 on Build GRCh38
rs753941529 Chr.20: 14326604-14326604 on Build GRCh38
rs371671990 Chr.20: 14326411-14326411 on Build GRCh38
rs147137056 Chr.20: 14326686-14326686 on Build GRCh38
rs747581282 Chr.20: 14326364-14326365 on Build GRCh38
rs775345656 Chr.20: 14326437-14326437 on Build GRCh38
rs774163669 Chr.20: 14326589-14326589 on Build GRCh38
rs781633669 Chr.20: 14326828-14326828 on Build GRCh38
rs140514613 Chr.20: 14326366-14326366 on Build GRCh38
rs141238925 Chr.20: 14326559-14326559 on Build GRCh38
rs139715667 Chr.20: 14326661-14326661 on Build GRCh38
rs755488912 Chr.20: 14326518-14326518 on Build GRCh38
rs151151641 Chr.20: 14326769-14326769 on Build GRCh38
rs750588557 Chr.20: 14326457-14326457 on Build GRCh38
rs148251977 Chr.20: 14326557-14326557 on Build GRCh38
rs377483086 Chr.20: 14326622-14326622 on Build GRCh38
rs1054484581 Chr.20: 14326605-14326605 on Build GRCh38
rs758767646 Chr.20: 14326553-14326553 on Build GRCh38
rs6135119 Chr.20: 14326660-14326660 on Build GRCh38
rs950461244 Chr.20: 14326825-14326825 on Build GRCh38
rs144067037 Chr.20: 14326461-14326461 on Build GRCh38
rs768621968 Chr.20: 14326720-14326720 on Build GRCh38
rs143179643 Chr.20: 14326742-14326742 on Build GRCh38
rs774110506 Chr.20: 14326721-14326721 on Build GRCh38
rs1056466152 Chr.20: 14326712-14326712 on Build GRCh38
rs778241174 Chr.20: 14326664-14326664 on Build GRCh38
rs150799973 Chr.20: 14326600-14326600 on Build GRCh38
rs776650907 Chr.20: 14326730-14326730 on Build GRCh38
rs770506040 Chr.20: 14326418-14326418 on Build GRCh38
rs36034779 Chr.20: 14326373-14326373 on Build GRCh38
rs398124654 Chr.20: 14326491-14326491 on Build GRCh38
rs780778475 Chr.20: 14326679-14326679 on Build GRCh38
rs911759953 Chr.20: 14326477-14326477 on Build GRCh38
rs980632823 Chr.20: 14326474-14326474 on Build GRCh38
rs751955471 Chr.20: 14326806-14326806 on Build GRCh38
rs376427607 Chr.20: 14326436-14326436 on Build GRCh38
rs564318429 Chr.20: 14326726-14326726 on Build GRCh38
rs775343139 Chr.20: 14326579-14326579 on Build GRCh38
rs115086094 Chr.20: 14326403-14326403 on Build GRCh38
rs983233462 Chr.20: 14326476-14326476 on Build GRCh38
rs751523905 Chr.20: 14326655-14326655 on Build GRCh38
rs763883408 Chr.20: 14326456-14326456 on Build GRCh38
rs749996521 Chr.20: 14326840-14326840 on Build GRCh38
rs780013962 Chr.20: 14326372-14326372 on Build GRCh38
rs768394306 Chr.20: 14326585-14326585 on Build GRCh38
rs779306670 Chr.20: 14326381-14326381 on Build GRCh38
rs936019282 Chr.20: 14326577-14326577 on Build GRCh38
rs372667293 Chr.20: 14326454-14326454 on Build GRCh38
rs776024502 Chr.20: 14326426-14326426 on Build GRCh38
rs753905921 Chr.20: 14326515-14326515 on Build GRCh38
rs76061726 Chr.20: 14326359-14326359 on Build GRCh38
rs6135120 Chr.20: 14326762-14326762 on Build GRCh38
rs562745017 Chr.20: 14326459-14326459 on Build GRCh38
rs771836971 Chr.20: 14326401-14326401 on Build GRCh38
rs201766195 Chr.20: 14326775-14326775 on Build GRCh38
rs1001761845 Chr.20: 14326695-14326695 on Build GRCh38
rs541810428 Chr.20: 14326523-14326523 on Build GRCh38
rs745543415 Chr.20: 14326684-14326684 on Build GRCh38
rs766443058 Chr.20: 14326511-14326511 on Build GRCh38

View Full Product Details