Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18487511-18488061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP9
Entrez Gene ID
10741
Gene Name
RB binding protein 9, serine hydrolase
Gene Aliases
BOG, RBBP10
UniGene
Hs.69330
Gene Chromosome Location
Chr.20: 18486540-18497250 on Build GRCh38
Transcripts
NM_006606.2 XM_005260652.2

SNP Detail

SNP ID Chromosome Location
rs913473985 Chr.20: 18487862-18487862 on Build GRCh38
rs897742472 Chr.20: 18488007-18488007 on Build GRCh38
rs879524861 Chr.20: 18487749-18487749 on Build GRCh38
rs993256814 Chr.20: 18487813-18487813 on Build GRCh38
rs988563364 Chr.20: 18487730-18487730 on Build GRCh38
rs752299515 Chr.20: 18487937-18487937 on Build GRCh38
rs147913483 Chr.20: 18487611-18487611 on Build GRCh38
rs968853758 Chr.20: 18487741-18487741 on Build GRCh38
rs946002038 Chr.20: 18487607-18487607 on Build GRCh38
rs149102477 Chr.20: 18488026-18488026 on Build GRCh38
rs895891912 Chr.20: 18487568-18487568 on Build GRCh38
rs542934458 Chr.20: 18487740-18487740 on Build GRCh38
rs79276132 Chr.20: 18488000-18488000 on Build GRCh38
rs560501020 Chr.20: 18487803-18487803 on Build GRCh38
rs979016828 Chr.20: 18487750-18487750 on Build GRCh38
rs886262826 Chr.20: 18487673-18487673 on Build GRCh38
rs957504734 Chr.20: 18487845-18487845 on Build GRCh38
rs767141832 Chr.20: 18487832-18487832 on Build GRCh38
rs542324272 Chr.20: 18487761-18487761 on Build GRCh38
rs968404247 Chr.20: 18487971-18487971 on Build GRCh38
rs900202126 Chr.20: 18487685-18487685 on Build GRCh38
rs199706406 Chr.20: 18487954-18487956 on Build GRCh38
rs993082434 Chr.20: 18488017-18488017 on Build GRCh38
rs190844098 Chr.20: 18487812-18487812 on Build GRCh38
rs925061574 Chr.20: 18487843-18487843 on Build GRCh38
rs946304727 Chr.20: 18487870-18487870 on Build GRCh38
rs990330281 Chr.20: 18487852-18487852 on Build GRCh38
rs890108281 Chr.20: 18487932-18487932 on Build GRCh38
rs557294705 Chr.20: 18487714-18487714 on Build GRCh38
rs139987376 Chr.20: 18487674-18487674 on Build GRCh38
rs568970619 Chr.20: 18488003-18488003 on Build GRCh38
rs997874625 Chr.20: 18488009-18488009 on Build GRCh38
rs930054879 Chr.20: 18487575-18487575 on Build GRCh38
rs930080868 Chr.20: 18487858-18487858 on Build GRCh38
rs1016188919 Chr.20: 18487684-18487684 on Build GRCh38
rs886313746 Chr.20: 18487868-18487868 on Build GRCh38
rs996245483 Chr.20: 18487706-18487706 on Build GRCh38
rs887875305 Chr.20: 18487969-18487969 on Build GRCh38
rs959227533 Chr.20: 18488014-18488014 on Build GRCh38
rs1030437976 Chr.20: 18487925-18487925 on Build GRCh38
rs900433762 Chr.20: 18487877-18487877 on Build GRCh38
rs941837161 Chr.20: 18487874-18487874 on Build GRCh38
rs530983192 Chr.20: 18488022-18488022 on Build GRCh38
rs565170441 Chr.20: 18487894-18487894 on Build GRCh38
rs544881642 Chr.20: 18488001-18488001 on Build GRCh38
rs917229569 Chr.20: 18487857-18487857 on Build GRCh38
rs185926307 Chr.20: 18487808-18487808 on Build GRCh38
rs1037507520 Chr.20: 18487875-18487875 on Build GRCh38
rs1029972371 Chr.20: 18487713-18487713 on Build GRCh38
rs145323684 Chr.20: 18487940-18487940 on Build GRCh38
rs1047161952 Chr.20: 18487864-18487864 on Build GRCh38

View Full Product Details