Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18527423-18527957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC23B
Entrez Gene ID
10483
Gene Name
Sec23 homolog B, coat complex II component
Gene Aliases
CDA-II, CDAII, CDAN2, CWS7, HEMPAS
UniGene
Hs.369373
Gene Chromosome Location
Chr.20: 18507544-18561415 on Build GRCh38
Transcripts
NM_001172745.1 NM_001172746.1 NM_006363.4 NM_032985.4 NM_032986.3 XM_017027593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6159125 c.1109G>T p.G370V Chr.20: 18527611-18527611 on Build GRCh38
1024907 c.1016G>A p.R339Q Chr.20: 18527518-18527518 on Build GRCh38
3799364 c.1066C>G p.Q356E Chr.20: 18527568-18527568 on Build GRCh38
5545754 c.1015C>T p.R339* Chr.20: 18527517-18527517 on Build GRCh38
1615372 c.1015C>G p.R339G Chr.20: 18527517-18527517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746779869 Chr.20: 18527634-18527634 on Build GRCh38
rs762005067 Chr.20: 18527491-18527491 on Build GRCh38
rs781104521 Chr.20: 18527588-18527588 on Build GRCh38
rs1034149881 Chr.20: 18527880-18527880 on Build GRCh38
rs575712961 Chr.20: 18527856-18527856 on Build GRCh38
rs7263397 Chr.20: 18527467-18527467 on Build GRCh38
rs754318349 Chr.20: 18527531-18527531 on Build GRCh38
rs761274385 Chr.20: 18527527-18527527 on Build GRCh38
rs745583678 Chr.20: 18527481-18527481 on Build GRCh38
rs145362985 Chr.20: 18527570-18527570 on Build GRCh38
rs140387877 Chr.20: 18527537-18527537 on Build GRCh38
rs933447279 Chr.20: 18527475-18527475 on Build GRCh38
rs150042408 Chr.20: 18527553-18527553 on Build GRCh38
rs1021739359 Chr.20: 18527622-18527622 on Build GRCh38
rs892002905 Chr.20: 18527532-18527532 on Build GRCh38
rs950615921 Chr.20: 18527751-18527751 on Build GRCh38
rs774678591 Chr.20: 18527609-18527609 on Build GRCh38
rs1054709239 Chr.20: 18527823-18527823 on Build GRCh38
rs538933592 Chr.20: 18527834-18527834 on Build GRCh38
rs371376726 Chr.20: 18527505-18527505 on Build GRCh38
rs1053289100 Chr.20: 18527477-18527477 on Build GRCh38
rs16979303 Chr.20: 18527711-18527711 on Build GRCh38
rs757193586 Chr.20: 18527580-18527580 on Build GRCh38
rs753056084 Chr.20: 18527643-18527643 on Build GRCh38
rs943006913 Chr.20: 18527898-18527898 on Build GRCh38
rs769456702 Chr.20: 18527607-18527607 on Build GRCh38
rs748353015 Chr.20: 18527610-18527610 on Build GRCh38
rs1033527300 Chr.20: 18527686-18527686 on Build GRCh38
rs776980592 Chr.20: 18527626-18527626 on Build GRCh38
rs891821857 Chr.20: 18527846-18527846 on Build GRCh38
rs758060427 Chr.20: 18527652-18527652 on Build GRCh38
rs781185323 Chr.20: 18527755-18527755 on Build GRCh38
rs1024418783 Chr.20: 18527870-18527870 on Build GRCh38
rs777288695 Chr.20: 18527565-18527565 on Build GRCh38
rs549175754 Chr.20: 18527635-18527635 on Build GRCh38
rs763670451 Chr.20: 18527657-18527657 on Build GRCh38
rs368960604 Chr.20: 18527517-18527517 on Build GRCh38
rs577659219 Chr.20: 18527734-18527734 on Build GRCh38
rs773426512 Chr.20: 18527618-18527618 on Build GRCh38
rs1049856968 Chr.20: 18527741-18527741 on Build GRCh38
rs779578318 Chr.20: 18527575-18527575 on Build GRCh38
rs748071441 Chr.20: 18527889-18527889 on Build GRCh38
rs974419704 Chr.20: 18527930-18527930 on Build GRCh38
rs753064087 Chr.20: 18527592-18527594 on Build GRCh38
rs764816865 Chr.20: 18527529-18527530 on Build GRCh38
rs760830538 Chr.20: 18527620-18527620 on Build GRCh38
rs951912875 Chr.20: 18527554-18527554 on Build GRCh38
rs534653213 Chr.20: 18527678-18527678 on Build GRCh38
rs557666284 Chr.20: 18527847-18527847 on Build GRCh38
rs746978431 Chr.20: 18527451-18527451 on Build GRCh38
rs960618255 Chr.20: 18527709-18527709 on Build GRCh38
rs17734573 Chr.20: 18527660-18527660 on Build GRCh38
rs769922478 Chr.20: 18527915-18527915 on Build GRCh38
rs1001381984 Chr.20: 18527876-18527876 on Build GRCh38
rs1026104501 Chr.20: 18527747-18527747 on Build GRCh38
rs767678738 Chr.20: 18527500-18527500 on Build GRCh38
rs772358387 Chr.20: 18527612-18527612 on Build GRCh38
rs764987635 Chr.20: 18527542-18527542 on Build GRCh38
rs940361454 Chr.20: 18527687-18527687 on Build GRCh38
rs1040556231 Chr.20: 18527697-18527697 on Build GRCh38
rs774426758 Chr.20: 18527486-18527486 on Build GRCh38
rs751138438 Chr.20: 18527658-18527658 on Build GRCh38
rs370161092 Chr.20: 18527606-18527606 on Build GRCh38
rs1000814810 Chr.20: 18527811-18527811 on Build GRCh38
rs908988030 Chr.20: 18527866-18527866 on Build GRCh38
rs768990257 Chr.20: 18527483-18527483 on Build GRCh38
rs767252820 Chr.20: 18527799-18527799 on Build GRCh38
rs765551971 Chr.20: 18527641-18527641 on Build GRCh38
rs971447139 Chr.20: 18527875-18527875 on Build GRCh38
rs767047492 Chr.20: 18527623-18527623 on Build GRCh38
rs758029042 Chr.20: 18527564-18527564 on Build GRCh38
rs758697091 Chr.20: 18527655-18527656 on Build GRCh38
rs766922478 Chr.20: 18527529-18527529 on Build GRCh38
rs770807409 Chr.20: 18527464-18527464 on Build GRCh38
rs984936938 Chr.20: 18527824-18527824 on Build GRCh38

View Full Product Details