Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 20512431-20512938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RALGAPA2
Entrez Gene ID
57186
Gene Name
Ral GTPase activating protein catalytic alpha subunit 2
Gene Aliases
AS250, C20orf74, bA287B20.1, dJ1049G11, dJ1049G11.4, p220
UniGene
Hs.472285
Gene Chromosome Location
Chr.20: 20389558-20712638 on Build GRCh38
Transcripts
NM_020343.3 XM_005260768.2 XM_006723598.1 XM_006723599.1 XM_011529309.1 XM_011529310.2 XM_011529311.2 XM_017027972.1 XM_017027973.1 XM_017027974.1 XM_017027975.1 XM_017027976.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5044193 c.4490C>A p.S1497* Chr.20: 20512879-20512879 on Build GRCh38
7203027 c.4703A>G p.N1568S Chr.20: 20512666-20512666 on Build GRCh38
5813555 c.4118A>T p.Q1373L Chr.20: 20512701-20512701 on Build GRCh38
5813552 c.4668A>T p.Q1556H Chr.20: 20512701-20512701 on Build GRCh38
3840554 c.3993G>A p.E1331E Chr.20: 20512826-20512826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775347855 Chr.20: 20512836-20512838 on Build GRCh38
rs1021047486 Chr.20: 20512646-20512646 on Build GRCh38
rs762693802 Chr.20: 20512741-20512741 on Build GRCh38
rs751872309 Chr.20: 20512533-20512533 on Build GRCh38
rs767720640 Chr.20: 20512743-20512743 on Build GRCh38
rs753913240 Chr.20: 20512642-20512642 on Build GRCh38
rs759284505 Chr.20: 20512467-20512467 on Build GRCh38
rs367755056 Chr.20: 20512867-20512867 on Build GRCh38
rs745836321 Chr.20: 20512804-20512804 on Build GRCh38
rs769385521 Chr.20: 20512811-20512811 on Build GRCh38
rs6046906 Chr.20: 20512555-20512555 on Build GRCh38
rs778982377 Chr.20: 20512672-20512672 on Build GRCh38
rs796997874 Chr.20: 20512701-20512701 on Build GRCh38
rs35039408 Chr.20: 20512578-20512578 on Build GRCh38
rs769475486 Chr.20: 20512736-20512736 on Build GRCh38
rs577898513 Chr.20: 20512759-20512759 on Build GRCh38
rs751875941 Chr.20: 20512913-20512913 on Build GRCh38
rs542368069 Chr.20: 20512878-20512878 on Build GRCh38
rs758472735 Chr.20: 20512907-20512907 on Build GRCh38
rs776774378 Chr.20: 20512730-20512730 on Build GRCh38
rs748829198 Chr.20: 20512836-20512836 on Build GRCh38
rs1007970796 Chr.20: 20512810-20512810 on Build GRCh38
rs780044972 Chr.20: 20512629-20512629 on Build GRCh38
rs768352542 Chr.20: 20512837-20512837 on Build GRCh38
rs776704308 Chr.20: 20512903-20512903 on Build GRCh38
rs758659819 Chr.20: 20512780-20512780 on Build GRCh38
rs762057126 Chr.20: 20512610-20512610 on Build GRCh38
rs886855360 Chr.20: 20512819-20512819 on Build GRCh38
rs781533887 Chr.20: 20512543-20512543 on Build GRCh38
rs1046808306 Chr.20: 20512466-20512466 on Build GRCh38
rs757704370 Chr.20: 20512534-20512534 on Build GRCh38
rs555401831 Chr.20: 20512692-20512692 on Build GRCh38
rs765390123 Chr.20: 20512767-20512767 on Build GRCh38
rs142558157 Chr.20: 20512870-20512870 on Build GRCh38
rs758089914 Chr.20: 20512681-20512681 on Build GRCh38
rs767961937 Chr.20: 20512612-20512612 on Build GRCh38
rs371187612 Chr.20: 20512576-20512576 on Build GRCh38
rs777362874 Chr.20: 20512782-20512782 on Build GRCh38
rs1017215691 Chr.20: 20512577-20512577 on Build GRCh38
rs766860894 Chr.20: 20512755-20512755 on Build GRCh38
rs764363005 Chr.20: 20512910-20512910 on Build GRCh38
rs760697605 Chr.20: 20512591-20512591 on Build GRCh38
rs764493541 Chr.20: 20512509-20512509 on Build GRCh38
rs151107257 Chr.20: 20512551-20512551 on Build GRCh38
rs746795634 Chr.20: 20512783-20512783 on Build GRCh38
rs759196773 Chr.20: 20512603-20512603 on Build GRCh38
rs755109566 Chr.20: 20512656-20512656 on Build GRCh38
rs764974375 Chr.20: 20512605-20512605 on Build GRCh38
rs372317603 Chr.20: 20512673-20512673 on Build GRCh38
rs948926609 Chr.20: 20512536-20512536 on Build GRCh38
rs777644982 Chr.20: 20512695-20512695 on Build GRCh38
rs765163239 Chr.20: 20512642-20512643 on Build GRCh38
rs770516542 Chr.20: 20512595-20512595 on Build GRCh38
rs781250615 Chr.20: 20512803-20512803 on Build GRCh38
rs556778025 Chr.20: 20512814-20512814 on Build GRCh38
rs938254222 Chr.20: 20512687-20512687 on Build GRCh38
rs45484900 Chr.20: 20512906-20512906 on Build GRCh38
rs745354515 Chr.20: 20512735-20512735 on Build GRCh38
rs6046905 Chr.20: 20512513-20512513 on Build GRCh38
rs553334757 Chr.20: 20512753-20512753 on Build GRCh38
rs868555203 Chr.20: 20512869-20512869 on Build GRCh38
rs749458611 Chr.20: 20512573-20512573 on Build GRCh38
rs752450870 Chr.20: 20512609-20512609 on Build GRCh38
rs769066945 Chr.20: 20512822-20512822 on Build GRCh38
rs746862945 Chr.20: 20512697-20512697 on Build GRCh38
rs368793860 Chr.20: 20512586-20512586 on Build GRCh38
rs757173652 Chr.20: 20512788-20512788 on Build GRCh38
rs368237333 Chr.20: 20512557-20512557 on Build GRCh38
rs773634279 Chr.20: 20512751-20512751 on Build GRCh38
rs182386610 Chr.20: 20512770-20512770 on Build GRCh38
rs773936107 Chr.20: 20512849-20512849 on Build GRCh38
rs747247446 Chr.20: 20512601-20512601 on Build GRCh38
rs375623322 Chr.20: 20512914-20512914 on Build GRCh38
rs369405077 Chr.20: 20512879-20512879 on Build GRCh38
rs374685110 Chr.20: 20512758-20512758 on Build GRCh38
rs750850573 Chr.20: 20512622-20512622 on Build GRCh38
rs1029638285 Chr.20: 20512596-20512596 on Build GRCh38
rs534889854 Chr.20: 20512589-20512589 on Build GRCh38
rs567564269 Chr.20: 20512580-20512580 on Build GRCh38
rs780506356 Chr.20: 20512715-20512715 on Build GRCh38
rs776293226 Chr.20: 20512597-20512597 on Build GRCh38
rs774971166 Chr.20: 20512737-20512737 on Build GRCh38
rs750984425 Chr.20: 20512545-20512545 on Build GRCh38
rs770689794 Chr.20: 20512821-20512821 on Build GRCh38
rs376120579 Chr.20: 20512875-20512875 on Build GRCh38
rs756631931 Chr.20: 20512628-20512628 on Build GRCh38
rs914794395 Chr.20: 20512493-20512493 on Build GRCh38
rs990440738 Chr.20: 20512490-20512490 on Build GRCh38

View Full Product Details