Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32793171-32793677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3B
Entrez Gene ID
1789
Gene Name
DNA methyltransferase 3 beta
Gene Aliases
ICF, ICF1, M.HsaIIIB
UniGene
Hs.643024
Gene Chromosome Location
Chr.20: 32762385-32809356 on Build GRCh38
Transcripts
NM_001207055.1 NM_001207056.1 NM_006892.3 NM_175848.1 NM_175849.1 NM_175850.2 XM_011528653.2 XM_011528654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6967309 c.1081A>G p.K361E Chr.20: 32793550-32793550 on Build GRCh38
6647845 c.1126G>A p.E376K Chr.20: 32793595-32793595 on Build GRCh38
6647870 c.1087C>T p.R363C Chr.20: 32793556-32793556 on Build GRCh38
723105 c.1088G>A p.R363H Chr.20: 32793557-32793557 on Build GRCh38
6954835 c.1090C>T p.R364C Chr.20: 32793559-32793559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776703292 Chr.20: 32793549-32793549 on Build GRCh38
rs765678596 Chr.20: 32793619-32793620 on Build GRCh38
rs755617625 Chr.20: 32793585-32793585 on Build GRCh38
rs556745799 Chr.20: 32793241-32793241 on Build GRCh38
rs940256742 Chr.20: 32793468-32793468 on Build GRCh38
rs759902349 Chr.20: 32793644-32793644 on Build GRCh38
rs983107833 Chr.20: 32793459-32793459 on Build GRCh38
rs756893516 Chr.20: 32793566-32793566 on Build GRCh38
rs1015371908 Chr.20: 32793462-32793462 on Build GRCh38
rs575147395 Chr.20: 32793262-32793262 on Build GRCh38
rs774827088 Chr.20: 32793509-32793509 on Build GRCh38
rs74598203 Chr.20: 32793309-32793309 on Build GRCh38
rs780723199 Chr.20: 32793574-32793574 on Build GRCh38
rs993021696 Chr.20: 32793546-32793546 on Build GRCh38
rs533519171 Chr.20: 32793357-32793357 on Build GRCh38
rs138596278 Chr.20: 32793557-32793557 on Build GRCh38
rs745375798 Chr.20: 32793505-32793505 on Build GRCh38
rs3787143 Chr.20: 32793264-32793264 on Build GRCh38
rs1046607429 Chr.20: 32793470-32793474 on Build GRCh38
rs987363168 Chr.20: 32793387-32793387 on Build GRCh38
rs572778050 Chr.20: 32793359-32793359 on Build GRCh38
rs752480624 Chr.20: 32793485-32793485 on Build GRCh38
rs752673059 Chr.20: 32793559-32793559 on Build GRCh38
rs765374165 Chr.20: 32793646-32793646 on Build GRCh38
rs759311169 Chr.20: 32793525-32793525 on Build GRCh38
rs964925858 Chr.20: 32793641-32793641 on Build GRCh38
rs772398833 Chr.20: 32793513-32793513 on Build GRCh38
rs55674025 Chr.20: 32793312-32793312 on Build GRCh38
rs766544393 Chr.20: 32793548-32793548 on Build GRCh38
rs760904691 Chr.20: 32793542-32793542 on Build GRCh38
rs773620147 Chr.20: 32793520-32793520 on Build GRCh38
rs770538114 Chr.20: 32793502-32793502 on Build GRCh38
rs745594445 Chr.20: 32793583-32793583 on Build GRCh38
rs74950975 Chr.20: 32793475-32793475 on Build GRCh38
rs1010898235 Chr.20: 32793567-32793567 on Build GRCh38
rs772592643 Chr.20: 32793607-32793607 on Build GRCh38
rs538471595 Chr.20: 32793474-32793474 on Build GRCh38
rs151190875 Chr.20: 32793413-32793413 on Build GRCh38
rs771123018 Chr.20: 32793616-32793616 on Build GRCh38
rs886056614 Chr.20: 32793553-32793553 on Build GRCh38
rs930323757 Chr.20: 32793442-32793442 on Build GRCh38
rs376325876 Chr.20: 32793219-32793219 on Build GRCh38
rs565516122 Chr.20: 32793282-32793282 on Build GRCh38
rs993419 Chr.20: 32793423-32793423 on Build GRCh38
rs758250830 Chr.20: 32793489-32793489 on Build GRCh38
rs960249368 Chr.20: 32793232-32793232 on Build GRCh38
rs996909227 Chr.20: 32793360-32793360 on Build GRCh38
rs535235787 Chr.20: 32793239-32793239 on Build GRCh38
rs150368389 Chr.20: 32793610-32793610 on Build GRCh38
rs1043099229 Chr.20: 32793250-32793250 on Build GRCh38
rs777774264 Chr.20: 32793496-32793496 on Build GRCh38
rs1052205099 Chr.20: 32793265-32793265 on Build GRCh38
rs201681031 Chr.20: 32793595-32793595 on Build GRCh38
rs538181918 Chr.20: 32793415-32793415 on Build GRCh38
rs776700133 Chr.20: 32793637-32793637 on Build GRCh38
rs186499177 Chr.20: 32793433-32793433 on Build GRCh38
rs776116311 Chr.20: 32793509-32793513 on Build GRCh38
rs751331541 Chr.20: 32793564-32793564 on Build GRCh38
rs543024541 Chr.20: 32793234-32793234 on Build GRCh38
rs776060020 Chr.20: 32793646-32793646 on Build GRCh38
rs769095842 Chr.20: 32793506-32793506 on Build GRCh38
rs1003936442 Chr.20: 32793429-32793429 on Build GRCh38
rs763959693 Chr.20: 32793563-32793563 on Build GRCh38
rs566593149 Chr.20: 32793340-32793340 on Build GRCh38
rs113400552 Chr.20: 32793556-32793556 on Build GRCh38
rs966592763 Chr.20: 32793374-32793374 on Build GRCh38
rs369336236 Chr.20: 32793560-32793560 on Build GRCh38
rs762375124 Chr.20: 32793510-32793510 on Build GRCh38
rs746641139 Chr.20: 32793499-32793499 on Build GRCh38
rs1020909881 Chr.20: 32793569-32793569 on Build GRCh38
rs964544151 Chr.20: 32793416-32793416 on Build GRCh38
rs201278419 Chr.20: 32793594-32793594 on Build GRCh38
rs934673816 Chr.20: 32793634-32793634 on Build GRCh38
rs759545374 Chr.20: 32793552-32793552 on Build GRCh38

View Full Product Details