Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35649512-35650010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE1
Entrez Gene ID
8904
Gene Name
copine 1
Gene Aliases
COPN1, CPN1
UniGene
Hs.246413
Gene Chromosome Location
Chr.20: 35626031-35664956 on Build GRCh38
Transcripts
NM_001198863.1 NM_003915.5 NM_152925.2 NM_152926.2 NM_152927.2 NM_152928.2 NR_037188.1 NM_001198838.1 NM_001198840.1 NM_006047.5 NM_152838.3

Gene Symbol
RBM12
Entrez Gene ID
10137
Gene Name
RNA binding motif protein 12
Gene Aliases
HRIHFB2091, SWAN
UniGene
Hs.700188
Gene Chromosome Location
Chr.20: 35648925-35664956 on Build GRCh38
Transcripts
NM_001198863.1 NM_003915.5 NM_152925.2 NM_152926.2 NM_152927.2 NM_152928.2 NR_037188.1 NM_001198838.1 NM_001198840.1 NM_006047.5 NM_152838.3

SNP Detail

SNP ID Chromosome Location
rs887450725 Chr.20: 35649600-35649600 on Build GRCh38
rs189394073 Chr.20: 35649558-35649558 on Build GRCh38
rs1015768489 Chr.20: 35649784-35649784 on Build GRCh38
rs945275839 Chr.20: 35649724-35649724 on Build GRCh38
rs771005091 Chr.20: 35649940-35649940 on Build GRCh38
rs551881535 Chr.20: 35649770-35649770 on Build GRCh38
rs959474767 Chr.20: 35649982-35649982 on Build GRCh38
rs531641752 Chr.20: 35649598-35649598 on Build GRCh38
rs926705691 Chr.20: 35649933-35649933 on Build GRCh38
rs972925865 Chr.20: 35649924-35649924 on Build GRCh38
rs1013407173 Chr.20: 35649707-35649707 on Build GRCh38
rs900210296 Chr.20: 35649760-35649760 on Build GRCh38
rs929038706 Chr.20: 35649960-35649960 on Build GRCh38
rs1048807535 Chr.20: 35649973-35649973 on Build GRCh38
rs758801264 Chr.20: 35649829-35649830 on Build GRCh38
rs561213997 Chr.20: 35649586-35649586 on Build GRCh38
rs954046416 Chr.20: 35649836-35649836 on Build GRCh38
rs753306409 Chr.20: 35649773-35649776 on Build GRCh38
rs748109549 Chr.20: 35649729-35649729 on Build GRCh38
rs944201257 Chr.20: 35649536-35649536 on Build GRCh38
rs566939212 Chr.20: 35649780-35649780 on Build GRCh38
rs912805502 Chr.20: 35649873-35649873 on Build GRCh38
rs182191544 Chr.20: 35649668-35649668 on Build GRCh38
rs982094776 Chr.20: 35649878-35649878 on Build GRCh38
rs539523237 Chr.20: 35649671-35649671 on Build GRCh38
rs952989207 Chr.20: 35649779-35649779 on Build GRCh38
rs1033573335 Chr.20: 35649661-35649661 on Build GRCh38
rs944324713 Chr.20: 35649874-35649874 on Build GRCh38
rs534442137 Chr.20: 35649886-35649886 on Build GRCh38
rs929088592 Chr.20: 35649572-35649572 on Build GRCh38
rs979927074 Chr.20: 35649792-35649792 on Build GRCh38
rs1037637139 Chr.20: 35649541-35649541 on Build GRCh38
rs997431664 Chr.20: 35649760-35649760 on Build GRCh38
rs1039213913 Chr.20: 35649758-35649759 on Build GRCh38
rs768706787 Chr.20: 35649663-35649663 on Build GRCh38
rs749501987 Chr.20: 35649565-35649565 on Build GRCh38
rs926703539 Chr.20: 35649631-35649631 on Build GRCh38
rs890114573 Chr.20: 35649756-35649756 on Build GRCh38
rs1008539409 Chr.20: 35649757-35649757 on Build GRCh38
rs1004407198 Chr.20: 35649784-35649784 on Build GRCh38
rs1032316694 Chr.20: 35649825-35649825 on Build GRCh38
rs371378890 Chr.20: 35649868-35649868 on Build GRCh38
rs918755074 Chr.20: 35649949-35649949 on Build GRCh38
rs577562372 Chr.20: 35649710-35649710 on Build GRCh38
rs550954231 Chr.20: 35649638-35649638 on Build GRCh38

View Full Product Details