Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36997697-36998203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBL1
Entrez Gene ID
5933
Gene Name
RB transcriptional corepressor like 1
Gene Aliases
CP107, PRB1, p107
UniGene
Hs.207745
Gene Chromosome Location
Chr.20: 36996349-37096215 on Build GRCh38
Transcripts
NM_001323281.1 NM_001323282.1 NM_002895.4 XM_006723842.2 XM_011528955.2 XM_017027992.1

SNP Detail

SNP ID Chromosome Location
rs906248995 Chr.20: 36997922-36997922 on Build GRCh38
rs1017648112 Chr.20: 36997917-36997917 on Build GRCh38
rs945622803 Chr.20: 36998083-36998083 on Build GRCh38
rs939537430 Chr.20: 36997843-36997843 on Build GRCh38
rs555784770 Chr.20: 36997793-36997793 on Build GRCh38
rs1011169060 Chr.20: 36997957-36997957 on Build GRCh38
rs186206858 Chr.20: 36997939-36997939 on Build GRCh38
rs772987318 Chr.20: 36997744-36997744 on Build GRCh38
rs951411078 Chr.20: 36997737-36997737 on Build GRCh38
rs947725759 Chr.20: 36997905-36997905 on Build GRCh38
rs6124523 Chr.20: 36998167-36998167 on Build GRCh38
rs980477065 Chr.20: 36998014-36998014 on Build GRCh38
rs983123879 Chr.20: 36998001-36998001 on Build GRCh38
rs181045011 Chr.20: 36997921-36997921 on Build GRCh38
rs908035297 Chr.20: 36998137-36998137 on Build GRCh38
rs1044849999 Chr.20: 36997798-36997798 on Build GRCh38
rs1046089184 Chr.20: 36998170-36998170 on Build GRCh38
rs977038682 Chr.20: 36997721-36997721 on Build GRCh38
rs1043712199 Chr.20: 36997889-36997889 on Build GRCh38
rs75998916 Chr.20: 36998023-36998023 on Build GRCh38
rs775871607 Chr.20: 36997980-36997980 on Build GRCh38
rs1139418 Chr.20: 36998090-36998090 on Build GRCh38
rs1007514813 Chr.20: 36998121-36998121 on Build GRCh38
rs189444997 Chr.20: 36997910-36997910 on Build GRCh38
rs916802419 Chr.20: 36998160-36998160 on Build GRCh38
rs903827049 Chr.20: 36998107-36998107 on Build GRCh38
rs368333594 Chr.20: 36998006-36998006 on Build GRCh38
rs116162813 Chr.20: 36997847-36997847 on Build GRCh38
rs371786602 Chr.20: 36998013-36998013 on Build GRCh38
rs926744137 Chr.20: 36997821-36997821 on Build GRCh38
rs914078939 Chr.20: 36998058-36998058 on Build GRCh38
rs571154571 Chr.20: 36998052-36998052 on Build GRCh38
rs922544054 Chr.20: 36997722-36997722 on Build GRCh38
rs1018166578 Chr.20: 36998005-36998005 on Build GRCh38
rs542878377 Chr.20: 36997949-36997949 on Build GRCh38
rs909574232 Chr.20: 36997749-36997753 on Build GRCh38
rs974000252 Chr.20: 36998154-36998154 on Build GRCh38
rs538211067 Chr.20: 36997864-36997864 on Build GRCh38
rs983522639 Chr.20: 36998105-36998105 on Build GRCh38
rs113618070 Chr.20: 36998053-36998053 on Build GRCh38
rs367675870 Chr.20: 36997777-36997777 on Build GRCh38
rs1037987415 Chr.20: 36997848-36997848 on Build GRCh38
rs111447513 Chr.20: 36997950-36997950 on Build GRCh38
rs964307709 Chr.20: 36998009-36998009 on Build GRCh38
rs942185461 Chr.20: 36998143-36998143 on Build GRCh38
rs1029475223 Chr.20: 36998175-36998175 on Build GRCh38
rs941065744 Chr.20: 36997763-36997763 on Build GRCh38
rs908088121 Chr.20: 36997806-36997806 on Build GRCh38
rs11475935 Chr.20: 36998124-36998124 on Build GRCh38

View Full Product Details