Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37365063-37365570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRC
Entrez Gene ID
6714
Gene Name
SRC proto-oncogene, non-receptor tyrosine kinase
Gene Aliases
ASV, SRC1, THC6, c-SRC, p60-Src
UniGene
Hs.195659
Gene Chromosome Location
Chr.20: 37344685-37405432 on Build GRCh38
Transcripts
NM_005417.4 NM_198291.2 XM_011529013.2 XM_017028024.1 XM_017028025.1 XM_017028026.1 XM_017028027.1

SNP Detail

SNP ID Chromosome Location
rs780644849 Chr.20: 37365158-37365158 on Build GRCh38
rs969050779 Chr.20: 37365243-37365243 on Build GRCh38
rs971529589 Chr.20: 37365219-37365219 on Build GRCh38
rs1022956795 Chr.20: 37365196-37365197 on Build GRCh38
rs112790817 Chr.20: 37365333-37365333 on Build GRCh38
rs3222151 Chr.20: 37365321-37365322 on Build GRCh38
rs899376762 Chr.20: 37365155-37365155 on Build GRCh38
rs897535599 Chr.20: 37365448-37365448 on Build GRCh38
rs988091109 Chr.20: 37365288-37365288 on Build GRCh38
rs946623864 Chr.20: 37365343-37365343 on Build GRCh38
rs1005116120 Chr.20: 37365129-37365129 on Build GRCh38
rs1034705838 Chr.20: 37365273-37365273 on Build GRCh38
rs1027910503 Chr.20: 37365157-37365157 on Build GRCh38
rs987911839 Chr.20: 37365339-37365339 on Build GRCh38
rs890726758 Chr.20: 37365111-37365111 on Build GRCh38
rs11483840 Chr.20: 37365491-37365492 on Build GRCh38
rs553666544 Chr.20: 37365479-37365479 on Build GRCh38
rs754524598 Chr.20: 37365443-37365443 on Build GRCh38
rs76783669 Chr.20: 37365152-37365152 on Build GRCh38
rs991498039 Chr.20: 37365164-37365164 on Build GRCh38
rs141695904 Chr.20: 37365320-37365325 on Build GRCh38
rs935168199 Chr.20: 37365325-37365326 on Build GRCh38
rs899301647 Chr.20: 37365517-37365517 on Build GRCh38
rs200644568 Chr.20: 37365329-37365330 on Build GRCh38
rs192970364 Chr.20: 37365430-37365430 on Build GRCh38
rs148409061 Chr.20: 37365213-37365213 on Build GRCh38
rs956593563 Chr.20: 37365282-37365282 on Build GRCh38
rs369858682 Chr.20: 37365196-37365196 on Build GRCh38
rs915172686 Chr.20: 37365341-37365341 on Build GRCh38
rs1050075882 Chr.20: 37365387-37365387 on Build GRCh38
rs1036613483 Chr.20: 37365494-37365494 on Build GRCh38
rs56153701 Chr.20: 37365366-37365371 on Build GRCh38
rs187940144 Chr.20: 37365414-37365414 on Build GRCh38
rs147422453 Chr.20: 37365093-37365093 on Build GRCh38
rs776847920 Chr.20: 37365332-37365333 on Build GRCh38
rs767888451 Chr.20: 37365089-37365089 on Build GRCh38
rs978833186 Chr.20: 37365335-37365335 on Build GRCh38
rs11697728 Chr.20: 37365098-37365098 on Build GRCh38
rs71187926 Chr.20: 37365368-37365371 on Build GRCh38
rs753467577 Chr.20: 37365412-37365412 on Build GRCh38
rs748421966 Chr.20: 37365332-37365340 on Build GRCh38
rs940941679 Chr.20: 37365472-37365472 on Build GRCh38
rs931978309 Chr.20: 37365370-37365370 on Build GRCh38
rs201651998 Chr.20: 37365331-37365332 on Build GRCh38
rs867706437 Chr.20: 37365391-37365391 on Build GRCh38
rs113006177 Chr.20: 37365285-37365403 on Build GRCh38
rs1040537599 Chr.20: 37365431-37365431 on Build GRCh38
rs767251996 Chr.20: 37365322-37365327 on Build GRCh38
rs915074556 Chr.20: 37365329-37365329 on Build GRCh38
rs754252388 Chr.20: 37365320-37365327 on Build GRCh38
rs927853173 Chr.20: 37365247-37365248 on Build GRCh38
rs1003636732 Chr.20: 37365245-37365245 on Build GRCh38
rs922025065 Chr.20: 37365337-37365337 on Build GRCh38

View Full Product Details