Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57563474-57563984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCK1
Entrez Gene ID
5105
Gene Name
phosphoenolpyruvate carboxykinase 1
Gene Aliases
PEPCK-C, PEPCK1, PEPCKC
UniGene
Hs.1872
Gene Chromosome Location
Chr.20: 57561081-57566461 on Build GRCh38
Transcripts
NM_002591.3 XM_011528839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6021964 c.831G>T p.K277N Chr.20: 57563597-57563597 on Build GRCh38
4561124 c.474G>A p.K158K Chr.20: 57563636-57563636 on Build GRCh38
6741420 c.526G>A p.V176I Chr.20: 57563688-57563688 on Build GRCh38
4620100 c.427G>A p.G143S Chr.20: 57563589-57563589 on Build GRCh38
6501838 c.942G>A p.W314* Chr.20: 57563708-57563708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770195437 Chr.20: 57563783-57563783 on Build GRCh38
rs899220454 Chr.20: 57563791-57563791 on Build GRCh38
rs772810175 Chr.20: 57563569-57563569 on Build GRCh38
rs972733476 Chr.20: 57563521-57563521 on Build GRCh38
rs749825944 Chr.20: 57563574-57563574 on Build GRCh38
rs908435945 Chr.20: 57563833-57563833 on Build GRCh38
rs747784635 Chr.20: 57563561-57563561 on Build GRCh38
rs1014509272 Chr.20: 57563835-57563835 on Build GRCh38
rs61760966 Chr.20: 57563630-57563630 on Build GRCh38
rs1047909135 Chr.20: 57563795-57563795 on Build GRCh38
rs376167846 Chr.20: 57563758-57563758 on Build GRCh38
rs756881385 Chr.20: 57563772-57563772 on Build GRCh38
rs369202361 Chr.20: 57563589-57563589 on Build GRCh38
rs201677393 Chr.20: 57563682-57563682 on Build GRCh38
rs994103099 Chr.20: 57563794-57563794 on Build GRCh38
rs760593327 Chr.20: 57563760-57563760 on Build GRCh38
rs778812472 Chr.20: 57563536-57563546 on Build GRCh38
rs371044564 Chr.20: 57563597-57563597 on Build GRCh38
rs574265290 Chr.20: 57563613-57563613 on Build GRCh38
rs758358426 Chr.20: 57563611-57563611 on Build GRCh38
rs201952951 Chr.20: 57563684-57563684 on Build GRCh38
rs28359546 Chr.20: 57563609-57563609 on Build GRCh38
rs28359547 Chr.20: 57563639-57563639 on Build GRCh38
rs754801137 Chr.20: 57563551-57563551 on Build GRCh38
rs753460370 Chr.20: 57563771-57563771 on Build GRCh38
rs775729245 Chr.20: 57563604-57563604 on Build GRCh38
rs147943264 Chr.20: 57563720-57563720 on Build GRCh38
rs773297524 Chr.20: 57563737-57563737 on Build GRCh38
rs913734575 Chr.20: 57563945-57563945 on Build GRCh38
rs762064699 Chr.20: 57563731-57563731 on Build GRCh38
rs769795153 Chr.20: 57563732-57563732 on Build GRCh38
rs758917878 Chr.20: 57563687-57563687 on Build GRCh38
rs1023186072 Chr.20: 57563855-57563855 on Build GRCh38
rs925015662 Chr.20: 57563740-57563740 on Build GRCh38
rs767446111 Chr.20: 57563676-57563676 on Build GRCh38
rs1012516112 Chr.20: 57563578-57563578 on Build GRCh38
rs759511459 Chr.20: 57563675-57563675 on Build GRCh38
rs774915631 Chr.20: 57563582-57563582 on Build GRCh38
rs150928523 Chr.20: 57563624-57563624 on Build GRCh38
rs1038211003 Chr.20: 57563788-57563788 on Build GRCh38
rs772226489 Chr.20: 57563716-57563717 on Build GRCh38
rs200991428 Chr.20: 57563608-57563608 on Build GRCh38
rs746384406 Chr.20: 57563703-57563703 on Build GRCh38
rs781052030 Chr.20: 57563556-57563556 on Build GRCh38
rs181986215 Chr.20: 57563776-57563776 on Build GRCh38
rs539178531 Chr.20: 57563914-57563914 on Build GRCh38
rs748437548 Chr.20: 57563588-57563588 on Build GRCh38
rs369795226 Chr.20: 57563766-57563766 on Build GRCh38
rs558731216 Chr.20: 57563581-57563581 on Build GRCh38
rs61760967 Chr.20: 57563664-57563664 on Build GRCh38
rs545773547 Chr.20: 57563651-57563651 on Build GRCh38
rs1001851819 Chr.20: 57563875-57563875 on Build GRCh38
rs8192708 Chr.20: 57563565-57563565 on Build GRCh38
rs755100704 Chr.20: 57563522-57563522 on Build GRCh38
rs958127474 Chr.20: 57563744-57563744 on Build GRCh38
rs779245706 Chr.20: 57563669-57563669 on Build GRCh38
rs772509055 Chr.20: 57563709-57563709 on Build GRCh38
rs904284609 Chr.20: 57563930-57563930 on Build GRCh38
rs148165198 Chr.20: 57563757-57563757 on Build GRCh38
rs748682003 Chr.20: 57563631-57563631 on Build GRCh38
rs777624810 Chr.20: 57563685-57563685 on Build GRCh38
rs200973243 Chr.20: 57563648-57563648 on Build GRCh38
rs754779680 Chr.20: 57563614-57563614 on Build GRCh38
rs772972715 Chr.20: 57563894-57563894 on Build GRCh38
rs28359548 Chr.20: 57563819-57563819 on Build GRCh38
rs747837917 Chr.20: 57563625-57563625 on Build GRCh38
rs866379995 Chr.20: 57563708-57563708 on Build GRCh38
rs752608038 Chr.20: 57563617-57563617 on Build GRCh38
rs753855895 Chr.20: 57563610-57563610 on Build GRCh38
rs770458871 Chr.20: 57563636-57563636 on Build GRCh38
rs1004069299 Chr.20: 57563527-57563527 on Build GRCh38
rs1056884636 Chr.20: 57563858-57563858 on Build GRCh38
rs200817092 Chr.20: 57563670-57563670 on Build GRCh38
rs866799714 Chr.20: 57563691-57563692 on Build GRCh38
rs774262797 Chr.20: 57563786-57563786 on Build GRCh38
rs957797137 Chr.20: 57563919-57563919 on Build GRCh38
rs768997627 Chr.20: 57563727-57563727 on Build GRCh38
rs776767788 Chr.20: 57563728-57563728 on Build GRCh38
rs373315287 Chr.20: 57563666-57563666 on Build GRCh38
rs201186470 Chr.20: 57563691-57563691 on Build GRCh38
rs554852774 Chr.20: 57563612-57563612 on Build GRCh38
rs758216764 Chr.20: 57563518-57563518 on Build GRCh38
rs752486702 Chr.20: 57563679-57563679 on Build GRCh38
rs375471431 Chr.20: 57563777-57563777 on Build GRCh38
rs368556535 Chr.20: 57563754-57563754 on Build GRCh38
rs750388522 Chr.20: 57563517-57563517 on Build GRCh38
rs186145640 Chr.20: 57563814-57563814 on Build GRCh38
rs1026529907 Chr.20: 57563535-57563535 on Build GRCh38
rs987957677 Chr.20: 57563920-57563920 on Build GRCh38
rs746717691 Chr.20: 57563547-57563547 on Build GRCh38
rs747438321 Chr.20: 57563721-57563721 on Build GRCh38
rs778186996 Chr.20: 57563690-57563690 on Build GRCh38
rs142034709 Chr.20: 57563871-57563871 on Build GRCh38
rs11552145 Chr.20: 57563592-57563592 on Build GRCh38
rs1016825680 Chr.20: 57563507-57563507 on Build GRCh38
rs200050361 Chr.20: 57563557-57563557 on Build GRCh38
rs965544560 Chr.20: 57563787-57563787 on Build GRCh38
rs984168742 Chr.20: 57563827-57563827 on Build GRCh38
rs45603039 Chr.20: 57563688-57563688 on Build GRCh38
rs45441592 Chr.20: 57563792-57563792 on Build GRCh38
rs779909324 Chr.20: 57563544-57563544 on Build GRCh38
rs147749756 Chr.20: 57563653-57563653 on Build GRCh38
rs772452343 Chr.20: 57563647-57563647 on Build GRCh38

View Full Product Details