Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57604603-57605103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBP1
Entrez Gene ID
81030
Gene Name
Z-DNA binding protein 1
Gene Aliases
C20orf183, DAI, DLM-1, DLM1
UniGene
Hs.302123
Gene Chromosome Location
Chr.20: 57603846-57620576 on Build GRCh38
Transcripts
NM_001160417.1 NM_001160418.1 NM_030776.2 NR_136660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3423745 c.1096C>T p.R366C Chr.20: 57604767-57604767 on Build GRCh38
6840086 c.1147C>T p.Q383* Chr.20: 57604716-57604716 on Build GRCh38
4982265 c.1148A>G p.Q383R Chr.20: 57604715-57604715 on Build GRCh38
4945726 c.1201C>G p.L401V Chr.20: 57604662-57604662 on Build GRCh38
3548171 c.1136G>A p.R379Q Chr.20: 57604727-57604727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988469589 Chr.20: 57605059-57605059 on Build GRCh38
rs772052598 Chr.20: 57604750-57604750 on Build GRCh38
rs34009889 Chr.20: 57604644-57604645 on Build GRCh38
rs774600480 Chr.20: 57604736-57604736 on Build GRCh38
rs988914063 Chr.20: 57604845-57604845 on Build GRCh38
rs374040785 Chr.20: 57604636-57604636 on Build GRCh38
rs1038459965 Chr.20: 57604896-57604896 on Build GRCh38
rs752773427 Chr.20: 57604761-57604763 on Build GRCh38
rs896712748 Chr.20: 57604964-57604964 on Build GRCh38
rs56345209 Chr.20: 57604878-57604878 on Build GRCh38
rs373855096 Chr.20: 57604737-57604737 on Build GRCh38
rs367553465 Chr.20: 57604657-57604657 on Build GRCh38
rs890808684 Chr.20: 57604946-57604946 on Build GRCh38
rs528159989 Chr.20: 57605032-57605032 on Build GRCh38
rs1008273415 Chr.20: 57604966-57604966 on Build GRCh38
rs753737913 Chr.20: 57604685-57604685 on Build GRCh38
rs747303081 Chr.20: 57604888-57604888 on Build GRCh38
rs111591569 Chr.20: 57604871-57604871 on Build GRCh38
rs765287829 Chr.20: 57604758-57604758 on Build GRCh38
rs767541414 Chr.20: 57604681-57604681 on Build GRCh38
rs755402343 Chr.20: 57604647-57604648 on Build GRCh38
rs780533884 Chr.20: 57604658-57604658 on Build GRCh38
rs781590658 Chr.20: 57604648-57604648 on Build GRCh38
rs751225768 Chr.20: 57604707-57604707 on Build GRCh38
rs754394724 Chr.20: 57604639-57604647 on Build GRCh38
rs758253059 Chr.20: 57604704-57604704 on Build GRCh38
rs755572308 Chr.20: 57604789-57604789 on Build GRCh38
rs779900652 Chr.20: 57604705-57604705 on Build GRCh38
rs564312535 Chr.20: 57604835-57604835 on Build GRCh38
rs746106294 Chr.20: 57604804-57604804 on Build GRCh38
rs772398526 Chr.20: 57604807-57604807 on Build GRCh38
rs907889475 Chr.20: 57604869-57604869 on Build GRCh38
rs773728374 Chr.20: 57604747-57604748 on Build GRCh38
rs559147964 Chr.20: 57604911-57604911 on Build GRCh38
rs778140366 Chr.20: 57604728-57604728 on Build GRCh38
rs920759008 Chr.20: 57604839-57604839 on Build GRCh38
rs774547110 Chr.20: 57604677-57604677 on Build GRCh38
rs761749701 Chr.20: 57604690-57604690 on Build GRCh38
rs368440797 Chr.20: 57605044-57605044 on Build GRCh38
rs754567510 Chr.20: 57604718-57604718 on Build GRCh38
rs751287186 Chr.20: 57604764-57604764 on Build GRCh38
rs752307843 Chr.20: 57604769-57604769 on Build GRCh38
rs983486692 Chr.20: 57604859-57604859 on Build GRCh38
rs368798376 Chr.20: 57604803-57604803 on Build GRCh38
rs768258120 Chr.20: 57604640-57604640 on Build GRCh38
rs903677130 Chr.20: 57604915-57604915 on Build GRCh38
rs749769260 Chr.20: 57604802-57604802 on Build GRCh38
rs999297175 Chr.20: 57604921-57604921 on Build GRCh38
rs754610408 Chr.20: 57604766-57604766 on Build GRCh38
rs141295310 Chr.20: 57604767-57604767 on Build GRCh38
rs201746175 Chr.20: 57604939-57604939 on Build GRCh38
rs925745735 Chr.20: 57604834-57604834 on Build GRCh38
rs780705384 Chr.20: 57604722-57604722 on Build GRCh38
rs771267202 Chr.20: 57604735-57604735 on Build GRCh38
rs772156181 Chr.20: 57604852-57604852 on Build GRCh38
rs762891234 Chr.20: 57604761-57604761 on Build GRCh38
rs187668196 Chr.20: 57604851-57604851 on Build GRCh38
rs758055000 Chr.20: 57604628-57604628 on Build GRCh38
rs748618471 Chr.20: 57604727-57604727 on Build GRCh38
rs897279074 Chr.20: 57604900-57604900 on Build GRCh38
rs1056892296 Chr.20: 57604961-57604961 on Build GRCh38
rs370676541 Chr.20: 57604866-57604866 on Build GRCh38
rs762942709 Chr.20: 57604818-57604818 on Build GRCh38
rs763168557 Chr.20: 57604672-57604672 on Build GRCh38
rs747154554 Chr.20: 57604808-57604808 on Build GRCh38
rs765746568 Chr.20: 57604722-57604723 on Build GRCh38
rs539737746 Chr.20: 57604662-57604662 on Build GRCh38
rs35727707 Chr.20: 57604752-57604752 on Build GRCh38
rs955812071 Chr.20: 57605058-57605058 on Build GRCh38
rs968184047 Chr.20: 57605033-57605033 on Build GRCh38
rs551102956 Chr.20: 57605067-57605067 on Build GRCh38
rs556864954 Chr.20: 57604679-57604679 on Build GRCh38
rs778381596 Chr.20: 57604790-57604790 on Build GRCh38
rs8118279 Chr.20: 57604819-57604819 on Build GRCh38
rs371321693 Chr.20: 57604694-57604694 on Build GRCh38
rs151096623 Chr.20: 57604763-57604763 on Build GRCh38
rs773304224 Chr.20: 57604809-57604809 on Build GRCh38
rs775400566 Chr.20: 57605031-57605031 on Build GRCh38
rs911063224 Chr.20: 57604892-57604892 on Build GRCh38
rs1027806913 Chr.20: 57605048-57605048 on Build GRCh38
rs779847276 Chr.20: 57604631-57604631 on Build GRCh38
rs576667622 Chr.20: 57604693-57604693 on Build GRCh38
rs941384960 Chr.20: 57604895-57604895 on Build GRCh38
rs578006214 Chr.20: 57604846-57604846 on Build GRCh38
rs773254372 Chr.20: 57604759-57604759 on Build GRCh38
rs1032708366 Chr.20: 57605052-57605052 on Build GRCh38
rs770999420 Chr.20: 57604664-57604664 on Build GRCh38
rs148550523 Chr.20: 57604674-57604674 on Build GRCh38
rs113866900 Chr.20: 57604668-57604668 on Build GRCh38

View Full Product Details