Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58651537-58652045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STX16
Entrez Gene ID
8675
Gene Name
syntaxin 16
Gene Aliases
SYN16
UniGene
Hs.307913
Gene Chromosome Location
Chr.20: 58651253-58679526 on Build GRCh38
Transcripts
NM_001001433.2 NM_001134772.2 NM_001134773.2 NM_001204868.1 NM_003763.5 NR_037941.1 NR_037942.1 NR_037943.1 NR_037945.1

Gene Symbol
STX16-NPEPL1
Entrez Gene ID
100534593
Gene Name
STX16-NPEPL1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.654868
Gene Chromosome Location
Chr.20: 58651253-58715844 on Build GRCh38
Transcripts
NM_001001433.2 NM_001134772.2 NM_001134773.2 NM_001204868.1 NM_003763.5 NR_037941.1 NR_037942.1 NR_037943.1 NR_037945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6525993 c.13C>A p.R5S Chr.20: 58652019-58652019 on Build GRCh38
5057451 c.31_33delTTG p.L13delL Chr.20: 58652037-58652039 on Build GRCh38
6525995 c.13C>A p.R5S Chr.20: 58652019-58652019 on Build GRCh38
5057452 c.31_33delTTG p.L13delL Chr.20: 58652037-58652039 on Build GRCh38
6525994 c.13C>A p.R5S Chr.20: 58652019-58652019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995467484 Chr.20: 58651598-58651598 on Build GRCh38
rs987357656 Chr.20: 58651767-58651767 on Build GRCh38
rs574028895 Chr.20: 58651702-58651702 on Build GRCh38
rs560227312 Chr.20: 58651932-58651932 on Build GRCh38
rs903788016 Chr.20: 58651945-58651945 on Build GRCh38
rs986531648 Chr.20: 58651986-58651986 on Build GRCh38
rs769621833 Chr.20: 58651968-58651968 on Build GRCh38
rs945781959 Chr.20: 58651724-58651724 on Build GRCh38
rs956659967 Chr.20: 58651985-58651985 on Build GRCh38
rs992144361 Chr.20: 58651560-58651560 on Build GRCh38
rs1047086651 Chr.20: 58651816-58651816 on Build GRCh38
rs917914847 Chr.20: 58651562-58651562 on Build GRCh38
rs372834496 Chr.20: 58651992-58651992 on Build GRCh38
rs1030691813 Chr.20: 58651983-58651984 on Build GRCh38
rs538009070 Chr.20: 58651976-58651976 on Build GRCh38
rs979997921 Chr.20: 58651601-58651601 on Build GRCh38
rs747535057 Chr.20: 58651776-58651776 on Build GRCh38
rs536036062 Chr.20: 58651912-58651912 on Build GRCh38
rs181473720 Chr.20: 58651944-58651944 on Build GRCh38
rs979140872 Chr.20: 58651700-58651700 on Build GRCh38
rs41296205 Chr.20: 58651657-58651657 on Build GRCh38
rs931285204 Chr.20: 58651779-58651779 on Build GRCh38
rs927176679 Chr.20: 58651651-58651651 on Build GRCh38
rs896287633 Chr.20: 58651931-58651931 on Build GRCh38
rs781149056 Chr.20: 58651960-58651960 on Build GRCh38
rs533175021 Chr.20: 58651824-58651824 on Build GRCh38
rs992197128 Chr.20: 58651989-58651989 on Build GRCh38
rs544167209 Chr.20: 58651714-58651714 on Build GRCh38
rs1006522264 Chr.20: 58651810-58651810 on Build GRCh38
rs1034882694 Chr.20: 58651644-58651645 on Build GRCh38
rs759024848 Chr.20: 58651990-58651990 on Build GRCh38
rs755911760 Chr.20: 58651976-58651976 on Build GRCh38
rs760960769 Chr.20: 58651999-58651999 on Build GRCh38
rs368572667 Chr.20: 58651847-58651847 on Build GRCh38
rs199908670 Chr.20: 58652001-58652001 on Build GRCh38
rs1005109379 Chr.20: 58651617-58651617 on Build GRCh38
rs886056845 Chr.20: 58651630-58651630 on Build GRCh38
rs554739090 Chr.20: 58651616-58651617 on Build GRCh38
rs549213992 Chr.20: 58651963-58651963 on Build GRCh38
rs200958706 Chr.20: 58651969-58651969 on Build GRCh38
rs1050781581 Chr.20: 58651730-58651730 on Build GRCh38
rs368579447 Chr.20: 58651982-58651982 on Build GRCh38
rs141733765 Chr.20: 58651652-58651652 on Build GRCh38
rs779868188 Chr.20: 58651984-58651985 on Build GRCh38
rs773804241 Chr.20: 58651985-58651985 on Build GRCh38
rs891565115 Chr.20: 58651718-58651718 on Build GRCh38
rs886056844 Chr.20: 58651598-58651598 on Build GRCh38
rs1039670086 Chr.20: 58651592-58651592 on Build GRCh38
rs557592494 Chr.20: 58651586-58651586 on Build GRCh38
rs942877747 Chr.20: 58651770-58651770 on Build GRCh38
rs572941998 Chr.20: 58651635-58651635 on Build GRCh38
rs879295717 Chr.20: 58651623-58651623 on Build GRCh38
rs562759179 Chr.20: 58651821-58651821 on Build GRCh38
rs967668107 Chr.20: 58651696-58651696 on Build GRCh38
rs898466614 Chr.20: 58651595-58651595 on Build GRCh38
rs950005713 Chr.20: 58651565-58651565 on Build GRCh38

View Full Product Details