Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 61975884-61976382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF4
Entrez Gene ID
6874
Gene Name
TATA-box binding protein associated factor 4
Gene Aliases
TAF2C, TAF2C1, TAF4A, TAFII130, TAFII135
UniGene
Hs.18857
Gene Chromosome Location
Chr.20: 61974798-62065810 on Build GRCh38
Transcripts
NM_003185.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4100016 c.3136A>C p.T1046P Chr.20: 61976290-61976290 on Build GRCh38
5629399 c.3150C>T p.F1050F Chr.20: 61976276-61976276 on Build GRCh38
4100015 c.3170G>A p.R1057Q Chr.20: 61976256-61976256 on Build GRCh38
6796362 c.3155G>A p.R1052Q Chr.20: 61976271-61976271 on Build GRCh38
1615803 c.3210A>G p.E1070E Chr.20: 61976216-61976216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142316115 Chr.20: 61976157-61976157 on Build GRCh38
rs757408374 Chr.20: 61976261-61976261 on Build GRCh38
rs772833914 Chr.20: 61976166-61976166 on Build GRCh38
rs774042619 Chr.20: 61976249-61976249 on Build GRCh38
rs751488620 Chr.20: 61976124-61976124 on Build GRCh38
rs368980072 Chr.20: 61976130-61976130 on Build GRCh38
rs775555039 Chr.20: 61976331-61976331 on Build GRCh38
rs891151079 Chr.20: 61976185-61976185 on Build GRCh38
rs1001475517 Chr.20: 61976064-61976064 on Build GRCh38
rs922888801 Chr.20: 61976040-61976040 on Build GRCh38
rs757496056 Chr.20: 61976141-61976141 on Build GRCh38
rs558431670 Chr.20: 61976294-61976294 on Build GRCh38
rs765471072 Chr.20: 61976263-61976263 on Build GRCh38
rs753845586 Chr.20: 61976349-61976351 on Build GRCh38
rs199751605 Chr.20: 61975919-61975919 on Build GRCh38
rs905746061 Chr.20: 61976042-61976042 on Build GRCh38
rs758590580 Chr.20: 61976274-61976274 on Build GRCh38
rs755279757 Chr.20: 61976358-61976358 on Build GRCh38
rs200944569 Chr.20: 61975923-61975923 on Build GRCh38
rs958114566 Chr.20: 61976285-61976285 on Build GRCh38
rs941802918 Chr.20: 61976319-61976319 on Build GRCh38
rs781580363 Chr.20: 61976361-61976361 on Build GRCh38
rs766164306 Chr.20: 61976243-61976243 on Build GRCh38
rs772745944 Chr.20: 61976312-61976312 on Build GRCh38
rs151249150 Chr.20: 61976255-61976255 on Build GRCh38
rs770509580 Chr.20: 61976323-61976323 on Build GRCh38
rs781558360 Chr.20: 61976298-61976298 on Build GRCh38
rs116249897 Chr.20: 61976013-61976013 on Build GRCh38
rs766141423 Chr.20: 61976122-61976122 on Build GRCh38
rs923407966 Chr.20: 61976281-61976281 on Build GRCh38
rs138216073 Chr.20: 61976012-61976012 on Build GRCh38
rs970650199 Chr.20: 61976254-61976254 on Build GRCh38
rs753993076 Chr.20: 61976131-61976131 on Build GRCh38
rs769076698 Chr.20: 61976151-61976151 on Build GRCh38
rs923460435 Chr.20: 61975971-61975971 on Build GRCh38
rs747521909 Chr.20: 61976150-61976150 on Build GRCh38
rs750550902 Chr.20: 61976334-61976334 on Build GRCh38
rs138160587 Chr.20: 61976330-61976330 on Build GRCh38
rs769267728 Chr.20: 61976301-61976301 on Build GRCh38
rs748896266 Chr.20: 61976317-61976317 on Build GRCh38
rs977973320 Chr.20: 61975940-61975940 on Build GRCh38
rs140373717 Chr.20: 61976273-61976273 on Build GRCh38
rs377312375 Chr.20: 61976344-61976344 on Build GRCh38
rs547139639 Chr.20: 61976117-61976117 on Build GRCh38
rs1037609770 Chr.20: 61976015-61976015 on Build GRCh38
rs1024881161 Chr.20: 61975920-61975920 on Build GRCh38
rs374820056 Chr.20: 61976149-61976149 on Build GRCh38
rs748707980 Chr.20: 61976300-61976300 on Build GRCh38
rs1047763720 Chr.20: 61976356-61976356 on Build GRCh38
rs758604523 Chr.20: 61976338-61976338 on Build GRCh38
rs963787088 Chr.20: 61976328-61976328 on Build GRCh38
rs780427584 Chr.20: 61976282-61976282 on Build GRCh38
rs933032876 Chr.20: 61975985-61975985 on Build GRCh38
rs930197874 Chr.20: 61976057-61976057 on Build GRCh38
rs760744623 Chr.20: 61976333-61976333 on Build GRCh38
rs772552759 Chr.20: 61976148-61976148 on Build GRCh38
rs373526782 Chr.20: 61976258-61976258 on Build GRCh38
rs1040725556 Chr.20: 61976024-61976024 on Build GRCh38
rs755458395 Chr.20: 61976296-61976296 on Build GRCh38
rs970614183 Chr.20: 61975924-61975924 on Build GRCh38
rs755467550 Chr.20: 61976043-61976043 on Build GRCh38
rs201002302 Chr.20: 61976324-61976324 on Build GRCh38
rs1047477447 Chr.20: 61976008-61976008 on Build GRCh38
rs201466968 Chr.20: 61976142-61976142 on Build GRCh38
rs751889689 Chr.20: 61976350-61976350 on Build GRCh38
rs575922992 Chr.20: 61976259-61976259 on Build GRCh38
rs369829443 Chr.20: 61976155-61976155 on Build GRCh38
rs907657688 Chr.20: 61976313-61976313 on Build GRCh38
rs372547782 Chr.20: 61976144-61976144 on Build GRCh38
rs966720450 Chr.20: 61976139-61976139 on Build GRCh38
rs150380471 Chr.20: 61976327-61976327 on Build GRCh38
rs1035482079 Chr.20: 61976111-61976111 on Build GRCh38
rs375321865 Chr.20: 61976125-61976125 on Build GRCh38
rs867475832 Chr.20: 61975928-61975928 on Build GRCh38
rs1014246348 Chr.20: 61976179-61976179 on Build GRCh38
rs143217243 Chr.20: 61976214-61976214 on Build GRCh38
rs989654614 Chr.20: 61976006-61976006 on Build GRCh38
rs769374020 Chr.20: 61976165-61976165 on Build GRCh38

View Full Product Details