Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85252128-85252665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf52
Entrez Gene ID
148423
Gene Name
chromosome 1 open reading frame 52
Gene Aliases
gm117
UniGene
Hs.26226
Gene Chromosome Location
Chr.1: 85249953-85259672 on Build GRCh38
Transcripts
NM_198077.3 NR_024113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
912701 c.520C>T p.P174S Chr.1: 85252658-85252658 on Build GRCh38
4635982 c.517G>C p.E173Q Chr.1: 85252661-85252661 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs980571156 Chr.1: 85252186-85252186 on Build GRCh38
rs933382037 Chr.1: 85252435-85252435 on Build GRCh38
rs1015780539 Chr.1: 85252504-85252504 on Build GRCh38
rs970861618 Chr.1: 85252183-85252183 on Build GRCh38
rs759467745 Chr.1: 85252639-85252639 on Build GRCh38
rs201037183 Chr.1: 85252625-85252625 on Build GRCh38
rs185442057 Chr.1: 85252293-85252293 on Build GRCh38
rs897709254 Chr.1: 85252520-85252520 on Build GRCh38
rs938813836 Chr.1: 85252296-85252296 on Build GRCh38
rs548395297 Chr.1: 85252559-85252559 on Build GRCh38
rs563891796 Chr.1: 85252261-85252261 on Build GRCh38
rs948710502 Chr.1: 85252204-85252204 on Build GRCh38
rs375368069 Chr.1: 85252396-85252396 on Build GRCh38
rs757506049 Chr.1: 85252526-85252526 on Build GRCh38
rs1044400345 Chr.1: 85252229-85252229 on Build GRCh38
rs923313532 Chr.1: 85252402-85252402 on Build GRCh38
rs12068494 Chr.1: 85252477-85252477 on Build GRCh38
rs753851941 Chr.1: 85252452-85252452 on Build GRCh38
rs1005973389 Chr.1: 85252511-85252511 on Build GRCh38
rs531703593 Chr.1: 85252536-85252539 on Build GRCh38
rs748533452 Chr.1: 85252612-85252612 on Build GRCh38
rs990397343 Chr.1: 85252257-85252257 on Build GRCh38
rs187540078 Chr.1: 85252188-85252188 on Build GRCh38
rs1007940926 Chr.1: 85252498-85252498 on Build GRCh38
rs900782479 Chr.1: 85252354-85252354 on Build GRCh38
rs745623593 Chr.1: 85252454-85252454 on Build GRCh38
rs1050497682 Chr.1: 85252483-85252483 on Build GRCh38
rs1037507746 Chr.1: 85252513-85252513 on Build GRCh38
rs891923745 Chr.1: 85252318-85252318 on Build GRCh38
rs142716073 Chr.1: 85252446-85252449 on Build GRCh38
rs928619843 Chr.1: 85252557-85252557 on Build GRCh38
rs988887311 Chr.1: 85252567-85252567 on Build GRCh38
rs958895852 Chr.1: 85252199-85252199 on Build GRCh38
rs1028026135 Chr.1: 85252449-85252449 on Build GRCh38
rs551475734 Chr.1: 85252544-85252546 on Build GRCh38
rs779493024 Chr.1: 85252582-85252582 on Build GRCh38
rs188668548 Chr.1: 85252549-85252549 on Build GRCh38
rs193268787 Chr.1: 85252245-85252245 on Build GRCh38
rs773610794 Chr.1: 85252619-85252619 on Build GRCh38
rs928709457 Chr.1: 85252263-85252263 on Build GRCh38
rs560517935 Chr.1: 85252299-85252299 on Build GRCh38
rs138660851 Chr.1: 85252490-85252490 on Build GRCh38
rs185605239 Chr.1: 85252203-85252203 on Build GRCh38
rs562971827 Chr.1: 85252548-85252548 on Build GRCh38
rs776807563 Chr.1: 85252638-85252638 on Build GRCh38
rs752513129 Chr.1: 85252434-85252434 on Build GRCh38
rs58178990 Chr.1: 85252452-85252455 on Build GRCh38
rs771014154 Chr.1: 85252626-85252626 on Build GRCh38
rs372116401 Chr.1: 85252236-85252236 on Build GRCh38
rs914788131 Chr.1: 85252312-85252312 on Build GRCh38
rs890859778 Chr.1: 85252485-85252485 on Build GRCh38
rs1053619705 Chr.1: 85252311-85252311 on Build GRCh38
rs1011769517 Chr.1: 85252333-85252333 on Build GRCh38
rs772548947 Chr.1: 85252616-85252616 on Build GRCh38
rs577203871 Chr.1: 85252401-85252401 on Build GRCh38
rs946326671 Chr.1: 85252337-85252337 on Build GRCh38

View Full Product Details