Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29066643-29067147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT8
Entrez Gene ID
10694
Gene Name
chaperonin containing TCP1 subunit 8
Gene Aliases
C21orf112, Cctq, D21S246, PRED71
UniGene
Hs.125113
Gene Chromosome Location
Chr.21: 29056322-29073797 on Build GRCh38
Transcripts
NM_001282907.1 NM_001282908.1 NM_001282909.1 NM_006585.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2841804 c.463C>T p.R155* Chr.21: 29066990-29066990 on Build GRCh38
4916641 c.441A>G p.V147V Chr.21: 29067012-29067012 on Build GRCh38
6426613 c.390A>T p.E130D Chr.21: 29067063-29067063 on Build GRCh38
4944600 c.597T>C p.V199V Chr.21: 29066743-29066743 on Build GRCh38
1029777 c.459C>T p.N153N Chr.21: 29066994-29066994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770034295 Chr.21: 29067020-29067020 on Build GRCh38
rs766233365 Chr.21: 29066952-29066952 on Build GRCh38
rs775548415 Chr.21: 29066812-29066812 on Build GRCh38
rs780436925 Chr.21: 29066996-29066996 on Build GRCh38
rs1032322384 Chr.21: 29066815-29066815 on Build GRCh38
rs781006894 Chr.21: 29066882-29066882 on Build GRCh38
rs150658487 Chr.21: 29066989-29066989 on Build GRCh38
rs760360277 Chr.21: 29067092-29067092 on Build GRCh38
rs766328524 Chr.21: 29067049-29067049 on Build GRCh38
rs1023763395 Chr.21: 29066709-29066709 on Build GRCh38
rs778245205 Chr.21: 29066784-29066784 on Build GRCh38
rs199688382 Chr.21: 29067011-29067011 on Build GRCh38
rs759948624 Chr.21: 29066878-29066878 on Build GRCh38
rs878885914 Chr.21: 29066769-29066769 on Build GRCh38
rs773886434 Chr.21: 29066798-29066798 on Build GRCh38
rs533997184 Chr.21: 29066848-29066848 on Build GRCh38
rs764327507 Chr.21: 29066821-29066821 on Build GRCh38
rs137862783 Chr.21: 29066891-29066891 on Build GRCh38
rs200522803 Chr.21: 29066892-29066892 on Build GRCh38
rs370516107 Chr.21: 29066819-29066819 on Build GRCh38
rs765056164 Chr.21: 29066935-29066935 on Build GRCh38
rs141938387 Chr.21: 29066988-29066988 on Build GRCh38
rs778824426 Chr.21: 29066758-29066758 on Build GRCh38
rs763655303 Chr.21: 29067096-29067096 on Build GRCh38
rs751063625 Chr.21: 29066875-29066875 on Build GRCh38
rs11088108 Chr.21: 29066708-29066708 on Build GRCh38
rs1001540054 Chr.21: 29066808-29066808 on Build GRCh38
rs765864738 Chr.21: 29066870-29066870 on Build GRCh38
rs746432471 Chr.21: 29066907-29066907 on Build GRCh38
rs781209136 Chr.21: 29067123-29067123 on Build GRCh38
rs186282453 Chr.21: 29067112-29067112 on Build GRCh38
rs758746044 Chr.21: 29067106-29067106 on Build GRCh38
rs561148305 Chr.21: 29066922-29066922 on Build GRCh38
rs373606062 Chr.21: 29066865-29066865 on Build GRCh38
rs759089567 Chr.21: 29067057-29067057 on Build GRCh38
rs905420592 Chr.21: 29066736-29066736 on Build GRCh38
rs905389640 Chr.21: 29066984-29066984 on Build GRCh38
rs367740906 Chr.21: 29066960-29066960 on Build GRCh38
rs375877034 Chr.21: 29066776-29066776 on Build GRCh38
rs372271542 Chr.21: 29066998-29066998 on Build GRCh38
rs149364731 Chr.21: 29067068-29067068 on Build GRCh38
rs189051142 Chr.21: 29066979-29066979 on Build GRCh38
rs375033335 Chr.21: 29066785-29066788 on Build GRCh38
rs763994165 Chr.21: 29066728-29066728 on Build GRCh38
rs776896116 Chr.21: 29066925-29066925 on Build GRCh38
rs756835717 Chr.21: 29066936-29066936 on Build GRCh38
rs748001135 Chr.21: 29066824-29066827 on Build GRCh38
rs771674921 Chr.21: 29066872-29066872 on Build GRCh38
rs746453310 Chr.21: 29067082-29067084 on Build GRCh38
rs762671489 Chr.21: 29067046-29067046 on Build GRCh38
rs1009982399 Chr.21: 29067042-29067042 on Build GRCh38
rs952505745 Chr.21: 29067118-29067118 on Build GRCh38
rs139906177 Chr.21: 29067014-29067014 on Build GRCh38
rs754281981 Chr.21: 29066807-29066807 on Build GRCh38
rs201031013 Chr.21: 29066772-29066772 on Build GRCh38
rs751025576 Chr.21: 29066954-29066954 on Build GRCh38
rs558654266 Chr.21: 29066775-29066775 on Build GRCh38
rs778937654 Chr.21: 29066806-29066808 on Build GRCh38
rs757815887 Chr.21: 29066866-29066866 on Build GRCh38
rs760352658 Chr.21: 29066712-29066712 on Build GRCh38
rs752274110 Chr.21: 29067091-29067091 on Build GRCh38
rs774223355 Chr.21: 29067056-29067056 on Build GRCh38
rs750739645 Chr.21: 29067103-29067103 on Build GRCh38
rs753033621 Chr.21: 29066689-29066689 on Build GRCh38
rs761608154 Chr.21: 29066840-29066840 on Build GRCh38
rs779367479 Chr.21: 29066985-29066985 on Build GRCh38
rs138494282 Chr.21: 29066983-29066983 on Build GRCh38
rs78904637 Chr.21: 29066913-29066913 on Build GRCh38
rs866862449 Chr.21: 29066756-29066756 on Build GRCh38
rs774508163 Chr.21: 29066671-29066671 on Build GRCh38
rs765144024 Chr.21: 29066863-29066863 on Build GRCh38
rs749676122 Chr.21: 29066794-29066794 on Build GRCh38
rs777264296 Chr.21: 29067005-29067007 on Build GRCh38
rs753653342 Chr.21: 29066747-29066747 on Build GRCh38
rs768216110 Chr.21: 29066780-29066780 on Build GRCh38
rs751263534 Chr.21: 29066905-29066905 on Build GRCh38
rs759002697 Chr.21: 29066836-29066836 on Build GRCh38
rs142398030 Chr.21: 29066951-29066951 on Build GRCh38
rs767190165 Chr.21: 29067078-29067078 on Build GRCh38
rs145895598 Chr.21: 29066959-29066959 on Build GRCh38
rs779536062 Chr.21: 29066902-29066902 on Build GRCh38
rs767624697 Chr.21: 29066681-29066681 on Build GRCh38
rs771509816 Chr.21: 29066796-29066796 on Build GRCh38
rs772904922 Chr.21: 29066947-29066947 on Build GRCh38
rs759533265 Chr.21: 29066802-29066802 on Build GRCh38
rs200022038 Chr.21: 29067120-29067121 on Build GRCh38
rs201421987 Chr.21: 29067118-29067119 on Build GRCh38
rs988188508 Chr.21: 29066841-29066841 on Build GRCh38
rs781495571 Chr.21: 29066669-29066670 on Build GRCh38
rs775059643 Chr.21: 29066799-29066799 on Build GRCh38
rs761591751 Chr.21: 29066928-29066928 on Build GRCh38
rs749877026 Chr.21: 29066759-29066759 on Build GRCh38
rs770586530 Chr.21: 29067118-29067121 on Build GRCh38
rs753697370 Chr.21: 29066670-29066673 on Build GRCh38
rs780605170 Chr.21: 29067111-29067111 on Build GRCh38
rs140880362 Chr.21: 29066783-29066786 on Build GRCh38
rs575611934 Chr.21: 29066908-29066908 on Build GRCh38
rs983998334 Chr.21: 29067120-29067120 on Build GRCh38
rs373906785 Chr.21: 29066679-29066679 on Build GRCh38
rs771632780 Chr.21: 29067119-29067120 on Build GRCh38
rs372559927 Chr.21: 29067024-29067024 on Build GRCh38
rs775596789 Chr.21: 29066910-29066910 on Build GRCh38
rs755027741 Chr.21: 29067121-29067121 on Build GRCh38
rs757144494 Chr.21: 29066754-29066754 on Build GRCh38
rs753560529 Chr.21: 29066824-29066824 on Build GRCh38
rs765513230 Chr.21: 29066879-29066879 on Build GRCh38

View Full Product Details