Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 25375584-25376079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRP5L
Entrez Gene ID
91355
Gene Name
LDL receptor related protein 5 like
Gene Aliases
-
UniGene
Hs.634058
Gene Chromosome Location
Chr.22: 25351418-25405410 on Build GRCh38
Transcripts
NM_001135772.1 XM_006724372.3 XM_017029083.1 XM_017029084.1 XM_017029085.1 XM_017029086.1 XM_017029088.1

SNP Detail

SNP ID Chromosome Location
rs549746250 Chr.22: 25376037-25376037 on Build GRCh38
rs371918268 Chr.22: 25376024-25376024 on Build GRCh38
rs911525153 Chr.22: 25375948-25375948 on Build GRCh38
rs367712766 Chr.22: 25375925-25375925 on Build GRCh38
rs905309087 Chr.22: 25376006-25376006 on Build GRCh38
rs1010203376 Chr.22: 25375891-25375891 on Build GRCh38
rs747875303 Chr.22: 25376007-25376007 on Build GRCh38
rs544718900 Chr.22: 25375915-25375915 on Build GRCh38
rs561076965 Chr.22: 25375957-25375957 on Build GRCh38
rs1003496551 Chr.22: 25375671-25375671 on Build GRCh38
rs373638117 Chr.22: 25375924-25375924 on Build GRCh38
rs1035267924 Chr.22: 25375688-25375688 on Build GRCh38
rs527505201 Chr.22: 25375902-25375902 on Build GRCh38
rs531613404 Chr.22: 25376020-25376020 on Build GRCh38
rs186278396 Chr.22: 25375705-25375705 on Build GRCh38
rs953127492 Chr.22: 25376049-25376049 on Build GRCh38
rs571485516 Chr.22: 25376057-25376057 on Build GRCh38
rs911843028 Chr.22: 25375748-25375748 on Build GRCh38
rs1044861885 Chr.22: 25376004-25376004 on Build GRCh38
rs554971344 Chr.22: 25375793-25375793 on Build GRCh38
rs867115655 Chr.22: 25375769-25375769 on Build GRCh38
rs982708628 Chr.22: 25375983-25375994 on Build GRCh38
rs147731767 Chr.22: 25375693-25375694 on Build GRCh38
rs904099680 Chr.22: 25376031-25376031 on Build GRCh38
rs1056479003 Chr.22: 25376022-25376022 on Build GRCh38
rs1034964114 Chr.22: 25375829-25375829 on Build GRCh38
rs577352693 Chr.22: 25375906-25375906 on Build GRCh38
rs930510817 Chr.22: 25375984-25375984 on Build GRCh38
rs988348973 Chr.22: 25375699-25375699 on Build GRCh38
rs572492084 Chr.22: 25375751-25375751 on Build GRCh38
rs978475427 Chr.22: 25375728-25375728 on Build GRCh38
rs968507964 Chr.22: 25375709-25375709 on Build GRCh38
rs1003528030 Chr.22: 25375825-25375825 on Build GRCh38
rs887238705 Chr.22: 25375979-25375979 on Build GRCh38
rs377135093 Chr.22: 25375944-25375944 on Build GRCh38
rs898964633 Chr.22: 25375775-25375775 on Build GRCh38
rs969321029 Chr.22: 25375640-25375640 on Build GRCh38
rs997047286 Chr.22: 25376034-25376034 on Build GRCh38
rs923004006 Chr.22: 25375745-25375745 on Build GRCh38
rs934191512 Chr.22: 25375749-25375749 on Build GRCh38
rs180789440 Chr.22: 25375633-25375633 on Build GRCh38
rs921602334 Chr.22: 25375734-25375734 on Build GRCh38
rs1036655172 Chr.22: 25375967-25375967 on Build GRCh38
rs1023011240 Chr.22: 25376030-25376031 on Build GRCh38
rs574018484 Chr.22: 25375867-25375867 on Build GRCh38
rs905278130 Chr.22: 25375806-25375806 on Build GRCh38
rs554276972 Chr.22: 25375737-25375737 on Build GRCh38
rs116919551 Chr.22: 25375792-25375792 on Build GRCh38
rs543703009 Chr.22: 25375879-25375879 on Build GRCh38
rs1051421811 Chr.22: 25375743-25375743 on Build GRCh38
rs1040127778 Chr.22: 25375742-25375742 on Build GRCh38
rs1045164926 Chr.22: 25375795-25375795 on Build GRCh38
rs1047217967 Chr.22: 25375754-25375754 on Build GRCh38
rs968585513 Chr.22: 25375904-25375904 on Build GRCh38

View Full Product Details