Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89382581-89383080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP6
Entrez Gene ID
163351
Gene Name
guanylate binding protein family member 6
Gene Aliases
-
UniGene
Hs.254338
Gene Chromosome Location
Chr.1: 89363877-89388160 on Build GRCh38
Transcripts
NM_001320257.1 NM_198460.2 XM_011540835.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3725504 c.1263C>A p.I421I Chr.1: 89382774-89382774 on Build GRCh38
4686561 c.1293G>A p.G431G Chr.1: 89382804-89382804 on Build GRCh38
6127911 c.1274G>T p.S425I Chr.1: 89382785-89382785 on Build GRCh38
4010460 c.1229T>G p.L410R Chr.1: 89382740-89382740 on Build GRCh38
6127912 c.1261A>G p.I421V Chr.1: 89382772-89382772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777194066 Chr.1: 89382659-89382659 on Build GRCh38
rs201186609 Chr.1: 89382814-89382814 on Build GRCh38
rs770161192 Chr.1: 89382871-89382871 on Build GRCh38
rs776719392 Chr.1: 89382717-89382722 on Build GRCh38
rs747259408 Chr.1: 89382758-89382758 on Build GRCh38
rs777752713 Chr.1: 89382847-89382847 on Build GRCh38
rs868235498 Chr.1: 89382803-89382803 on Build GRCh38
rs916556528 Chr.1: 89382878-89382878 on Build GRCh38
rs746169173 Chr.1: 89382861-89382861 on Build GRCh38
rs199680832 Chr.1: 89382882-89382882 on Build GRCh38
rs749862782 Chr.1: 89382876-89382876 on Build GRCh38
rs760532126 Chr.1: 89382917-89382917 on Build GRCh38
rs761408621 Chr.1: 89382901-89382901 on Build GRCh38
rs767281208 Chr.1: 89382706-89382706 on Build GRCh38
rs763889875 Chr.1: 89382708-89382709 on Build GRCh38
rs149263534 Chr.1: 89382875-89382875 on Build GRCh38
rs762395528 Chr.1: 89382795-89382795 on Build GRCh38
rs746249258 Chr.1: 89382779-89382779 on Build GRCh38
rs113532469 Chr.1: 89382793-89382793 on Build GRCh38
rs752988441 Chr.1: 89382884-89382884 on Build GRCh38
rs371076563 Chr.1: 89382976-89382976 on Build GRCh38
rs756170596 Chr.1: 89382723-89382723 on Build GRCh38
rs758497962 Chr.1: 89382842-89382842 on Build GRCh38
rs766360544 Chr.1: 89382838-89382838 on Build GRCh38
rs949448716 Chr.1: 89383049-89383049 on Build GRCh38
rs928081907 Chr.1: 89382836-89382836 on Build GRCh38
rs760079354 Chr.1: 89382660-89382660 on Build GRCh38
rs535198266 Chr.1: 89383024-89383025 on Build GRCh38
rs964307184 Chr.1: 89382949-89382949 on Build GRCh38
rs372567088 Chr.1: 89382687-89382687 on Build GRCh38
rs60606690 Chr.1: 89382786-89382786 on Build GRCh38
rs939373504 Chr.1: 89382637-89382637 on Build GRCh38
rs1040099680 Chr.1: 89382696-89382696 on Build GRCh38
rs142589232 Chr.1: 89382848-89382848 on Build GRCh38
rs376383791 Chr.1: 89382664-89382664 on Build GRCh38
rs149161305 Chr.1: 89382818-89382818 on Build GRCh38
rs555078891 Chr.1: 89382857-89382857 on Build GRCh38
rs895083249 Chr.1: 89382646-89382646 on Build GRCh38
rs750194971 Chr.1: 89382817-89382817 on Build GRCh38
rs577902812 Chr.1: 89382941-89382941 on Build GRCh38
rs1007037558 Chr.1: 89382972-89382972 on Build GRCh38
rs267598761 Chr.1: 89382864-89382864 on Build GRCh38
rs761804959 Chr.1: 89382879-89382880 on Build GRCh38
rs750184642 Chr.1: 89382662-89382662 on Build GRCh38
rs566111153 Chr.1: 89382802-89382802 on Build GRCh38
rs201707768 Chr.1: 89382896-89382896 on Build GRCh38
rs765443522 Chr.1: 89382928-89382928 on Build GRCh38
rs995129835 Chr.1: 89382993-89382993 on Build GRCh38
rs935173941 Chr.1: 89382936-89382936 on Build GRCh38
rs78680245 Chr.1: 89382734-89382734 on Build GRCh38
rs898151751 Chr.1: 89382989-89382989 on Build GRCh38
rs902400150 Chr.1: 89382928-89382928 on Build GRCh38
rs780162722 Chr.1: 89382727-89382727 on Build GRCh38
rs376124592 Chr.1: 89382952-89382952 on Build GRCh38
rs113138844 Chr.1: 89382718-89382718 on Build GRCh38
rs1006576056 Chr.1: 89382740-89382740 on Build GRCh38
rs762477460 Chr.1: 89382673-89382673 on Build GRCh38
rs761413265 Chr.1: 89382811-89382811 on Build GRCh38
rs201343031 Chr.1: 89382641-89382641 on Build GRCh38
rs946608828 Chr.1: 89382647-89382647 on Build GRCh38
rs989997917 Chr.1: 89382926-89382926 on Build GRCh38
rs748171558 Chr.1: 89382728-89382728 on Build GRCh38
rs747228403 Chr.1: 89382850-89382850 on Build GRCh38
rs998739283 Chr.1: 89382888-89382888 on Build GRCh38
rs10733091 Chr.1: 89382998-89382998 on Build GRCh38
rs1017972174 Chr.1: 89382974-89382974 on Build GRCh38
rs778040669 Chr.1: 89382749-89382749 on Build GRCh38
rs957313588 Chr.1: 89382925-89382925 on Build GRCh38
rs773312485 Chr.1: 89382620-89382620 on Build GRCh38
rs774050207 Chr.1: 89382807-89382807 on Build GRCh38
rs879129180 Chr.1: 89382798-89382798 on Build GRCh38
rs750344779 Chr.1: 89382714-89382714 on Build GRCh38
rs1014091920 Chr.1: 89382868-89382868 on Build GRCh38
rs151241468 Chr.1: 89382829-89382829 on Build GRCh38
rs531273768 Chr.1: 89382688-89382688 on Build GRCh38
rs771584497 Chr.1: 89382903-89382903 on Build GRCh38
rs138122124 Chr.1: 89382840-89382840 on Build GRCh38
rs910206142 Chr.1: 89382672-89382672 on Build GRCh38
rs201346130 Chr.1: 89382809-89382809 on Build GRCh38
rs560167346 Chr.1: 89382975-89382975 on Build GRCh38
rs1043571543 Chr.1: 89382656-89382656 on Build GRCh38
rs774935804 Chr.1: 89382669-89382669 on Build GRCh38
rs556395717 Chr.1: 89382913-89382913 on Build GRCh38
rs765323147 Chr.1: 89382615-89382615 on Build GRCh38
rs771292871 Chr.1: 89382768-89382768 on Build GRCh38
rs1050856523 Chr.1: 89382942-89382942 on Build GRCh38
rs747342519 Chr.1: 89382640-89382640 on Build GRCh38
rs571463495 Chr.1: 89382773-89382773 on Build GRCh38
rs542276146 Chr.1: 89383033-89383033 on Build GRCh38
rs909364883 Chr.1: 89382993-89382994 on Build GRCh38
rs950913959 Chr.1: 89383036-89383036 on Build GRCh38
rs763731680 Chr.1: 89382680-89382680 on Build GRCh38
rs377052539 Chr.1: 89382776-89382776 on Build GRCh38

View Full Product Details